Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23181905-23182398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCR
Entrez Gene ID
613
Gene Name
BCR, RhoGEF and GTPase activating protein
Gene Aliases
ALL, BCR1, CML, D22S11, D22S662, PHL
UniGene
Hs.517461
Gene Chromosome Location
Chr.22: 23180365-23318037 on Build GRCh38
Transcripts
NM_004327.3 NM_021574.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7168345 c.964G>C p.E322Q Chr.22: 23181924-23181924 on Build GRCh38
3309452 c.1218C>T p.G406G Chr.22: 23182178-23182178 on Build GRCh38
6873274 c.1177C>T p.R393W Chr.22: 23182137-23182137 on Build GRCh38
4439320 c.1180C>T p.H394Y Chr.22: 23182140-23182140 on Build GRCh38
3309449 c.1122G>A p.S374S Chr.22: 23182082-23182082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763492884 Chr.22: 23182113-23182113 on Build GRCh38
rs777193556 Chr.22: 23182062-23182062 on Build GRCh38
rs779931858 Chr.22: 23182136-23182136 on Build GRCh38
rs747219172 Chr.22: 23182105-23182105 on Build GRCh38
rs753694900 Chr.22: 23182137-23182137 on Build GRCh38
rs745787464 Chr.22: 23182011-23182011 on Build GRCh38
rs1006919237 Chr.22: 23182310-23182310 on Build GRCh38
rs56321828 Chr.22: 23182199-23182199 on Build GRCh38
rs568261332 Chr.22: 23182291-23182291 on Build GRCh38
rs755590125 Chr.22: 23181962-23181962 on Build GRCh38
rs1025485408 Chr.22: 23182293-23182293 on Build GRCh38
rs760238340 Chr.22: 23182269-23182269 on Build GRCh38
rs376162963 Chr.22: 23182081-23182081 on Build GRCh38
rs775817981 Chr.22: 23182109-23182109 on Build GRCh38
rs759322122 Chr.22: 23182278-23182278 on Build GRCh38
rs995844793 Chr.22: 23182245-23182245 on Build GRCh38
rs34361676 Chr.22: 23182144-23182145 on Build GRCh38
rs746120867 Chr.22: 23182154-23182154 on Build GRCh38
rs774467555 Chr.22: 23181940-23181940 on Build GRCh38
rs777281405 Chr.22: 23182112-23182113 on Build GRCh38
rs766073647 Chr.22: 23182179-23182179 on Build GRCh38
rs755855526 Chr.22: 23182131-23182131 on Build GRCh38
rs149419186 Chr.22: 23182144-23182144 on Build GRCh38
rs766826731 Chr.22: 23182051-23182051 on Build GRCh38
rs199978062 Chr.22: 23181964-23181964 on Build GRCh38
rs772764527 Chr.22: 23182267-23182267 on Build GRCh38
rs755908407 Chr.22: 23182091-23182091 on Build GRCh38
rs373071713 Chr.22: 23182071-23182071 on Build GRCh38
rs774792375 Chr.22: 23182236-23182236 on Build GRCh38
rs965438936 Chr.22: 23182160-23182160 on Build GRCh38
rs767089893 Chr.22: 23182117-23182117 on Build GRCh38
rs779799231 Chr.22: 23182095-23182095 on Build GRCh38
rs764826594 Chr.22: 23181977-23181977 on Build GRCh38
rs530146563 Chr.22: 23182115-23182115 on Build GRCh38
rs35866798 Chr.22: 23182174-23182175 on Build GRCh38
rs888677122 Chr.22: 23182305-23182305 on Build GRCh38
rs575719304 Chr.22: 23181995-23181995 on Build GRCh38
rs377165672 Chr.22: 23181943-23181943 on Build GRCh38
rs765768358 Chr.22: 23182066-23182066 on Build GRCh38
rs1028702479 Chr.22: 23182250-23182250 on Build GRCh38
rs781046421 Chr.22: 23182231-23182231 on Build GRCh38
rs760115322 Chr.22: 23182063-23182063 on Build GRCh38
rs1040101656 Chr.22: 23182317-23182317 on Build GRCh38
rs372259892 Chr.22: 23182188-23182188 on Build GRCh38
rs565490262 Chr.22: 23181978-23181978 on Build GRCh38
rs746063214 Chr.22: 23182208-23182208 on Build GRCh38
rs6003541 Chr.22: 23182279-23182279 on Build GRCh38
rs146690317 Chr.22: 23181983-23181983 on Build GRCh38
rs750090348 Chr.22: 23182120-23182120 on Build GRCh38
rs772208382 Chr.22: 23182036-23182036 on Build GRCh38
rs969485901 Chr.22: 23182301-23182301 on Build GRCh38
rs781008414 Chr.22: 23182004-23182004 on Build GRCh38
rs949578672 Chr.22: 23182029-23182029 on Build GRCh38
rs1037535852 Chr.22: 23182026-23182026 on Build GRCh38
rs901295805 Chr.22: 23182328-23182328 on Build GRCh38
rs753258409 Chr.22: 23182068-23182068 on Build GRCh38
rs760893146 Chr.22: 23181951-23181951 on Build GRCh38
rs755000316 Chr.22: 23182138-23182138 on Build GRCh38
rs760417296 Chr.22: 23182172-23182172 on Build GRCh38
rs140632061 Chr.22: 23182215-23182215 on Build GRCh38
rs759918664 Chr.22: 23182107-23182107 on Build GRCh38
rs752215466 Chr.22: 23181979-23181979 on Build GRCh38
rs773488857 Chr.22: 23181948-23181948 on Build GRCh38
rs998273294 Chr.22: 23182335-23182335 on Build GRCh38
rs138970278 Chr.22: 23182102-23182102 on Build GRCh38
rs771425636 Chr.22: 23182110-23182111 on Build GRCh38
rs767700273 Chr.22: 23181944-23181944 on Build GRCh38
rs752648171 Chr.22: 23182290-23182290 on Build GRCh38
rs143923270 Chr.22: 23182166-23182166 on Build GRCh38
rs753732580 Chr.22: 23182183-23182183 on Build GRCh38
rs558805510 Chr.22: 23181932-23181932 on Build GRCh38
rs754732943 Chr.22: 23182185-23182185 on Build GRCh38
rs757926987 Chr.22: 23181980-23181980 on Build GRCh38
rs780281584 Chr.22: 23182223-23182223 on Build GRCh38
rs746521333 Chr.22: 23182104-23182105 on Build GRCh38
rs760995586 Chr.22: 23182048-23182048 on Build GRCh38
rs199823582 Chr.22: 23182227-23182227 on Build GRCh38
rs962539286 Chr.22: 23182283-23182283 on Build GRCh38
rs766873720 Chr.22: 23181953-23181953 on Build GRCh38
rs773260605 Chr.22: 23182043-23182043 on Build GRCh38
rs781511904 Chr.22: 23182149-23182149 on Build GRCh38
rs772813429 Chr.22: 23182167-23182167 on Build GRCh38
rs150295769 Chr.22: 23182069-23182069 on Build GRCh38
rs967278261 Chr.22: 23181958-23181958 on Build GRCh38
rs769785338 Chr.22: 23182021-23182021 on Build GRCh38
rs776161479 Chr.22: 23182277-23182277 on Build GRCh38
rs759920763 Chr.22: 23182152-23182154 on Build GRCh38
rs147089762 Chr.22: 23182147-23182147 on Build GRCh38
rs775849476 Chr.22: 23182284-23182284 on Build GRCh38
rs772823202 Chr.22: 23181995-23181997 on Build GRCh38
rs776997501 Chr.22: 23182106-23182106 on Build GRCh38
rs765163453 Chr.22: 23182186-23182186 on Build GRCh38
rs138456927 Chr.22: 23182155-23182155 on Build GRCh38
rs374432951 Chr.22: 23182111-23182111 on Build GRCh38
rs749272031 Chr.22: 23182098-23182098 on Build GRCh38
rs750288785 Chr.22: 23182082-23182082 on Build GRCh38
rs772007472 Chr.22: 23182100-23182100 on Build GRCh38
rs765754155 Chr.22: 23182182-23182183 on Build GRCh38
rs560606583 Chr.22: 23182191-23182191 on Build GRCh38
rs769276337 Chr.22: 23182162-23182162 on Build GRCh38
rs375509594 Chr.22: 23182235-23182235 on Build GRCh38
rs370770316 Chr.22: 23182087-23182087 on Build GRCh38
rs781458682 Chr.22: 23182202-23182202 on Build GRCh38
rs771541146 Chr.22: 23182257-23182257 on Build GRCh38
rs763359824 Chr.22: 23182159-23182159 on Build GRCh38
rs930105176 Chr.22: 23182285-23182285 on Build GRCh38
rs962222060 Chr.22: 23182061-23182061 on Build GRCh38
rs756954804 Chr.22: 23182072-23182072 on Build GRCh38
rs140119537 Chr.22: 23181997-23181997 on Build GRCh38
rs761256537 Chr.22: 23182164-23182164 on Build GRCh38
rs887433747 Chr.22: 23182260-23182260 on Build GRCh38
rs370557490 Chr.22: 23182022-23182022 on Build GRCh38
rs762318426 Chr.22: 23182070-23182070 on Build GRCh38
rs770213414 Chr.22: 23182108-23182108 on Build GRCh38
rs754365155 Chr.22: 23181954-23181954 on Build GRCh38
rs75489603 Chr.22: 23182078-23182078 on Build GRCh38
rs763553519 Chr.22: 23182112-23182112 on Build GRCh38
rs747018681 Chr.22: 23182146-23182146 on Build GRCh38
rs952234964 Chr.22: 23181959-23181959 on Build GRCh38
rs748111546 Chr.22: 23182024-23182024 on Build GRCh38
rs34195371 Chr.22: 23182211-23182212 on Build GRCh38
rs765988278 Chr.22: 23182271-23182271 on Build GRCh38

View Full Product Details