Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23253708-23254206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCR
Entrez Gene ID
613
Gene Name
BCR, RhoGEF and GTPase activating protein
Gene Aliases
ALL, BCR1, CML, D22S11, D22S662, PHL
UniGene
Hs.517461
Gene Chromosome Location
Chr.22: 23180365-23318037 on Build GRCh38
Transcripts
NM_004327.3 NM_021574.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4751236 c.1427C>T p.A476V Chr.22: 23253946-23253946 on Build GRCh38
4978514 c.1415G>A p.G472D Chr.22: 23253934-23253934 on Build GRCh38
6343664 c.1287G>T p.S429S Chr.22: 23253806-23253806 on Build GRCh38
3309455 c.1292G>A p.G431E Chr.22: 23253811-23253811 on Build GRCh38
1327309 c.1365C>G p.Y455* Chr.22: 23253884-23253884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs60094959 Chr.22: 23253929-23253929 on Build GRCh38
rs776026251 Chr.22: 23253784-23253784 on Build GRCh38
rs770360146 Chr.22: 23253753-23253754 on Build GRCh38
rs113473889 Chr.22: 23253896-23253896 on Build GRCh38
rs768132628 Chr.22: 23253800-23253800 on Build GRCh38
rs754083866 Chr.22: 23253960-23253960 on Build GRCh38
rs141440561 Chr.22: 23253844-23253844 on Build GRCh38
rs766124888 Chr.22: 23253817-23253817 on Build GRCh38
rs772886566 Chr.22: 23253881-23253881 on Build GRCh38
rs926719409 Chr.22: 23253883-23253883 on Build GRCh38
rs757426575 Chr.22: 23253833-23253833 on Build GRCh38
rs368824679 Chr.22: 23253858-23253858 on Build GRCh38
rs748687790 Chr.22: 23253920-23253920 on Build GRCh38
rs55938746 Chr.22: 23253887-23253887 on Build GRCh38
rs751622658 Chr.22: 23253828-23253828 on Build GRCh38
rs142408079 Chr.22: 23254096-23254096 on Build GRCh38
rs375606988 Chr.22: 23254176-23254176 on Build GRCh38
rs959485079 Chr.22: 23254086-23254086 on Build GRCh38
rs762847989 Chr.22: 23253950-23253950 on Build GRCh38
rs775483533 Chr.22: 23254015-23254015 on Build GRCh38
rs755299554 Chr.22: 23253962-23253962 on Build GRCh38
rs770075898 Chr.22: 23253776-23253776 on Build GRCh38
rs915556853 Chr.22: 23254062-23254062 on Build GRCh38
rs546478464 Chr.22: 23254130-23254130 on Build GRCh38
rs567775066 Chr.22: 23253956-23253956 on Build GRCh38
rs961558920 Chr.22: 23254172-23254172 on Build GRCh38
rs777203588 Chr.22: 23253927-23253927 on Build GRCh38
rs746445769 Chr.22: 23253994-23253994 on Build GRCh38
rs904005714 Chr.22: 23254045-23254045 on Build GRCh38
rs779346105 Chr.22: 23253915-23253915 on Build GRCh38
rs112376118 Chr.22: 23254180-23254180 on Build GRCh38
rs767622070 Chr.22: 23253951-23253951 on Build GRCh38
rs979386120 Chr.22: 23254100-23254100 on Build GRCh38
rs765208555 Chr.22: 23253895-23253895 on Build GRCh38
rs926152330 Chr.22: 23254109-23254109 on Build GRCh38
rs759409214 Chr.22: 23253893-23253893 on Build GRCh38
rs776435380 Chr.22: 23253939-23253939 on Build GRCh38
rs202097532 Chr.22: 23253790-23253790 on Build GRCh38
rs750536964 Chr.22: 23253952-23253952 on Build GRCh38
rs369217201 Chr.22: 23253802-23253802 on Build GRCh38
rs747217069 Chr.22: 23253753-23253753 on Build GRCh38
rs948975593 Chr.22: 23253877-23253877 on Build GRCh38
rs755020236 Chr.22: 23253821-23253821 on Build GRCh38
rs756471631 Chr.22: 23253842-23253842 on Build GRCh38
rs780334612 Chr.22: 23253843-23253843 on Build GRCh38
rs1021131773 Chr.22: 23253845-23253845 on Build GRCh38
rs992520020 Chr.22: 23254061-23254061 on Build GRCh38
rs137940685 Chr.22: 23254060-23254060 on Build GRCh38
rs753230073 Chr.22: 23253974-23253974 on Build GRCh38
rs781378436 Chr.22: 23253806-23253806 on Build GRCh38
rs1043266272 Chr.22: 23253880-23253880 on Build GRCh38
rs760291423 Chr.22: 23253885-23253885 on Build GRCh38
rs777263198 Chr.22: 23253789-23253790 on Build GRCh38
rs201526307 Chr.22: 23253980-23253980 on Build GRCh38
rs750638137 Chr.22: 23253905-23253905 on Build GRCh38
rs370367734 Chr.22: 23253862-23253862 on Build GRCh38
rs767534953 Chr.22: 23253902-23253902 on Build GRCh38
rs1021517358 Chr.22: 23254041-23254041 on Build GRCh38
rs1009835377 Chr.22: 23254035-23254035 on Build GRCh38
rs769218763 Chr.22: 23253846-23253846 on Build GRCh38
rs749245847 Chr.22: 23254017-23254017 on Build GRCh38
rs371224222 Chr.22: 23253946-23253946 on Build GRCh38
rs753789104 Chr.22: 23253820-23253820 on Build GRCh38
rs765633104 Chr.22: 23253964-23253964 on Build GRCh38
rs769690797 Chr.22: 23254011-23254011 on Build GRCh38
rs149249967 Chr.22: 23253737-23253737 on Build GRCh38
rs781275452 Chr.22: 23253755-23253755 on Build GRCh38
rs756349433 Chr.22: 23253907-23253907 on Build GRCh38
rs1033757096 Chr.22: 23254075-23254075 on Build GRCh38
rs373981091 Chr.22: 23253827-23253827 on Build GRCh38
rs778044913 Chr.22: 23253750-23253750 on Build GRCh38
rs139201536 Chr.22: 23253947-23253947 on Build GRCh38
rs954673077 Chr.22: 23253747-23253747 on Build GRCh38
rs979531098 Chr.22: 23253863-23253863 on Build GRCh38
rs780951780 Chr.22: 23254000-23254000 on Build GRCh38
rs570703729 Chr.22: 23253921-23253921 on Build GRCh38
rs56092735 Chr.22: 23253809-23253809 on Build GRCh38
rs758892323 Chr.22: 23253979-23253979 on Build GRCh38
rs772155976 Chr.22: 23254055-23254055 on Build GRCh38
rs966353095 Chr.22: 23253836-23253836 on Build GRCh38
rs577194214 Chr.22: 23253735-23253735 on Build GRCh38
rs568592995 Chr.22: 23254053-23254053 on Build GRCh38
rs760814778 Chr.22: 23253955-23253955 on Build GRCh38
rs757619294 Chr.22: 23253754-23253754 on Build GRCh38
rs113693210 Chr.22: 23253886-23253886 on Build GRCh38
rs547409270 Chr.22: 23253908-23253908 on Build GRCh38
rs771506995 Chr.22: 23253867-23253867 on Build GRCh38
rs761454338 Chr.22: 23253803-23253803 on Build GRCh38
rs182865213 Chr.22: 23253805-23253805 on Build GRCh38
rs563008753 Chr.22: 23253759-23253759 on Build GRCh38
rs760472511 Chr.22: 23253810-23253810 on Build GRCh38
rs549357360 Chr.22: 23253940-23253940 on Build GRCh38
rs745555992 Chr.22: 23254003-23254003 on Build GRCh38
rs934802691 Chr.22: 23254159-23254159 on Build GRCh38
rs536333041 Chr.22: 23254034-23254034 on Build GRCh38
rs145968600 Chr.22: 23254160-23254160 on Build GRCh38
rs377682973 Chr.22: 23253932-23253932 on Build GRCh38
rs772780462 Chr.22: 23253912-23253912 on Build GRCh38
rs755352820 Chr.22: 23253910-23253910 on Build GRCh38
rs140148416 Chr.22: 23254175-23254175 on Build GRCh38
rs577456032 Chr.22: 23254104-23254104 on Build GRCh38
rs756812114 Chr.22: 23253999-23253999 on Build GRCh38
rs761911300 Chr.22: 23253898-23253898 on Build GRCh38
rs750641994 Chr.22: 23253834-23253834 on Build GRCh38
rs746444281 Chr.22: 23254016-23254020 on Build GRCh38

View Full Product Details