Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23608315-23608835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRICH1
Entrez Gene ID
51233
Gene Name
aspartate rich 1
Gene Aliases
C22orf43, KB-208E9.1
UniGene
Hs.517466
Gene Chromosome Location
Chr.22: 23580872-23637732 on Build GRCh38
Transcripts
NM_016449.3 XM_011530206.1 XM_011530207.2 XM_011530208.2 XM_011530209.2 XM_011530210.2

SNP Detail

SNP ID Chromosome Location
rs980346583 Chr.22: 23608637-23608637 on Build GRCh38
rs1016639970 Chr.22: 23608516-23608516 on Build GRCh38
rs143075146 Chr.22: 23608491-23608491 on Build GRCh38
rs937881457 Chr.22: 23608430-23608430 on Build GRCh38
rs764382742 Chr.22: 23608780-23608780 on Build GRCh38
rs772968973 Chr.22: 23608720-23608720 on Build GRCh38
rs1057475213 Chr.22: 23608686-23608686 on Build GRCh38
rs745968240 Chr.22: 23608756-23608756 on Build GRCh38
rs755738223 Chr.22: 23608338-23608338 on Build GRCh38
rs148363479 Chr.22: 23608590-23608590 on Build GRCh38
rs1016554149 Chr.22: 23608350-23608350 on Build GRCh38
rs77396288 Chr.22: 23608378-23608378 on Build GRCh38
rs1037776165 Chr.22: 23608705-23608705 on Build GRCh38
rs938836235 Chr.22: 23608469-23608469 on Build GRCh38
rs771276675 Chr.22: 23608522-23608522 on Build GRCh38
rs987680358 Chr.22: 23608579-23608579 on Build GRCh38
rs147123796 Chr.22: 23608768-23608768 on Build GRCh38
rs753374890 Chr.22: 23608710-23608710 on Build GRCh38
rs547782443 Chr.22: 23608439-23608439 on Build GRCh38
rs778805098 Chr.22: 23608713-23608713 on Build GRCh38
rs989514265 Chr.22: 23608391-23608391 on Build GRCh38
rs923660058 Chr.22: 23608383-23608383 on Build GRCh38
rs761201642 Chr.22: 23608689-23608689 on Build GRCh38
rs981793781 Chr.22: 23608623-23608623 on Build GRCh38
rs993679186 Chr.22: 23608778-23608778 on Build GRCh38
rs539110157 Chr.22: 23608531-23608531 on Build GRCh38
rs1044622773 Chr.22: 23608447-23608447 on Build GRCh38
rs553506774 Chr.22: 23608515-23608515 on Build GRCh38
rs766127514 Chr.22: 23608517-23608518 on Build GRCh38
rs776844581 Chr.22: 23608568-23608568 on Build GRCh38
rs1027016950 Chr.22: 23608782-23608782 on Build GRCh38
rs373938982 Chr.22: 23608771-23608771 on Build GRCh38
rs544571070 Chr.22: 23608607-23608607 on Build GRCh38
rs903182675 Chr.22: 23608482-23608482 on Build GRCh38
rs1007754524 Chr.22: 23608560-23608560 on Build GRCh38
rs753280554 Chr.22: 23608360-23608360 on Build GRCh38
rs989244948 Chr.22: 23608602-23608602 on Build GRCh38
rs376018525 Chr.22: 23608789-23608789 on Build GRCh38
rs762024226 Chr.22: 23608793-23608793 on Build GRCh38
rs2298372 Chr.22: 23608459-23608459 on Build GRCh38
rs938877507 Chr.22: 23608671-23608671 on Build GRCh38
rs947681396 Chr.22: 23608442-23608442 on Build GRCh38
rs775698267 Chr.22: 23608769-23608769 on Build GRCh38
rs978452023 Chr.22: 23608707-23608707 on Build GRCh38
rs535169186 Chr.22: 23608601-23608601 on Build GRCh38
rs55871531 Chr.22: 23608634-23608634 on Build GRCh38
rs777688616 Chr.22: 23608506-23608506 on Build GRCh38
rs371103176 Chr.22: 23608696-23608696 on Build GRCh38
rs914945620 Chr.22: 23608611-23608611 on Build GRCh38
rs886495664 Chr.22: 23608501-23608501 on Build GRCh38
rs1033421095 Chr.22: 23608633-23608633 on Build GRCh38
rs935024040 Chr.22: 23608384-23608384 on Build GRCh38
rs140540619 Chr.22: 23608631-23608631 on Build GRCh38
rs1016458936 Chr.22: 23608586-23608586 on Build GRCh38
rs376290792 Chr.22: 23608715-23608715 on Build GRCh38
rs371379626 Chr.22: 23608529-23608529 on Build GRCh38
rs2298371 Chr.22: 23608395-23608395 on Build GRCh38
rs945306507 Chr.22: 23608468-23608468 on Build GRCh38
rs943445714 Chr.22: 23608761-23608761 on Build GRCh38
rs577245542 Chr.22: 23608559-23608559 on Build GRCh38
rs748165472 Chr.22: 23608739-23608739 on Build GRCh38
rs570385653 Chr.22: 23608339-23608339 on Build GRCh38
rs954543626 Chr.22: 23608574-23608574 on Build GRCh38
rs370073879 Chr.22: 23608351-23608351 on Build GRCh38
rs540185507 Chr.22: 23608669-23608669 on Build GRCh38
rs373047643 Chr.22: 23608714-23608714 on Build GRCh38
rs575706491 Chr.22: 23608595-23608595 on Build GRCh38
rs889097012 Chr.22: 23608731-23608731 on Build GRCh38

View Full Product Details