Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23616711-23617220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRICH1
Entrez Gene ID
51233
Gene Name
aspartate rich 1
Gene Aliases
C22orf43, KB-208E9.1
UniGene
Hs.517466
Gene Chromosome Location
Chr.22: 23580872-23637732 on Build GRCh38
Transcripts
NM_016449.3 XM_011530206.1 XM_011530207.2 XM_011530208.2 XM_011530209.2 XM_011530210.2 XM_017028813.1

SNP Detail

SNP ID Chromosome Location
rs1021381536 Chr.22: 23617157-23617157 on Build GRCh38
rs954209752 Chr.22: 23616965-23616965 on Build GRCh38
rs998717149 Chr.22: 23617167-23617167 on Build GRCh38
rs771863366 Chr.22: 23616859-23616859 on Build GRCh38
rs188901449 Chr.22: 23617097-23617097 on Build GRCh38
rs754158489 Chr.22: 23617150-23617150 on Build GRCh38
rs977346309 Chr.22: 23616737-23616737 on Build GRCh38
rs140585648 Chr.22: 23616790-23616790 on Build GRCh38
rs578048910 Chr.22: 23616872-23616872 on Build GRCh38
rs761826797 Chr.22: 23616892-23616892 on Build GRCh38
rs780265402 Chr.22: 23616818-23616818 on Build GRCh38
rs756114441 Chr.22: 23616803-23616826 on Build GRCh38
rs372522604 Chr.22: 23616924-23616924 on Build GRCh38
rs200150486 Chr.22: 23616896-23616896 on Build GRCh38
rs765123401 Chr.22: 23616884-23616884 on Build GRCh38
rs549135381 Chr.22: 23616854-23616854 on Build GRCh38
rs916881040 Chr.22: 23617055-23617055 on Build GRCh38
rs910122557 Chr.22: 23617026-23617026 on Build GRCh38
rs777892830 Chr.22: 23616925-23616926 on Build GRCh38
rs756263921 Chr.22: 23616907-23616907 on Build GRCh38
rs1044142593 Chr.22: 23617057-23617057 on Build GRCh38
rs769250752 Chr.22: 23616841-23616841 on Build GRCh38
rs756565765 Chr.22: 23616809-23616809 on Build GRCh38
rs778596949 Chr.22: 23616849-23616849 on Build GRCh38
rs1013978666 Chr.22: 23616800-23616800 on Build GRCh38
rs151331742 Chr.22: 23616785-23616785 on Build GRCh38
rs760242229 Chr.22: 23616866-23616866 on Build GRCh38
rs755563291 Chr.22: 23616922-23616922 on Build GRCh38
rs759514985 Chr.22: 23616881-23616881 on Build GRCh38
rs376396400 Chr.22: 23616840-23616840 on Build GRCh38
rs747611486 Chr.22: 23616857-23616857 on Build GRCh38
rs748081607 Chr.22: 23616889-23616891 on Build GRCh38
rs905732293 Chr.22: 23617090-23617090 on Build GRCh38
rs375305188 Chr.22: 23616899-23616899 on Build GRCh38
rs754111486 Chr.22: 23616920-23616920 on Build GRCh38
rs533201424 Chr.22: 23616783-23616783 on Build GRCh38
rs142379053 Chr.22: 23616758-23616758 on Build GRCh38
rs572347429 Chr.22: 23617166-23617166 on Build GRCh38
rs564042062 Chr.22: 23616964-23616964 on Build GRCh38
rs529191261 Chr.22: 23616834-23616834 on Build GRCh38
rs751664198 Chr.22: 23616885-23616885 on Build GRCh38
rs369857225 Chr.22: 23616909-23616909 on Build GRCh38
rs534696116 Chr.22: 23616905-23616905 on Build GRCh38
rs891737534 Chr.22: 23617099-23617099 on Build GRCh38
rs369513827 Chr.22: 23616861-23616861 on Build GRCh38
rs1032912299 Chr.22: 23616902-23616902 on Build GRCh38
rs369398425 Chr.22: 23616951-23616951 on Build GRCh38
rs1009997501 Chr.22: 23616945-23616945 on Build GRCh38
rs958055880 Chr.22: 23616926-23616926 on Build GRCh38
rs772073824 Chr.22: 23616921-23616921 on Build GRCh38
rs140931048 Chr.22: 23616744-23616744 on Build GRCh38
rs145209361 Chr.22: 23616994-23616994 on Build GRCh38
rs374860508 Chr.22: 23616771-23616771 on Build GRCh38
rs932913790 Chr.22: 23616747-23616747 on Build GRCh38
rs184177364 Chr.22: 23616962-23616962 on Build GRCh38
rs779503500 Chr.22: 23616923-23616923 on Build GRCh38
rs1010117592 Chr.22: 23617105-23617105 on Build GRCh38
rs535799820 Chr.22: 23617002-23617002 on Build GRCh38
rs371714689 Chr.22: 23617005-23617005 on Build GRCh38
rs772849274 Chr.22: 23616845-23616846 on Build GRCh38
rs150057438 Chr.22: 23616867-23616867 on Build GRCh38
rs940247954 Chr.22: 23617027-23617027 on Build GRCh38
rs373500135 Chr.22: 23616948-23616948 on Build GRCh38
rs1054263617 Chr.22: 23617098-23617098 on Build GRCh38
rs1037123672 Chr.22: 23617034-23617034 on Build GRCh38

View Full Product Details