Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50579884-50580382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHKB
Entrez Gene ID
1120
Gene Name
choline kinase beta
Gene Aliases
CHETK, CHKL, CK, CKB, CKEKB, EK, EKB, MDCMC
UniGene
Hs.654827
Gene Chromosome Location
Chr.22: 50578958-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Gene Symbol
CHKB-CPT1B
Entrez Gene ID
386593
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
Gene Aliases
CHKL-CPT1B
UniGene
Hs.439777
Gene Chromosome Location
Chr.22: 50568861-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361191 c.925C>G p.Q309E Chr.22: 50579976-50579976 on Build GRCh38
3066147 c.772G>A p.D258N Chr.22: 50580236-50580236 on Build GRCh38
3414219 c.871G>A p.E291K Chr.22: 50580030-50580030 on Build GRCh38
4105235 c.865A>G p.T289A Chr.22: 50580036-50580036 on Build GRCh38
5934114 c.847G>A p.E283K Chr.22: 50580054-50580054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140020889 Chr.22: 50580031-50580031 on Build GRCh38
rs778897184 Chr.22: 50580233-50580233 on Build GRCh38
rs569158018 Chr.22: 50580120-50580120 on Build GRCh38
rs765702456 Chr.22: 50580281-50580281 on Build GRCh38
rs767551132 Chr.22: 50580309-50580309 on Build GRCh38
rs139059552 Chr.22: 50579976-50579976 on Build GRCh38
rs771856963 Chr.22: 50580242-50580242 on Build GRCh38
rs769598051 Chr.22: 50580002-50580002 on Build GRCh38
rs757635030 Chr.22: 50580056-50580056 on Build GRCh38
rs779131876 Chr.22: 50579985-50579985 on Build GRCh38
rs774282967 Chr.22: 50580129-50580129 on Build GRCh38
rs368960649 Chr.22: 50580091-50580091 on Build GRCh38
rs192728067 Chr.22: 50580278-50580278 on Build GRCh38
rs143061031 Chr.22: 50579993-50579993 on Build GRCh38
rs770978511 Chr.22: 50580232-50580232 on Build GRCh38
rs1009219003 Chr.22: 50580117-50580117 on Build GRCh38
rs984131563 Chr.22: 50579925-50579925 on Build GRCh38
rs387907069 Chr.22: 50579979-50579979 on Build GRCh38
rs755024138 Chr.22: 50580225-50580225 on Build GRCh38
rs751219415 Chr.22: 50579944-50579944 on Build GRCh38
rs1028826818 Chr.22: 50580193-50580193 on Build GRCh38
rs768336190 Chr.22: 50580344-50580344 on Build GRCh38
rs780230314 Chr.22: 50580092-50580092 on Build GRCh38
rs1030395882 Chr.22: 50580257-50580257 on Build GRCh38
rs202048376 Chr.22: 50580194-50580194 on Build GRCh38
rs775547378 Chr.22: 50579996-50579996 on Build GRCh38
rs777595453 Chr.22: 50580109-50580109 on Build GRCh38
rs752801856 Chr.22: 50580313-50580313 on Build GRCh38
rs765505160 Chr.22: 50580336-50580337 on Build GRCh38
rs539357838 Chr.22: 50580347-50580347 on Build GRCh38
rs754327744 Chr.22: 50580048-50580048 on Build GRCh38
rs760833908 Chr.22: 50580058-50580058 on Build GRCh38
rs573700239 Chr.22: 50580237-50580237 on Build GRCh38
rs764474062 Chr.22: 50580045-50580045 on Build GRCh38
rs776962773 Chr.22: 50580268-50580268 on Build GRCh38
rs763332214 Chr.22: 50580288-50580288 on Build GRCh38
rs760131828 Chr.22: 50580143-50580143 on Build GRCh38
rs779897037 Chr.22: 50580338-50580338 on Build GRCh38
rs375021148 Chr.22: 50580159-50580159 on Build GRCh38
rs901172601 Chr.22: 50580151-50580151 on Build GRCh38
rs762307633 Chr.22: 50580171-50580171 on Build GRCh38
rs751732782 Chr.22: 50580302-50580302 on Build GRCh38
rs375639229 Chr.22: 50579965-50579965 on Build GRCh38
rs992511759 Chr.22: 50579931-50579931 on Build GRCh38
rs760310819 Chr.22: 50580263-50580263 on Build GRCh38
rs140514 Chr.22: 50580150-50580150 on Build GRCh38
rs776195737 Chr.22: 50580274-50580274 on Build GRCh38
rs747565422 Chr.22: 50580106-50580107 on Build GRCh38
rs916899523 Chr.22: 50579995-50579995 on Build GRCh38
rs374030717 Chr.22: 50579949-50579949 on Build GRCh38
rs745748373 Chr.22: 50580329-50580329 on Build GRCh38
rs952790208 Chr.22: 50580255-50580255 on Build GRCh38
rs771824459 Chr.22: 50580132-50580132 on Build GRCh38
rs370202172 Chr.22: 50580008-50580008 on Build GRCh38
rs549171476 Chr.22: 50579964-50579964 on Build GRCh38
rs777082533 Chr.22: 50580285-50580286 on Build GRCh38
rs751758581 Chr.22: 50580210-50580210 on Build GRCh38
rs770853530 Chr.22: 50580127-50580127 on Build GRCh38
rs759319573 Chr.22: 50580130-50580130 on Build GRCh38
rs750714699 Chr.22: 50580072-50580072 on Build GRCh38
rs368365086 Chr.22: 50580229-50580229 on Build GRCh38
rs775101863 Chr.22: 50580134-50580134 on Build GRCh38
rs550228390 Chr.22: 50580100-50580100 on Build GRCh38
rs761289961 Chr.22: 50580043-50580043 on Build GRCh38
rs766099951 Chr.22: 50579939-50579939 on Build GRCh38
rs747030221 Chr.22: 50580099-50580099 on Build GRCh38
rs531581370 Chr.22: 50580087-50580087 on Build GRCh38
rs757507890 Chr.22: 50579983-50579983 on Build GRCh38
rs778253283 Chr.22: 50579975-50579975 on Build GRCh38
rs367644242 Chr.22: 50580325-50580325 on Build GRCh38
rs199997732 Chr.22: 50580145-50580145 on Build GRCh38
rs376879066 Chr.22: 50580303-50580303 on Build GRCh38
rs773669615 Chr.22: 50580287-50580287 on Build GRCh38
rs147485527 Chr.22: 50579999-50579999 on Build GRCh38
rs771452948 Chr.22: 50580152-50580152 on Build GRCh38
rs74605815 Chr.22: 50580135-50580135 on Build GRCh38
rs762690035 Chr.22: 50580020-50580020 on Build GRCh38
rs144789175 Chr.22: 50580305-50580305 on Build GRCh38
rs752126398 Chr.22: 50579912-50579912 on Build GRCh38
rs201148042 Chr.22: 50580205-50580205 on Build GRCh38
rs776158594 Chr.22: 50580356-50580356 on Build GRCh38
rs767076133 Chr.22: 50579962-50579962 on Build GRCh38
rs766710559 Chr.22: 50580289-50580289 on Build GRCh38
rs766225706 Chr.22: 50580028-50580028 on Build GRCh38
rs899946738 Chr.22: 50580235-50580235 on Build GRCh38
rs750764003 Chr.22: 50580198-50580198 on Build GRCh38
rs759899367 Chr.22: 50580293-50580295 on Build GRCh38
rs370329322 Chr.22: 50579929-50579929 on Build GRCh38
rs1000710213 Chr.22: 50580170-50580170 on Build GRCh38
rs752131662 Chr.22: 50580042-50580042 on Build GRCh38
rs757296475 Chr.22: 50580318-50580318 on Build GRCh38
rs374544105 Chr.22: 50580030-50580030 on Build GRCh38
rs781264108 Chr.22: 50580228-50580228 on Build GRCh38
rs961307091 Chr.22: 50580285-50580285 on Build GRCh38
rs577904303 Chr.22: 50580340-50580340 on Build GRCh38
rs775024706 Chr.22: 50580244-50580244 on Build GRCh38
rs555772646 Chr.22: 50580334-50580334 on Build GRCh38

View Full Product Details