Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50582111-50582635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHKB
Entrez Gene ID
1120
Gene Name
choline kinase beta
Gene Aliases
CHETK, CHKL, CK, CKB, CKEKB, EK, EKB, MDCMC
UniGene
Hs.654827
Gene Chromosome Location
Chr.22: 50578958-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Gene Symbol
CHKB-CPT1B
Entrez Gene ID
386593
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
Gene Aliases
CHKL-CPT1B
UniGene
Hs.439777
Gene Chromosome Location
Chr.22: 50568861-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6497754 c.310C>T p.R104W Chr.22: 50582272-50582272 on Build GRCh38
7361192 c.184C>T p.R62C Chr.22: 50582598-50582598 on Build GRCh38
3668638 c.316T>G p.Y106D Chr.22: 50582266-50582266 on Build GRCh38
4842563 c.298G>A p.E100K Chr.22: 50582284-50582284 on Build GRCh38
4569063 c.148T>C p.Y50H Chr.22: 50582634-50582634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs180979987 Chr.22: 50582333-50582333 on Build GRCh38
rs548235784 Chr.22: 50582440-50582440 on Build GRCh38
rs765251030 Chr.22: 50582553-50582553 on Build GRCh38
rs772390023 Chr.22: 50582602-50582602 on Build GRCh38
rs146163970 Chr.22: 50582319-50582319 on Build GRCh38
rs864309593 Chr.22: 50582565-50582565 on Build GRCh38
rs532210178 Chr.22: 50582308-50582308 on Build GRCh38
rs779772104 Chr.22: 50582482-50582482 on Build GRCh38
rs748097708 Chr.22: 50582229-50582229 on Build GRCh38
rs1037339903 Chr.22: 50582331-50582331 on Build GRCh38
rs774840848 Chr.22: 50582587-50582587 on Build GRCh38
rs990126817 Chr.22: 50582209-50582209 on Build GRCh38
rs574732831 Chr.22: 50582167-50582167 on Build GRCh38
rs1003521884 Chr.22: 50582180-50582180 on Build GRCh38
rs147043849 Chr.22: 50582598-50582598 on Build GRCh38
rs750199673 Chr.22: 50582480-50582480 on Build GRCh38
rs963300240 Chr.22: 50582491-50582491 on Build GRCh38
rs13055012 Chr.22: 50582402-50582402 on Build GRCh38
rs757118700 Chr.22: 50582310-50582310 on Build GRCh38
rs767762723 Chr.22: 50582532-50582532 on Build GRCh38
rs773555751 Chr.22: 50582503-50582503 on Build GRCh38
rs770798469 Chr.22: 50582235-50582235 on Build GRCh38
rs774714523 Chr.22: 50582520-50582520 on Build GRCh38
rs140867913 Chr.22: 50582307-50582307 on Build GRCh38
rs1048786810 Chr.22: 50582449-50582449 on Build GRCh38
rs373168311 Chr.22: 50582155-50582155 on Build GRCh38
rs1012006163 Chr.22: 50582415-50582415 on Build GRCh38
rs546857626 Chr.22: 50582527-50582527 on Build GRCh38
rs954051094 Chr.22: 50582407-50582408 on Build GRCh38
rs370162577 Chr.22: 50582538-50582538 on Build GRCh38
rs923090455 Chr.22: 50582297-50582297 on Build GRCh38
rs1036112409 Chr.22: 50582216-50582216 on Build GRCh38
rs765518636 Chr.22: 50582282-50582282 on Build GRCh38
rs138208848 Chr.22: 50582192-50582192 on Build GRCh38
rs766423227 Chr.22: 50582293-50582293 on Build GRCh38
rs770361712 Chr.22: 50582417-50582442 on Build GRCh38
rs986782734 Chr.22: 50582489-50582489 on Build GRCh38
rs746555609 Chr.22: 50582408-50582433 on Build GRCh38
rs549810043 Chr.22: 50582473-50582473 on Build GRCh38
rs370820899 Chr.22: 50582531-50582531 on Build GRCh38
rs865952423 Chr.22: 50582299-50582299 on Build GRCh38
rs371960270 Chr.22: 50582275-50582275 on Build GRCh38
rs141533752 Chr.22: 50582586-50582586 on Build GRCh38
rs894412168 Chr.22: 50582244-50582244 on Build GRCh38
rs868464194 Chr.22: 50582272-50582272 on Build GRCh38
rs951019529 Chr.22: 50582413-50582432 on Build GRCh38
rs924682608 Chr.22: 50582149-50582149 on Build GRCh38
rs967408054 Chr.22: 50582259-50582259 on Build GRCh38
rs894738702 Chr.22: 50582582-50582582 on Build GRCh38
rs778622382 Chr.22: 50582479-50582479 on Build GRCh38
rs763126650 Chr.22: 50582465-50582465 on Build GRCh38
rs973087628 Chr.22: 50582495-50582495 on Build GRCh38
rs930373080 Chr.22: 50582447-50582447 on Build GRCh38
rs762303788 Chr.22: 50582379-50582379 on Build GRCh38
rs373182054 Chr.22: 50582378-50582378 on Build GRCh38
rs773607675 Chr.22: 50582438-50582438 on Build GRCh38
rs775814577 Chr.22: 50582606-50582606 on Build GRCh38
rs770091386 Chr.22: 50582512-50582512 on Build GRCh38
rs967459033 Chr.22: 50582443-50582443 on Build GRCh38
rs932865412 Chr.22: 50582171-50582171 on Build GRCh38
rs1022703157 Chr.22: 50582422-50582422 on Build GRCh38
rs919021435 Chr.22: 50582446-50582446 on Build GRCh38
rs534912766 Chr.22: 50582481-50582481 on Build GRCh38
rs770307691 Chr.22: 50582502-50582502 on Build GRCh38
rs774901785 Chr.22: 50582343-50582343 on Build GRCh38
rs765651340 Chr.22: 50582414-50582414 on Build GRCh38
rs552799306 Chr.22: 50582467-50582467 on Build GRCh38
rs751273046 Chr.22: 50582563-50582563 on Build GRCh38
rs746369567 Chr.22: 50582505-50582506 on Build GRCh38
rs868783642 Chr.22: 50582590-50582590 on Build GRCh38
rs771547711 Chr.22: 50582243-50582243 on Build GRCh38
rs750678988 Chr.22: 50582289-50582289 on Build GRCh38
rs770234068 Chr.22: 50582553-50582553 on Build GRCh38
rs1003166428 Chr.22: 50582387-50582387 on Build GRCh38
rs1021559166 Chr.22: 50582425-50582425 on Build GRCh38
rs745454865 Chr.22: 50582240-50582240 on Build GRCh38
rs764013683 Chr.22: 50582614-50582614 on Build GRCh38
rs376846624 Chr.22: 50582303-50582303 on Build GRCh38
rs1037129513 Chr.22: 50582435-50582436 on Build GRCh38
rs80067609 Chr.22: 50582566-50582566 on Build GRCh38
rs1025792290 Chr.22: 50582143-50582143 on Build GRCh38
rs868312203 Chr.22: 50582335-50582335 on Build GRCh38
rs769141695 Chr.22: 50582571-50582571 on Build GRCh38
rs529663693 Chr.22: 50582366-50582366 on Build GRCh38
rs778353288 Chr.22: 50582572-50582572 on Build GRCh38
rs1051288311 Chr.22: 50582175-50582175 on Build GRCh38
rs775693513 Chr.22: 50582583-50582584 on Build GRCh38
rs918910953 Chr.22: 50582504-50582504 on Build GRCh38
rs758217425 Chr.22: 50582562-50582562 on Build GRCh38
rs754605636 Chr.22: 50582564-50582564 on Build GRCh38
rs547495727 Chr.22: 50582376-50582376 on Build GRCh38
rs763394280 Chr.22: 50582511-50582511 on Build GRCh38
rs1056451549 Chr.22: 50582411-50582411 on Build GRCh38
rs894691937 Chr.22: 50582412-50582412 on Build GRCh38
rs990109854 Chr.22: 50582152-50582152 on Build GRCh38
rs957397761 Chr.22: 50582150-50582150 on Build GRCh38
rs907355659 Chr.22: 50582374-50582374 on Build GRCh38
rs746600504 Chr.22: 50582483-50582483 on Build GRCh38
rs748707816 Chr.22: 50582490-50582490 on Build GRCh38
rs86337 Chr.22: 50582239-50582239 on Build GRCh38
rs1047288115 Chr.22: 50582373-50582373 on Build GRCh38
rs977531813 Chr.22: 50582135-50582136 on Build GRCh38
rs749686701 Chr.22: 50582573-50582573 on Build GRCh38
rs564786154 Chr.22: 50582305-50582305 on Build GRCh38
rs768222640 Chr.22: 50582485-50582485 on Build GRCh38
rs757428570 Chr.22: 50582291-50582291 on Build GRCh38
rs543205547 Chr.22: 50582258-50582258 on Build GRCh38
rs760830828 Chr.22: 50582613-50582613 on Build GRCh38
rs753890036 Chr.22: 50582470-50582470 on Build GRCh38
rs560829170 Chr.22: 50582206-50582206 on Build GRCh38
rs751630140 Chr.22: 50582295-50582295 on Build GRCh38
rs570346128 Chr.22: 50582448-50582448 on Build GRCh38
rs537780707 Chr.22: 50582466-50582466 on Build GRCh38
rs747691808 Chr.22: 50582568-50582568 on Build GRCh38
rs929019284 Chr.22: 50582348-50582348 on Build GRCh38
rs369633837 Chr.22: 50582267-50582267 on Build GRCh38
rs775052422 Chr.22: 50582245-50582245 on Build GRCh38
rs761881789 Chr.22: 50582545-50582545 on Build GRCh38
rs768098104 Chr.22: 50582372-50582372 on Build GRCh38
rs756101150 Chr.22: 50582231-50582231 on Build GRCh38
rs757247185 Chr.22: 50582471-50582471 on Build GRCh38
rs547428711 Chr.22: 50582320-50582320 on Build GRCh38
rs958954555 Chr.22: 50582196-50582196 on Build GRCh38
rs767634040 Chr.22: 50582468-50582468 on Build GRCh38
rs998884192 Chr.22: 50582453-50582453 on Build GRCh38
rs750271570 Chr.22: 50582554-50582554 on Build GRCh38
rs13054973 Chr.22: 50582346-50582346 on Build GRCh38
rs746545584 Chr.22: 50582357-50582357 on Build GRCh38

View Full Product Details