Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 2571338-2571851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN4
Entrez Gene ID
152330
Gene Name
contactin 4
Gene Aliases
AXCAM, BIG-2
UniGene
Hs.298705
Gene Chromosome Location
Chr.3: 2098771-3059080 on Build GRCh38
Transcripts
NM_001206955.1 NM_175607.2 XM_006713004.3 XM_011533425.2 XM_011533426.2 XM_011533427.2 XM_011533428.2 XM_011533429.2 XM_011533430.2 XM_017005782.1 XM_017005783.1 XM_017005784.1 XM_017005785.1 XM_017005786.1 XM_017005787.1 XM_017005788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1043406 c.15G>A p.W5* Chr.3: 2571518-2571518 on Build GRCh38
2948897 c.10C>T p.P4S Chr.3: 2571513-2571513 on Build GRCh38
243761 c.18A>G p.E6E Chr.3: 2571521-2571521 on Build GRCh38
3592122 c.16G>A p.E6K Chr.3: 2571519-2571519 on Build GRCh38
1043405 c.10C>G p.P4A Chr.3: 2571513-2571513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758593322 Chr.3: 2571543-2571543 on Build GRCh38
rs757181173 Chr.3: 2571756-2571756 on Build GRCh38
rs990202663 Chr.3: 2571726-2571726 on Build GRCh38
rs148938086 Chr.3: 2571679-2571679 on Build GRCh38
rs868380627 Chr.3: 2571819-2571819 on Build GRCh38
rs760163203 Chr.3: 2571600-2571600 on Build GRCh38
rs766800101 Chr.3: 2571584-2571584 on Build GRCh38
rs1001626241 Chr.3: 2571667-2571667 on Build GRCh38
rs781184190 Chr.3: 2571806-2571806 on Build GRCh38
rs189339886 Chr.3: 2571400-2571400 on Build GRCh38
rs189603935 Chr.3: 2571655-2571655 on Build GRCh38
rs923215535 Chr.3: 2571799-2571799 on Build GRCh38
rs570307282 Chr.3: 2571487-2571487 on Build GRCh38
rs906045137 Chr.3: 2571664-2571664 on Build GRCh38
rs976106434 Chr.3: 2571780-2571780 on Build GRCh38
rs945997967 Chr.3: 2571759-2571759 on Build GRCh38
rs1024486196 Chr.3: 2571422-2571422 on Build GRCh38
rs773239321 Chr.3: 2571391-2571391 on Build GRCh38
rs112813310 Chr.3: 2571548-2571548 on Build GRCh38
rs757980763 Chr.3: 2571447-2571447 on Build GRCh38
rs745609573 Chr.3: 2571743-2571743 on Build GRCh38
rs553409555 Chr.3: 2571656-2571656 on Build GRCh38
rs749234886 Chr.3: 2571722-2571722 on Build GRCh38
rs567675764 Chr.3: 2571393-2571393 on Build GRCh38
rs780118857 Chr.3: 2571494-2571494 on Build GRCh38
rs371314379 Chr.3: 2571365-2571365 on Build GRCh38
rs768917769 Chr.3: 2571512-2571512 on Build GRCh38
rs891521004 Chr.3: 2571388-2571388 on Build GRCh38
rs377736175 Chr.3: 2571454-2571454 on Build GRCh38
rs775005762 Chr.3: 2571595-2571595 on Build GRCh38
rs180795954 Chr.3: 2571673-2571673 on Build GRCh38
rs773472183 Chr.3: 2571562-2571562 on Build GRCh38
rs372773679 Chr.3: 2571503-2571503 on Build GRCh38
rs763421080 Chr.3: 2571467-2571467 on Build GRCh38
rs572278259 Chr.3: 2571645-2571645 on Build GRCh38
rs955510932 Chr.3: 2571528-2571528 on Build GRCh38
rs538749809 Chr.3: 2571498-2571498 on Build GRCh38
rs573595744 Chr.3: 2571672-2571672 on Build GRCh38
rs778610345 Chr.3: 2571492-2571492 on Build GRCh38
rs974550411 Chr.3: 2571737-2571737 on Build GRCh38
rs957458401 Chr.3: 2571710-2571710 on Build GRCh38
rs776991426 Chr.3: 2571534-2571534 on Build GRCh38
rs780102836 Chr.3: 2571627-2571627 on Build GRCh38
rs1034496424 Chr.3: 2571688-2571688 on Build GRCh38
rs758286283 Chr.3: 2571493-2571493 on Build GRCh38
rs1005871113 Chr.3: 2571380-2571380 on Build GRCh38
rs1026044978 Chr.3: 2571506-2571506 on Build GRCh38
rs762257805 Chr.3: 2571458-2571458 on Build GRCh38
rs1008314203 Chr.3: 2571699-2571699 on Build GRCh38
rs750822964 Chr.3: 2571466-2571466 on Build GRCh38
rs369433480 Chr.3: 2571392-2571392 on Build GRCh38
rs763477802 Chr.3: 2571567-2571567 on Build GRCh38
rs184419013 Chr.3: 2571640-2571640 on Build GRCh38
rs964500567 Chr.3: 2571596-2571596 on Build GRCh38
rs931899738 Chr.3: 2571809-2571809 on Build GRCh38
rs971934352 Chr.3: 2571429-2571429 on Build GRCh38
rs751815804 Chr.3: 2571641-2571641 on Build GRCh38
rs925831324 Chr.3: 2571626-2571626 on Build GRCh38
rs756869060 Chr.3: 2571403-2571403 on Build GRCh38
rs974665151 Chr.3: 2571610-2571610 on Build GRCh38
rs763748449 Chr.3: 2571479-2571479 on Build GRCh38
rs757040256 Chr.3: 2571489-2571489 on Build GRCh38
rs531615293 Chr.3: 2571684-2571684 on Build GRCh38
rs901678946 Chr.3: 2571536-2571536 on Build GRCh38
rs73001953 Chr.3: 2571729-2571729 on Build GRCh38
rs192393093 Chr.3: 2571459-2571459 on Build GRCh38
rs115253404 Chr.3: 2571687-2571687 on Build GRCh38
rs113847954 Chr.3: 2571658-2571658 on Build GRCh38
rs143698827 Chr.3: 2571739-2571739 on Build GRCh38
rs377155230 Chr.3: 2571539-2571539 on Build GRCh38
rs199722823 Chr.3: 2571471-2571471 on Build GRCh38
rs752180644 Chr.3: 2571475-2571475 on Build GRCh38

View Full Product Details