Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 2866613-2867129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN4
Entrez Gene ID
152330
Gene Name
contactin 4
Gene Aliases
AXCAM, BIG-2
UniGene
Hs.298705
Gene Chromosome Location
Chr.3: 2098771-3059080 on Build GRCh38
Transcripts
NM_001206955.1 NM_175607.2 XM_006713004.3 XM_011533425.2 XM_011533426.2 XM_011533427.2 XM_011533428.2 XM_011533429.2 XM_011533430.2 XM_011533431.2 XM_017005782.1 XM_017005783.1 XM_017005784.1 XM_017005785.1 XM_017005786.1 XM_017005787.1 XM_017005788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1422295 c.472A>G p.I158V Chr.3: 2866769-2866769 on Build GRCh38
6510833 c.457C>A p.L153M Chr.3: 2866754-2866754 on Build GRCh38
4502547 c.615C>T p.V205V Chr.3: 2866912-2866912 on Build GRCh38
6995288 c.518delC p.S173fs*12 Chr.3: 2866815-2866815 on Build GRCh38
730368 c.491C>A p.S164Y Chr.3: 2866788-2866788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771520571 Chr.3: 2866754-2866754 on Build GRCh38
rs781645462 Chr.3: 2866726-2866726 on Build GRCh38
rs777931140 Chr.3: 2866972-2866972 on Build GRCh38
rs764831080 Chr.3: 2866647-2866647 on Build GRCh38
rs775009474 Chr.3: 2866756-2866756 on Build GRCh38
rs371379463 Chr.3: 2866930-2866930 on Build GRCh38
rs35239662 Chr.3: 2866983-2866983 on Build GRCh38
rs770034746 Chr.3: 2866830-2866830 on Build GRCh38
rs765118168 Chr.3: 2866805-2866805 on Build GRCh38
rs776112548 Chr.3: 2866862-2866862 on Build GRCh38
rs988373715 Chr.3: 2866657-2866657 on Build GRCh38
rs575538921 Chr.3: 2866658-2866658 on Build GRCh38
rs749754841 Chr.3: 2866839-2866839 on Build GRCh38
rs757962159 Chr.3: 2866737-2866737 on Build GRCh38
rs771430690 Chr.3: 2866840-2866840 on Build GRCh38
rs768026136 Chr.3: 2866996-2866996 on Build GRCh38
rs145978491 Chr.3: 2866661-2866661 on Build GRCh38
rs867681859 Chr.3: 2866857-2866857 on Build GRCh38
rs1051071741 Chr.3: 2866747-2866747 on Build GRCh38
rs529765389 Chr.3: 2866776-2866776 on Build GRCh38
rs971111582 Chr.3: 2866838-2866838 on Build GRCh38
rs929736696 Chr.3: 2866950-2866950 on Build GRCh38
rs766409827 Chr.3: 2866902-2866902 on Build GRCh38
rs755077756 Chr.3: 2866921-2866921 on Build GRCh38
rs774923326 Chr.3: 2866846-2866846 on Build GRCh38
rs200056087 Chr.3: 2866762-2866762 on Build GRCh38
rs536722862 Chr.3: 2867028-2867028 on Build GRCh38
rs6789163 Chr.3: 2867010-2867010 on Build GRCh38
rs145596001 Chr.3: 2866894-2866894 on Build GRCh38
rs946549840 Chr.3: 2866687-2866687 on Build GRCh38
rs780219181 Chr.3: 2866819-2866819 on Build GRCh38
rs1038429199 Chr.3: 2867093-2867093 on Build GRCh38
rs927231156 Chr.3: 2866893-2866893 on Build GRCh38
rs1009671362 Chr.3: 2867090-2867090 on Build GRCh38
rs372701946 Chr.3: 2866777-2866777 on Build GRCh38
rs987873278 Chr.3: 2866936-2866936 on Build GRCh38
rs975327898 Chr.3: 2866691-2866691 on Build GRCh38
rs769007358 Chr.3: 2866718-2866718 on Build GRCh38
rs148903705 Chr.3: 2867020-2867020 on Build GRCh38
rs377048968 Chr.3: 2866895-2866895 on Build GRCh38
rs556668862 Chr.3: 2867034-2867034 on Build GRCh38
rs780326374 Chr.3: 2866703-2866703 on Build GRCh38
rs369680620 Chr.3: 2866889-2866889 on Build GRCh38
rs182008043 Chr.3: 2866709-2866709 on Build GRCh38
rs35687367 Chr.3: 2866853-2866853 on Build GRCh38
rs940024960 Chr.3: 2867088-2867088 on Build GRCh38
rs748448958 Chr.3: 2866742-2866742 on Build GRCh38
rs751618459 Chr.3: 2866918-2866918 on Build GRCh38
rs559646321 Chr.3: 2866874-2866874 on Build GRCh38
rs186538995 Chr.3: 2866790-2866790 on Build GRCh38
rs528938000 Chr.3: 2866818-2866818 on Build GRCh38
rs564373471 Chr.3: 2866684-2866684 on Build GRCh38
rs975675461 Chr.3: 2867005-2867005 on Build GRCh38
rs919752805 Chr.3: 2866937-2866937 on Build GRCh38
rs770260587 Chr.3: 2866745-2866745 on Build GRCh38
rs1053964369 Chr.3: 2867076-2867076 on Build GRCh38
rs571862221 Chr.3: 2866974-2866974 on Build GRCh38
rs149729477 Chr.3: 2866828-2866828 on Build GRCh38
rs138674717 Chr.3: 2866929-2866929 on Build GRCh38
rs779384838 Chr.3: 2866977-2866977 on Build GRCh38
rs1001107269 Chr.3: 2867039-2867039 on Build GRCh38
rs757901304 Chr.3: 2867058-2867058 on Build GRCh38
rs1018920181 Chr.3: 2866802-2866802 on Build GRCh38
rs1001065119 Chr.3: 2866825-2866825 on Build GRCh38
rs890754105 Chr.3: 2866723-2866723 on Build GRCh38
rs752814497 Chr.3: 2866942-2866942 on Build GRCh38
rs867197562 Chr.3: 2866813-2866813 on Build GRCh38
rs756388039 Chr.3: 2866954-2866954 on Build GRCh38
rs940146400 Chr.3: 2866796-2866796 on Build GRCh38
rs768218638 Chr.3: 2866860-2866860 on Build GRCh38
rs6442761 Chr.3: 2866651-2866651 on Build GRCh38
rs189432684 Chr.3: 2866646-2866646 on Build GRCh38
rs201525733 Chr.3: 2866829-2866829 on Build GRCh38
rs750302381 Chr.3: 2866806-2866806 on Build GRCh38
rs778888279 Chr.3: 2866834-2866834 on Build GRCh38
rs763565870 Chr.3: 2866753-2866753 on Build GRCh38
rs200414214 Chr.3: 2866823-2866823 on Build GRCh38
rs761504879 Chr.3: 2866803-2866803 on Build GRCh38
rs377711635 Chr.3: 2867042-2867042 on Build GRCh38

View Full Product Details