Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 8627509-8628013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSUH2
Entrez Gene ID
51066
Gene Name
ssu-2 homolog (C. elegans)
Gene Aliases
C3orf32, SSU-2, fls485
UniGene
Hs.736935
Gene Chromosome Location
Chr.3: 8619400-8745035 on Build GRCh38
Transcripts
NM_001256748.1 NM_001256749.1 NM_015931.2 NR_046358.1 XM_011533774.1 XM_017006510.1 XM_017006511.1 XM_017006512.1 XM_017006513.1 XM_017006514.1 XM_017006515.1 XM_017006516.1 XM_017006517.1 XM_017006518.1 XM_017006519.1 XM_017006520.1 XM_017006521.1 XM_017006522.1 XM_017006523.1 XM_017006524.1 XM_017006525.1 XM_017006526.1 XM_017006527.1 XM_017006528.1 XM_017006529.1 XM_017006530.1 XM_017006531.1 XM_017006532.1 XM_017006533.1 XM_017006534.1 XM_017006535.1 XM_017006536.1 XM_017006537.1 XM_017006538.1 XM_017006539.1 XM_017006540.1 XM_017006541.1 XM_017006542.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5865349 c.610G>A p.G204R Chr.3: 8627762-8627762 on Build GRCh38
2827993 c.554G>A p.R185H Chr.3: 8627752-8627752 on Build GRCh38
4782189 c.674+2T>A p.? Chr.3: 8627696-8627696 on Build GRCh38
4657330 c.605G>A p.R202Q Chr.3: 8627701-8627701 on Build GRCh38
4789339 c.594G>C p.R198R Chr.3: 8627778-8627778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013993293 Chr.3: 8627797-8627797 on Build GRCh38
rs766780350 Chr.3: 8627763-8627763 on Build GRCh38
rs750734268 Chr.3: 8627713-8627713 on Build GRCh38
rs145642860 Chr.3: 8627715-8627715 on Build GRCh38
rs935560033 Chr.3: 8627978-8627978 on Build GRCh38
rs943426698 Chr.3: 8627924-8627924 on Build GRCh38
rs565569011 Chr.3: 8627811-8627811 on Build GRCh38
rs191081487 Chr.3: 8627863-8627863 on Build GRCh38
rs113481784 Chr.3: 8627673-8627673 on Build GRCh38
rs760731511 Chr.3: 8627701-8627701 on Build GRCh38
rs972653531 Chr.3: 8627613-8627613 on Build GRCh38
rs545196684 Chr.3: 8627674-8627674 on Build GRCh38
rs777431535 Chr.3: 8627683-8627683 on Build GRCh38
rs398051543 Chr.3: 8627685-8627685 on Build GRCh38
rs537633392 Chr.3: 8627533-8627533 on Build GRCh38
rs778665877 Chr.3: 8627726-8627726 on Build GRCh38
rs752000182 Chr.3: 8627765-8627765 on Build GRCh38
rs891572142 Chr.3: 8627779-8627779 on Build GRCh38
rs755740096 Chr.3: 8627676-8627676 on Build GRCh38
rs745647362 Chr.3: 8627732-8627732 on Build GRCh38
rs923390007 Chr.3: 8627930-8627930 on Build GRCh38
rs142057178 Chr.3: 8627716-8627716 on Build GRCh38
rs952768411 Chr.3: 8627883-8627883 on Build GRCh38
rs951926551 Chr.3: 8627839-8627839 on Build GRCh38
rs376748108 Chr.3: 8627795-8627795 on Build GRCh38
rs763094413 Chr.3: 8627762-8627762 on Build GRCh38
rs1533001 Chr.3: 8627675-8627675 on Build GRCh38
rs556143253 Chr.3: 8627573-8627573 on Build GRCh38
rs556008210 Chr.3: 8627882-8627882 on Build GRCh38
rs750835627 Chr.3: 8627672-8627672 on Build GRCh38
rs944809029 Chr.3: 8627881-8627881 on Build GRCh38
rs141418100 Chr.3: 8627812-8627812 on Build GRCh38
rs765085847 Chr.3: 8627730-8627731 on Build GRCh38
rs541706925 Chr.3: 8627606-8627606 on Build GRCh38
rs916912268 Chr.3: 8627604-8627604 on Build GRCh38
rs79240383 Chr.3: 8627709-8627709 on Build GRCh38
rs745723419 Chr.3: 8627695-8627695 on Build GRCh38
rs74609126 Chr.3: 8627602-8627602 on Build GRCh38
rs754891583 Chr.3: 8627834-8627834 on Build GRCh38
rs768186436 Chr.3: 8627787-8627787 on Build GRCh38
rs932009111 Chr.3: 8627920-8627920 on Build GRCh38
rs979712041 Chr.3: 8627825-8627825 on Build GRCh38
rs186261883 Chr.3: 8627607-8627607 on Build GRCh38
rs753420595 Chr.3: 8627723-8627723 on Build GRCh38
rs776051085 Chr.3: 8627697-8627697 on Build GRCh38
rs537458474 Chr.3: 8627899-8627899 on Build GRCh38
rs757515526 Chr.3: 8627656-8627656 on Build GRCh38
rs531444231 Chr.3: 8627687-8627687 on Build GRCh38
rs147863430 Chr.3: 8627810-8627810 on Build GRCh38
rs377451237 Chr.3: 8627780-8627780 on Build GRCh38
rs756835646 Chr.3: 8627724-8627724 on Build GRCh38
rs768475071 Chr.3: 8627772-8627772 on Build GRCh38
rs762005243 Chr.3: 8627747-8627747 on Build GRCh38
rs553606925 Chr.3: 8627961-8627961 on Build GRCh38
rs1031430623 Chr.3: 8627599-8627599 on Build GRCh38
rs369635093 Chr.3: 8627820-8627820 on Build GRCh38
rs778522394 Chr.3: 8627691-8627691 on Build GRCh38
rs779751625 Chr.3: 8627658-8627658 on Build GRCh38
rs980026908 Chr.3: 8627611-8627611 on Build GRCh38
rs369427525 Chr.3: 8627745-8627745 on Build GRCh38
rs777374175 Chr.3: 8627717-8627717 on Build GRCh38
rs752469476 Chr.3: 8627718-8627718 on Build GRCh38
rs769878746 Chr.3: 8627752-8627752 on Build GRCh38
rs779838642 Chr.3: 8627735-8627735 on Build GRCh38
rs933319167 Chr.3: 8627939-8627939 on Build GRCh38
rs535363974 Chr.3: 8627958-8627958 on Build GRCh38
rs6784347 Chr.3: 8627872-8627872 on Build GRCh38
rs1030804668 Chr.3: 8627696-8627696 on Build GRCh38
rs1052632175 Chr.3: 8627979-8627979 on Build GRCh38
rs770648653 Chr.3: 8627688-8627688 on Build GRCh38
rs754118506 Chr.3: 8627650-8627650 on Build GRCh38
rs768615616 Chr.3: 8627738-8627738 on Build GRCh38
rs746833566 Chr.3: 8627831-8627831 on Build GRCh38
rs756821414 Chr.3: 8627695-8627695 on Build GRCh38
rs551229165 Chr.3: 8627838-8627838 on Build GRCh38
rs183398630 Chr.3: 8627975-8627975 on Build GRCh38
rs549084219 Chr.3: 8627568-8627568 on Build GRCh38
rs776228275 Chr.3: 8627697-8627706 on Build GRCh38
rs374521608 Chr.3: 8627753-8627753 on Build GRCh38
rs355055 Chr.3: 8627929-8627929 on Build GRCh38
rs753255727 Chr.3: 8627796-8627796 on Build GRCh38
rs151201843 Chr.3: 8627702-8627702 on Build GRCh38
rs892033887 Chr.3: 8627986-8627986 on Build GRCh38
rs377685481 Chr.3: 8627734-8627734 on Build GRCh38
rs976210894 Chr.3: 8627886-8627886 on Build GRCh38
rs776697436 Chr.3: 8627706-8627706 on Build GRCh38
rs756856940 Chr.3: 8627798-8627798 on Build GRCh38
rs373708901 Chr.3: 8627708-8627708 on Build GRCh38
rs139675767 Chr.3: 8627761-8627761 on Build GRCh38
rs5846608 Chr.3: 8627682-8627682 on Build GRCh38
rs796642660 Chr.3: 8627601-8627601 on Build GRCh38
rs6784066 Chr.3: 8627635-8627635 on Build GRCh38
rs746882683 Chr.3: 8627737-8627737 on Build GRCh38

View Full Product Details