Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9705372-9705866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE9
Entrez Gene ID
151835
Gene Name
copine family member 9
Gene Aliases
-
UniGene
Hs.121335
Gene Chromosome Location
Chr.3: 9703807-9729908 on Build GRCh38
Transcripts
NM_001308388.1 NM_153635.2 XM_011533386.2 XM_011533388.2 XM_011533389.2 XM_017005747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4327993 c.290C>T p.S97F Chr.3: 9705493-9705493 on Build GRCh38
7247857 c.298-10G>C p.? Chr.3: 9705708-9705708 on Build GRCh38
7247858 c.298-10G>C p.? Chr.3: 9705708-9705708 on Build GRCh38
1669948 c.290C>T p.S97F Chr.3: 9705493-9705493 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372633475 Chr.3: 9705503-9705503 on Build GRCh38
rs73021421 Chr.3: 9705643-9705643 on Build GRCh38
rs927196324 Chr.3: 9705625-9705625 on Build GRCh38
rs199890278 Chr.3: 9705445-9705445 on Build GRCh38
rs781038710 Chr.3: 9705737-9705737 on Build GRCh38
rs375034299 Chr.3: 9705825-9705825 on Build GRCh38
rs574855653 Chr.3: 9705725-9705725 on Build GRCh38
rs566258212 Chr.3: 9705768-9705768 on Build GRCh38
rs766523332 Chr.3: 9705529-9705529 on Build GRCh38
rs761238652 Chr.3: 9705539-9705539 on Build GRCh38
rs748031680 Chr.3: 9705633-9705633 on Build GRCh38
rs773333474 Chr.3: 9705510-9705510 on Build GRCh38
rs371534251 Chr.3: 9705419-9705420 on Build GRCh38
rs78883514 Chr.3: 9705708-9705708 on Build GRCh38
rs376855412 Chr.3: 9705522-9705522 on Build GRCh38
rs768741640 Chr.3: 9705458-9705458 on Build GRCh38
rs913024936 Chr.3: 9705518-9705518 on Build GRCh38
rs1050490098 Chr.3: 9705789-9705789 on Build GRCh38
rs767848325 Chr.3: 9705534-9705534 on Build GRCh38
rs930336497 Chr.3: 9705778-9705778 on Build GRCh38
rs748090409 Chr.3: 9705493-9705493 on Build GRCh38
rs771164883 Chr.3: 9705766-9705766 on Build GRCh38
rs1045655788 Chr.3: 9705612-9705612 on Build GRCh38
rs778855159 Chr.3: 9705489-9705489 on Build GRCh38
rs1037625832 Chr.3: 9705779-9705779 on Build GRCh38
rs540857064 Chr.3: 9705395-9705395 on Build GRCh38
rs1031459745 Chr.3: 9705499-9705499 on Build GRCh38
rs764712331 Chr.3: 9705549-9705549 on Build GRCh38
rs1055041641 Chr.3: 9705655-9705655 on Build GRCh38
rs556863883 Chr.3: 9705839-9705839 on Build GRCh38
rs950026098 Chr.3: 9705574-9705574 on Build GRCh38
rs565574640 Chr.3: 9705408-9705408 on Build GRCh38
rs758124402 Chr.3: 9705681-9705681 on Build GRCh38
rs776381529 Chr.3: 9705464-9705464 on Build GRCh38
rs759848118 Chr.3: 9705533-9705533 on Build GRCh38
rs540181988 Chr.3: 9705418-9705419 on Build GRCh38
rs754866144 Chr.3: 9705723-9705723 on Build GRCh38
rs562477220 Chr.3: 9705521-9705521 on Build GRCh38
rs777722337 Chr.3: 9705761-9705761 on Build GRCh38
rs748096178 Chr.3: 9705758-9705758 on Build GRCh38
rs751420010 Chr.3: 9705721-9705721 on Build GRCh38
rs749890819 Chr.3: 9705676-9705676 on Build GRCh38
rs532895364 Chr.3: 9705424-9705424 on Build GRCh38
rs897294172 Chr.3: 9705784-9705784 on Build GRCh38
rs890058665 Chr.3: 9705816-9705816 on Build GRCh38
rs780006927 Chr.3: 9705425-9705425 on Build GRCh38
rs749326522 Chr.3: 9705765-9705765 on Build GRCh38
rs746139548 Chr.3: 9705715-9705715 on Build GRCh38
rs927771012 Chr.3: 9705637-9705637 on Build GRCh38
rs981966947 Chr.3: 9705624-9705624 on Build GRCh38

View Full Product Details