Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9712586-9713086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE9
Entrez Gene ID
151835
Gene Name
copine family member 9
Gene Aliases
-
UniGene
Hs.121335
Gene Chromosome Location
Chr.3: 9703807-9729908 on Build GRCh38
Transcripts
NM_001308388.1 NM_153635.2 XM_011533386.2 XM_011533388.2 XM_011533389.2 XM_017005747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1049062 c.545C>T p.T182M Chr.3: 9712828-9712828 on Build GRCh38
6469151 c.627T>C p.A209A Chr.3: 9713056-9713056 on Build GRCh38
1049064 c.628C>T p.L210L Chr.3: 9713057-9713057 on Build GRCh38
5400127 c.492C>T p.F164F Chr.3: 9712775-9712775 on Build GRCh38
2949789 c.501A>G p.K167K Chr.3: 9712784-9712784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147141975 Chr.3: 9712908-9712908 on Build GRCh38
rs144773387 Chr.3: 9712688-9712688 on Build GRCh38
rs778280074 Chr.3: 9712857-9712857 on Build GRCh38
rs187310 Chr.3: 9712650-9712650 on Build GRCh38
rs776440574 Chr.3: 9712763-9712763 on Build GRCh38
rs759743369 Chr.3: 9712996-9712996 on Build GRCh38
rs1018535612 Chr.3: 9712736-9712736 on Build GRCh38
rs369902762 Chr.3: 9712870-9712870 on Build GRCh38
rs369328540 Chr.3: 9712793-9712793 on Build GRCh38
rs543337789 Chr.3: 9712745-9712745 on Build GRCh38
rs368887169 Chr.3: 9712938-9712938 on Build GRCh38
rs757812463 Chr.3: 9713032-9713032 on Build GRCh38
rs1057451782 Chr.3: 9712638-9712638 on Build GRCh38
rs781519920 Chr.3: 9712831-9712831 on Build GRCh38
rs114886261 Chr.3: 9712937-9712937 on Build GRCh38
rs758550800 Chr.3: 9712679-9712679 on Build GRCh38
rs770178491 Chr.3: 9712834-9712842 on Build GRCh38
rs375304283 Chr.3: 9712760-9712760 on Build GRCh38
rs757191477 Chr.3: 9712647-9712647 on Build GRCh38
rs369200151 Chr.3: 9712907-9712907 on Build GRCh38
rs372168341 Chr.3: 9712651-9712651 on Build GRCh38
rs748488074 Chr.3: 9712693-9712693 on Build GRCh38
rs779238273 Chr.3: 9713040-9713040 on Build GRCh38
rs766536649 Chr.3: 9712817-9712817 on Build GRCh38
rs778214429 Chr.3: 9713016-9713016 on Build GRCh38
rs752944991 Chr.3: 9712838-9712838 on Build GRCh38
rs751529724 Chr.3: 9712822-9712822 on Build GRCh38
rs570939905 Chr.3: 9712944-9712944 on Build GRCh38
rs1008340188 Chr.3: 9712876-9712876 on Build GRCh38
rs773905027 Chr.3: 9712725-9712725 on Build GRCh38
rs758726029 Chr.3: 9712908-9712908 on Build GRCh38
rs1018756309 Chr.3: 9712882-9712882 on Build GRCh38
rs148300122 Chr.3: 9712947-9712947 on Build GRCh38
rs188891650 Chr.3: 9713023-9713023 on Build GRCh38
rs749727725 Chr.3: 9712743-9712743 on Build GRCh38
rs753570592 Chr.3: 9712636-9712636 on Build GRCh38
rs762792198 Chr.3: 9712802-9712802 on Build GRCh38
rs1035336795 Chr.3: 9712967-9712967 on Build GRCh38
rs770450996 Chr.3: 9712708-9712708 on Build GRCh38
rs905826428 Chr.3: 9712625-9712625 on Build GRCh38
rs768318945 Chr.3: 9712931-9712931 on Build GRCh38
rs376329459 Chr.3: 9712860-9712860 on Build GRCh38
rs763934079 Chr.3: 9712635-9712635 on Build GRCh38
rs372450025 Chr.3: 9712998-9712998 on Build GRCh38
rs781695937 Chr.3: 9712692-9712692 on Build GRCh38
rs753934673 Chr.3: 9712746-9712746 on Build GRCh38
rs751685389 Chr.3: 9712984-9712984 on Build GRCh38
rs764990547 Chr.3: 9712792-9712792 on Build GRCh38
rs183493275 Chr.3: 9712960-9712960 on Build GRCh38
rs375168515 Chr.3: 9712661-9712661 on Build GRCh38
rs756587753 Chr.3: 9712848-9712848 on Build GRCh38
rs938699729 Chr.3: 9712632-9712632 on Build GRCh38
rs760242134 Chr.3: 9712612-9712612 on Build GRCh38
rs146586055 Chr.3: 9712824-9712824 on Build GRCh38
rs9827361 Chr.3: 9712875-9712875 on Build GRCh38
rs763381372 Chr.3: 9712955-9712955 on Build GRCh38
rs779964680 Chr.3: 9712682-9712682 on Build GRCh38
rs768190119 Chr.3: 9713053-9713053 on Build GRCh38
rs930039324 Chr.3: 9712940-9712940 on Build GRCh38
rs149771786 Chr.3: 9712928-9712928 on Build GRCh38
rs746351427 Chr.3: 9713051-9713051 on Build GRCh38
rs561992622 Chr.3: 9712784-9712784 on Build GRCh38
rs776310726 Chr.3: 9712932-9712932 on Build GRCh38
rs753244902 Chr.3: 9712866-9712866 on Build GRCh38
rs562415577 Chr.3: 9712667-9712667 on Build GRCh38
rs753036508 Chr.3: 9713011-9713011 on Build GRCh38
rs767674647 Chr.3: 9712999-9712999 on Build GRCh38
rs756393346 Chr.3: 9713014-9713014 on Build GRCh38
rs190521882 Chr.3: 9712828-9712828 on Build GRCh38
rs760386532 Chr.3: 9712756-9712756 on Build GRCh38
rs762955623 Chr.3: 9712951-9712951 on Build GRCh38
rs564101817 Chr.3: 9712670-9712670 on Build GRCh38
rs143275352 Chr.3: 9712888-9712888 on Build GRCh38

View Full Product Details