Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9942971-9943469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRELD1
Entrez Gene ID
78987
Gene Name
cysteine rich with EGF like domains 1
Gene Aliases
AVSD2, CIRRIN
UniGene
Hs.9383
Gene Chromosome Location
Chr.3: 9933788-9945413 on Build GRCh38
Transcripts
NM_001031717.3 NM_001077415.2 NM_015513.4 XM_011534108.1 XM_017007175.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2951108 c.972C>T p.T324T Chr.3: 9943439-9943439 on Build GRCh38
5753141 c.833G>A p.C278Y Chr.3: 9943092-9943092 on Build GRCh38
5757987 c.954C>T p.N318N Chr.3: 9943421-9943421 on Build GRCh38
7280624 c.873G>A p.K291K Chr.3: 9943132-9943132 on Build GRCh38
341944 c.979G>T p.G327C Chr.3: 9943446-9943446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008947024 Chr.3: 9943250-9943250 on Build GRCh38
rs374104985 Chr.3: 9943216-9943216 on Build GRCh38
rs554925730 Chr.3: 9943061-9943061 on Build GRCh38
rs1033251868 Chr.3: 9943359-9943359 on Build GRCh38
rs749915183 Chr.3: 9943411-9943411 on Build GRCh38
rs760897938 Chr.3: 9943038-9943038 on Build GRCh38
rs200714636 Chr.3: 9943374-9943374 on Build GRCh38
rs34842990 Chr.3: 9943129-9943130 on Build GRCh38
rs370127844 Chr.3: 9943065-9943065 on Build GRCh38
rs374333743 Chr.3: 9943445-9943445 on Build GRCh38
rs755381475 Chr.3: 9943067-9943067 on Build GRCh38
rs79223485 Chr.3: 9943171-9943171 on Build GRCh38
rs759473511 Chr.3: 9943426-9943426 on Build GRCh38
rs983476737 Chr.3: 9943087-9943087 on Build GRCh38
rs767862346 Chr.3: 9943200-9943200 on Build GRCh38
rs1057421795 Chr.3: 9943229-9943229 on Build GRCh38
rs765462584 Chr.3: 9943414-9943414 on Build GRCh38
rs772178038 Chr.3: 9943026-9943026 on Build GRCh38
rs756718838 Chr.3: 9943203-9943203 on Build GRCh38
rs28942092 Chr.3: 9943399-9943399 on Build GRCh38
rs534019383 Chr.3: 9943146-9943146 on Build GRCh38
rs947142551 Chr.3: 9943164-9943164 on Build GRCh38
rs760118784 Chr.3: 9943199-9943199 on Build GRCh38
rs763909257 Chr.3: 9943421-9943424 on Build GRCh38
rs746951044 Chr.3: 9943122-9943122 on Build GRCh38
rs779279495 Chr.3: 9943187-9943187 on Build GRCh38
rs771099812 Chr.3: 9943379-9943379 on Build GRCh38
rs752297888 Chr.3: 9943443-9943444 on Build GRCh38
rs776511431 Chr.3: 9943044-9943044 on Build GRCh38
rs199785910 Chr.3: 9943053-9943053 on Build GRCh38
rs116780838 Chr.3: 9943169-9943169 on Build GRCh38
rs761187806 Chr.3: 9943403-9943403 on Build GRCh38
rs575057703 Chr.3: 9943308-9943308 on Build GRCh38
rs916328885 Chr.3: 9943004-9943004 on Build GRCh38
rs775848791 Chr.3: 9943401-9943401 on Build GRCh38
rs769745964 Chr.3: 9943341-9943341 on Build GRCh38
rs768797943 Chr.3: 9943040-9943040 on Build GRCh38
rs1023653653 Chr.3: 9943280-9943280 on Build GRCh38
rs771191734 Chr.3: 9943446-9943446 on Build GRCh38
rs780956328 Chr.3: 9943335-9943335 on Build GRCh38
rs772094283 Chr.3: 9943172-9943172 on Build GRCh38
rs756956925 Chr.3: 9943097-9943097 on Build GRCh38
rs777339615 Chr.3: 9943069-9943069 on Build GRCh38
rs760717626 Chr.3: 9943271-9943271 on Build GRCh38
rs755981922 Chr.3: 9943440-9943440 on Build GRCh38
rs373820286 Chr.3: 9943048-9943048 on Build GRCh38
rs367972424 Chr.3: 9943340-9943340 on Build GRCh38
rs765041298 Chr.3: 9943205-9943205 on Build GRCh38
rs544520546 Chr.3: 9943254-9943254 on Build GRCh38
rs758060687 Chr.3: 9943215-9943215 on Build GRCh38
rs773216920 Chr.3: 9943372-9943372 on Build GRCh38
rs895618882 Chr.3: 9943259-9943259 on Build GRCh38
rs576984269 Chr.3: 9943183-9943183 on Build GRCh38
rs951653246 Chr.3: 9943074-9943074 on Build GRCh38
rs62245481 Chr.3: 9943013-9943013 on Build GRCh38
rs751414991 Chr.3: 9943427-9943427 on Build GRCh38
rs750115424 Chr.3: 9943206-9943206 on Build GRCh38
rs996611527 Chr.3: 9943132-9943132 on Build GRCh38
rs563014562 Chr.3: 9943268-9943268 on Build GRCh38
rs753428326 Chr.3: 9943202-9943202 on Build GRCh38
rs1023683410 Chr.3: 9943020-9943020 on Build GRCh38
rs746736174 Chr.3: 9943334-9943334 on Build GRCh38
rs370824760 Chr.3: 9943391-9943391 on Build GRCh38
rs944195374 Chr.3: 9943119-9943119 on Build GRCh38
rs76764016 Chr.3: 9943412-9943412 on Build GRCh38
rs766742034 Chr.3: 9943173-9943173 on Build GRCh38
rs201083358 Chr.3: 9943114-9943114 on Build GRCh38
rs536223283 Chr.3: 9943055-9943055 on Build GRCh38
rs1019888360 Chr.3: 9943426-9943426 on Build GRCh38
rs754728781 Chr.3: 9943439-9943439 on Build GRCh38
rs902883115 Chr.3: 9943196-9943196 on Build GRCh38
rs748150498 Chr.3: 9943154-9943154 on Build GRCh38
rs983020063 Chr.3: 9943418-9943418 on Build GRCh38
rs776799121 Chr.3: 9943149-9943149 on Build GRCh38
rs371166779 Chr.3: 9943032-9943032 on Build GRCh38
rs978881304 Chr.3: 9943278-9943278 on Build GRCh38
rs1020316171 Chr.3: 9943256-9943256 on Build GRCh38
rs974064584 Chr.3: 9943028-9943028 on Build GRCh38
rs778399642 Chr.3: 9943106-9943106 on Build GRCh38
rs745725769 Chr.3: 9943109-9943109 on Build GRCh38
rs774719172 Chr.3: 9943383-9943383 on Build GRCh38
rs138442570 Chr.3: 9943306-9943307 on Build GRCh38
rs753734694 Chr.3: 9943405-9943405 on Build GRCh38
rs773044321 Chr.3: 9943260-9943260 on Build GRCh38
rs969332306 Chr.3: 9943022-9943022 on Build GRCh38

View Full Product Details