Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10090278-10090810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCD2
Entrez Gene ID
2177
Gene Name
Fanconi anemia complementation group D2
Gene Aliases
FA-D2, FA4, FACD, FAD, FAD2, FANCD
UniGene
Hs.208388
Gene Chromosome Location
Chr.3: 10026387-10101937 on Build GRCh38
Transcripts
NM_001018115.2 NM_001319984.1 NM_033084.4 NM_173472.1

Gene Symbol
FANCD2OS
Entrez Gene ID
115795
Gene Name
FANCD2 opposite strand
Gene Aliases
C3orf24
UniGene
Hs.190413
Gene Chromosome Location
Chr.3: 10081320-10108603 on Build GRCh38
Transcripts
NM_001018115.2 NM_001319984.1 NM_033084.4 NM_173472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6207162 c.3714G>A p.M1238I Chr.3: 10090322-10090322 on Build GRCh38
5963290 c.3777+5G>C p.? Chr.3: 10090390-10090390 on Build GRCh38
4975872 c.3707G>T p.R1236L Chr.3: 10090315-10090315 on Build GRCh38
1417256 c.3738_3739insA p.I1249fs*2 Chr.3: 10090346-10090347 on Build GRCh38
4991808 c.3756C>A p.G1252G Chr.3: 10090364-10090364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757808149 Chr.3: 10090356-10090356 on Build GRCh38
rs77529198 Chr.3: 10090467-10090467 on Build GRCh38
rs750130327 Chr.3: 10090343-10090343 on Build GRCh38
rs958137591 Chr.3: 10090612-10090617 on Build GRCh38
rs368999017 Chr.3: 10090390-10090390 on Build GRCh38
rs1021994462 Chr.3: 10090488-10090488 on Build GRCh38
rs774421283 Chr.3: 10090398-10090398 on Build GRCh38
rs972834689 Chr.3: 10090685-10090685 on Build GRCh38
rs779242727 Chr.3: 10090379-10090379 on Build GRCh38
rs764775864 Chr.3: 10090342-10090342 on Build GRCh38
rs746017476 Chr.3: 10090410-10090410 on Build GRCh38
rs940930676 Chr.3: 10090682-10090682 on Build GRCh38
rs1047237177 Chr.3: 10090393-10090393 on Build GRCh38
rs803342 Chr.3: 10090444-10090444 on Build GRCh38
rs368684162 Chr.3: 10090376-10090376 on Build GRCh38
rs775831214 Chr.3: 10090319-10090319 on Build GRCh38
rs982268437 Chr.3: 10090652-10090652 on Build GRCh38
rs138480262 Chr.3: 10090542-10090542 on Build GRCh38
rs56036549 Chr.3: 10090577-10090577 on Build GRCh38
rs56102803 Chr.3: 10090728-10090728 on Build GRCh38
rs371651879 Chr.3: 10090521-10090521 on Build GRCh38
rs776029741 Chr.3: 10090437-10090437 on Build GRCh38
rs747942447 Chr.3: 10090366-10090366 on Build GRCh38
rs967328313 Chr.3: 10090498-10090498 on Build GRCh38
rs977618164 Chr.3: 10090505-10090505 on Build GRCh38
rs960752642 Chr.3: 10090573-10090573 on Build GRCh38
rs773716319 Chr.3: 10090444-10090447 on Build GRCh38
rs767806932 Chr.3: 10090318-10090318 on Build GRCh38
rs535842006 Chr.3: 10090528-10090528 on Build GRCh38
rs762915519 Chr.3: 10090300-10090300 on Build GRCh38
rs770863196 Chr.3: 10090385-10090385 on Build GRCh38
rs963702682 Chr.3: 10090471-10090471 on Build GRCh38
rs771721585 Chr.3: 10090386-10090389 on Build GRCh38
rs568515612 Chr.3: 10090359-10090359 on Build GRCh38
rs1032323912 Chr.3: 10090562-10090562 on Build GRCh38
rs949649696 Chr.3: 10090320-10090320 on Build GRCh38
rs549383233 Chr.3: 10090741-10090741 on Build GRCh38
rs537878728 Chr.3: 10090622-10090622 on Build GRCh38
rs143964628 Chr.3: 10090709-10090709 on Build GRCh38
rs781344292 Chr.3: 10090362-10090362 on Build GRCh38
rs1035547094 Chr.3: 10090519-10090519 on Build GRCh38
rs772442959 Chr.3: 10090650-10090650 on Build GRCh38
rs1040858089 Chr.3: 10090785-10090785 on Build GRCh38
rs761089162 Chr.3: 10090321-10090321 on Build GRCh38
rs902397199 Chr.3: 10090511-10090511 on Build GRCh38
rs772558102 Chr.3: 10090417-10090417 on Build GRCh38
rs190275884 Chr.3: 10090673-10090673 on Build GRCh38
rs1011444431 Chr.3: 10090478-10090478 on Build GRCh38
rs374940277 Chr.3: 10090378-10090378 on Build GRCh38
rs1023848158 Chr.3: 10090646-10090646 on Build GRCh38
rs77391196 Chr.3: 10090472-10090472 on Build GRCh38
rs55891340 Chr.3: 10090504-10090504 on Build GRCh38
rs943879739 Chr.3: 10090784-10090784 on Build GRCh38
rs994778998 Chr.3: 10090380-10090380 on Build GRCh38
rs1017880445 Chr.3: 10090470-10090470 on Build GRCh38
rs1019954618 Chr.3: 10090503-10090503 on Build GRCh38
rs573953089 Chr.3: 10090749-10090749 on Build GRCh38
rs121917786 Chr.3: 10090315-10090315 on Build GRCh38
rs187313930 Chr.3: 10090619-10090619 on Build GRCh38
rs558364451 Chr.3: 10090620-10090620 on Build GRCh38
rs113633744 Chr.3: 10090776-10090776 on Build GRCh38
rs370370618 Chr.3: 10090529-10090529 on Build GRCh38
rs771078251 Chr.3: 10090314-10090314 on Build GRCh38
rs760054945 Chr.3: 10090316-10090316 on Build GRCh38
rs748337156 Chr.3: 10090506-10090506 on Build GRCh38
rs55716118 Chr.3: 10090735-10090735 on Build GRCh38
rs999951188 Chr.3: 10090546-10090546 on Build GRCh38
rs150540355 Chr.3: 10090744-10090744 on Build GRCh38
rs974315136 Chr.3: 10090651-10090651 on Build GRCh38

View Full Product Details