Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196706519-196707029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP19
Entrez Gene ID
84984
Gene Name
centrosomal protein 19
Gene Aliases
C3orf34, MOSPGF
UniGene
Hs.282800
Gene Chromosome Location
Chr.3: 196706273-196712294 on Build GRCh38
Transcripts
NM_032898.4 XM_005269370.4 XM_011513246.2

SNP Detail

SNP ID Chromosome Location
rs1021446905 Chr.3: 196706931-196706931 on Build GRCh38
rs893785943 Chr.3: 196706881-196706881 on Build GRCh38
rs59114665 Chr.3: 196706635-196706635 on Build GRCh38
rs902930159 Chr.3: 196706694-196706694 on Build GRCh38
rs931066925 Chr.3: 196706733-196706733 on Build GRCh38
rs914821162 Chr.3: 196706790-196706790 on Build GRCh38
rs529935439 Chr.3: 196706892-196706892 on Build GRCh38
rs570226607 Chr.3: 196706654-196706654 on Build GRCh38
rs1011720469 Chr.3: 196706680-196706680 on Build GRCh38
rs1041678462 Chr.3: 196706545-196706545 on Build GRCh38
rs181258050 Chr.3: 196706865-196706865 on Build GRCh38
rs7646550 Chr.3: 196706550-196706550 on Build GRCh38
rs947659960 Chr.3: 196706803-196706803 on Build GRCh38
rs996490215 Chr.3: 196706959-196706959 on Build GRCh38
rs188697990 Chr.3: 196706760-196706760 on Build GRCh38
rs71621289 Chr.3: 196706783-196706785 on Build GRCh38
rs1041887949 Chr.3: 196706804-196706804 on Build GRCh38
rs73892924 Chr.3: 196706817-196706817 on Build GRCh38
rs1015405254 Chr.3: 196706751-196706751 on Build GRCh38
rs1019684389 Chr.3: 196706719-196706720 on Build GRCh38
rs564155516 Chr.3: 196706757-196706757 on Build GRCh38
rs888360085 Chr.3: 196706978-196706978 on Build GRCh38
rs967597605 Chr.3: 196706607-196706607 on Build GRCh38
rs1020135568 Chr.3: 196706587-196706587 on Build GRCh38
rs944195127 Chr.3: 196706750-196706750 on Build GRCh38
rs187077574 Chr.3: 196706985-196706985 on Build GRCh38
rs937352982 Chr.3: 196706608-196706608 on Build GRCh38
rs536206035 Chr.3: 196706602-196706602 on Build GRCh38
rs558586057 Chr.3: 196706556-196706556 on Build GRCh38
rs556846529 Chr.3: 196706574-196706574 on Build GRCh38
rs928530762 Chr.3: 196706641-196706641 on Build GRCh38
rs1007998230 Chr.3: 196706697-196706697 on Build GRCh38
rs757747606 Chr.3: 196706741-196706741 on Build GRCh38
rs141655480 Chr.3: 196706737-196706737 on Build GRCh38
rs142939945 Chr.3: 196706544-196706544 on Build GRCh38
rs903052532 Chr.3: 196706934-196706934 on Build GRCh38
rs147684020 Chr.3: 196706691-196706691 on Build GRCh38
rs1046126530 Chr.3: 196706575-196706576 on Build GRCh38
rs1056029105 Chr.3: 196706623-196706623 on Build GRCh38
rs7616240 Chr.3: 196706752-196706752 on Build GRCh38
rs146272086 Chr.3: 196706720-196706720 on Build GRCh38
rs77890712 Chr.3: 196706957-196706957 on Build GRCh38
rs937319852 Chr.3: 196706780-196706780 on Build GRCh38
rs543851425 Chr.3: 196706572-196706572 on Build GRCh38
rs899467475 Chr.3: 196706730-196706730 on Build GRCh38
rs760915333 Chr.3: 196706657-196706657 on Build GRCh38
rs75357825 Chr.3: 196706785-196706785 on Build GRCh38
rs1026642230 Chr.3: 196706971-196706971 on Build GRCh38
rs9840915 Chr.3: 196706570-196706570 on Build GRCh38
rs79234712 Chr.3: 196706928-196706928 on Build GRCh38
rs997021883 Chr.3: 196706761-196706761 on Build GRCh38
rs751049134 Chr.3: 196706759-196706759 on Build GRCh38
rs919561452 Chr.3: 196706731-196706731 on Build GRCh38
rs142454352 Chr.3: 196706702-196706702 on Build GRCh38
rs996799153 Chr.3: 196706569-196706569 on Build GRCh38
rs764374135 Chr.3: 196706690-196706690 on Build GRCh38
rs1047230784 Chr.3: 196706735-196706735 on Build GRCh38
rs60302538 Chr.3: 196706849-196706849 on Build GRCh38

View Full Product Details