Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 440122-440631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA11P
Entrez Gene ID
79963
Gene Name
ATP binding cassette subfamily A member 11, pseudogene
Gene Aliases
ABCA11, EST1133530
UniGene
Hs.677493
Gene Chromosome Location
Chr.4: 425435-474209 on Build GRCh38
Transcripts
NR_002451.2 NM_133474.3

Gene Symbol
ZNF721
Entrez Gene ID
170960
Gene Name
zinc finger protein 721
Gene Aliases
-
UniGene
Hs.428360
Gene Chromosome Location
Chr.4: 439984-499171 on Build GRCh38
Transcripts
NR_002451.2 NM_133474.3

SNP Detail

SNP ID Chromosome Location
rs887088091 Chr.4: 440370-440370 on Build GRCh38
rs1045944797 Chr.4: 440251-440251 on Build GRCh38
rs397745975 Chr.4: 440185-440185 on Build GRCh38
rs1032181205 Chr.4: 440471-440471 on Build GRCh38
rs1056007696 Chr.4: 440299-440299 on Build GRCh38
rs765358077 Chr.4: 440525-440525 on Build GRCh38
rs775724845 Chr.4: 440550-440553 on Build GRCh38
rs577773458 Chr.4: 440209-440209 on Build GRCh38
rs957783288 Chr.4: 440151-440151 on Build GRCh38
rs577489515 Chr.4: 440544-440544 on Build GRCh38
rs1045887 Chr.4: 440290-440290 on Build GRCh38
rs1047072163 Chr.4: 440355-440355 on Build GRCh38
rs79377789 Chr.4: 440163-440163 on Build GRCh38
rs146779980 Chr.4: 440456-440456 on Build GRCh38
rs1019941182 Chr.4: 440394-440394 on Build GRCh38
rs143449501 Chr.4: 440555-440555 on Build GRCh38
rs571985007 Chr.4: 440422-440422 on Build GRCh38
rs913679059 Chr.4: 440510-440510 on Build GRCh38
rs574400411 Chr.4: 440307-440307 on Build GRCh38
rs547914828 Chr.4: 440387-440388 on Build GRCh38
rs544923673 Chr.4: 440213-440213 on Build GRCh38
rs937610908 Chr.4: 440586-440586 on Build GRCh38
rs527625994 Chr.4: 440388-440389 on Build GRCh38
rs970355866 Chr.4: 440532-440532 on Build GRCh38
rs957757352 Chr.4: 440474-440474 on Build GRCh38
rs781828678 Chr.4: 440520-440520 on Build GRCh38
rs35069635 Chr.4: 440184-440184 on Build GRCh38
rs995598266 Chr.4: 440346-440346 on Build GRCh38
rs35230303 Chr.4: 440545-440546 on Build GRCh38
rs139982108 Chr.4: 440477-440477 on Build GRCh38
rs987731292 Chr.4: 440167-440167 on Build GRCh38
rs937498468 Chr.4: 440297-440297 on Build GRCh38
rs981834913 Chr.4: 440537-440537 on Build GRCh38
rs898534681 Chr.4: 440308-440308 on Build GRCh38
rs553724946 Chr.4: 440400-440400 on Build GRCh38
rs946280873 Chr.4: 440225-440225 on Build GRCh38
rs187547729 Chr.4: 440388-440388 on Build GRCh38

View Full Product Details