Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 792296-792785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPLX1
Entrez Gene ID
10815
Gene Name
complexin 1
Gene Aliases
CPX-I, CPX1
UniGene
Hs.478930
Gene Chromosome Location
Chr.4: 784957-826157 on Build GRCh38
Transcripts
NM_006651.3 XM_011513391.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5701127 c.65_66insG p.D23fs*1 Chr.4: 792574-792575 on Build GRCh38
5732410 c.172G>A p.E58K Chr.4: 792468-792468 on Build GRCh38
4003153 c.32-5A>G p.? Chr.4: 792613-792613 on Build GRCh38
5662544 c.154T>A p.Y52N Chr.4: 792486-792486 on Build GRCh38
5803716 c.175C>A p.R59S Chr.4: 792465-792465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74519932 Chr.4: 792459-792459 on Build GRCh38
rs953386163 Chr.4: 792762-792762 on Build GRCh38
rs998408587 Chr.4: 792360-792360 on Build GRCh38
rs745821806 Chr.4: 792516-792516 on Build GRCh38
rs373179110 Chr.4: 792406-792406 on Build GRCh38
rs561994728 Chr.4: 792424-792424 on Build GRCh38
rs373896763 Chr.4: 792543-792543 on Build GRCh38
rs755432411 Chr.4: 792472-792472 on Build GRCh38
rs371042405 Chr.4: 792456-792456 on Build GRCh38
rs1036759703 Chr.4: 792562-792562 on Build GRCh38
rs375498137 Chr.4: 792350-792350 on Build GRCh38
rs749616705 Chr.4: 792384-792384 on Build GRCh38
rs762707703 Chr.4: 792565-792567 on Build GRCh38
rs760717134 Chr.4: 792623-792623 on Build GRCh38
rs775090510 Chr.4: 792457-792457 on Build GRCh38
rs941391684 Chr.4: 792584-792584 on Build GRCh38
rs746939470 Chr.4: 792655-792655 on Build GRCh38
rs1050140307 Chr.4: 792515-792515 on Build GRCh38
rs1051338763 Chr.4: 792671-792671 on Build GRCh38
rs932551815 Chr.4: 792485-792485 on Build GRCh38
rs757504656 Chr.4: 792561-792561 on Build GRCh38
rs551494928 Chr.4: 792549-792549 on Build GRCh38
rs757556064 Chr.4: 792636-792636 on Build GRCh38
rs765495551 Chr.4: 792638-792638 on Build GRCh38
rs543655831 Chr.4: 792749-792749 on Build GRCh38
rs772092684 Chr.4: 792518-792518 on Build GRCh38
rs781284680 Chr.4: 792477-792477 on Build GRCh38
rs758831727 Chr.4: 792413-792413 on Build GRCh38
rs772717921 Chr.4: 792497-792529 on Build GRCh38
rs778881257 Chr.4: 792575-792575 on Build GRCh38
rs751884989 Chr.4: 792550-792550 on Build GRCh38
rs941492500 Chr.4: 792347-792347 on Build GRCh38
rs759157151 Chr.4: 792454-792454 on Build GRCh38
rs752366371 Chr.4: 792398-792398 on Build GRCh38
rs780025703 Chr.4: 792582-792582 on Build GRCh38
rs535706262 Chr.4: 792621-792621 on Build GRCh38
rs909997470 Chr.4: 792339-792339 on Build GRCh38
rs965869781 Chr.4: 792743-792743 on Build GRCh38
rs774713077 Chr.4: 792606-792606 on Build GRCh38
rs763262947 Chr.4: 792547-792547 on Build GRCh38
rs541047416 Chr.4: 792463-792463 on Build GRCh38
rs760171732 Chr.4: 792522-792522 on Build GRCh38
rs989601416 Chr.4: 792732-792732 on Build GRCh38
rs769372085 Chr.4: 792604-792604 on Build GRCh38
rs372963634 Chr.4: 792466-792466 on Build GRCh38
rs750758191 Chr.4: 792409-792409 on Build GRCh38
rs745590886 Chr.4: 792576-792576 on Build GRCh38
rs767976894 Chr.4: 792616-792616 on Build GRCh38
rs751692728 Chr.4: 792426-792426 on Build GRCh38
rs760044462 Chr.4: 792551-792551 on Build GRCh38
rs756306382 Chr.4: 792484-792484 on Build GRCh38
rs749055234 Chr.4: 792445-792445 on Build GRCh38
rs769407191 Chr.4: 792393-792394 on Build GRCh38
rs199531432 Chr.4: 792595-792595 on Build GRCh38
rs768248598 Chr.4: 792529-792529 on Build GRCh38
rs1019971138 Chr.4: 792416-792416 on Build GRCh38
rs2306251 Chr.4: 792613-792613 on Build GRCh38
rs752757823 Chr.4: 792482-792482 on Build GRCh38
rs970904969 Chr.4: 792682-792682 on Build GRCh38
rs779042489 Chr.4: 792508-792508 on Build GRCh38
rs370561245 Chr.4: 792554-792554 on Build GRCh38
rs900008828 Chr.4: 792359-792359 on Build GRCh38
rs747832797 Chr.4: 792601-792601 on Build GRCh38
rs546240562 Chr.4: 792366-792366 on Build GRCh38
rs934185217 Chr.4: 792662-792662 on Build GRCh38
rs768176752 Chr.4: 792592-792592 on Build GRCh38
rs958109914 Chr.4: 792728-792728 on Build GRCh38
rs368950761 Chr.4: 792355-792355 on Build GRCh38
rs1027164711 Chr.4: 792423-792425 on Build GRCh38
rs1054358258 Chr.4: 792331-792331 on Build GRCh38
rs553761470 Chr.4: 792674-792674 on Build GRCh38
rs750299210 Chr.4: 792645-792645 on Build GRCh38
rs746371966 Chr.4: 792541-792543 on Build GRCh38
rs767957006 Chr.4: 792553-792553 on Build GRCh38
rs909896685 Chr.4: 792538-792538 on Build GRCh38
rs557963884 Chr.4: 792372-792372 on Build GRCh38
rs770021210 Chr.4: 792440-792440 on Build GRCh38
rs774426352 Chr.4: 792449-792449 on Build GRCh38
rs951400865 Chr.4: 792469-792469 on Build GRCh38
rs2306250 Chr.4: 792720-792720 on Build GRCh38
rs748344625 Chr.4: 792429-792429 on Build GRCh38
rs770818939 Chr.4: 792500-792500 on Build GRCh38
rs999917177 Chr.4: 792428-792428 on Build GRCh38
rs191893022 Chr.4: 792702-792702 on Build GRCh38
rs558257499 Chr.4: 792709-792709 on Build GRCh38
rs781445446 Chr.4: 792427-792427 on Build GRCh38
rs759104063 Chr.4: 792383-792383 on Build GRCh38
rs921919043 Chr.4: 792738-792738 on Build GRCh38
rs1000123879 Chr.4: 792719-792719 on Build GRCh38
rs778044492 Chr.4: 792487-792487 on Build GRCh38
rs376378419 Chr.4: 792442-792442 on Build GRCh38
rs758260996 Chr.4: 792651-792651 on Build GRCh38
rs531966510 Chr.4: 792588-792588 on Build GRCh38
rs200282920 Chr.4: 792521-792521 on Build GRCh38
rs576236702 Chr.4: 792385-792385 on Build GRCh38
rs779893229 Chr.4: 792652-792652 on Build GRCh38
rs749104326 Chr.4: 792490-792490 on Build GRCh38
rs569648725 Chr.4: 792579-792579 on Build GRCh38
rs896808047 Chr.4: 792404-792404 on Build GRCh38
rs936686586 Chr.4: 792321-792321 on Build GRCh38
rs764256636 Chr.4: 792629-792629 on Build GRCh38
rs767345763 Chr.4: 792396-792396 on Build GRCh38
rs750301545 Chr.4: 792368-792368 on Build GRCh38
rs1025139736 Chr.4: 792443-792443 on Build GRCh38
rs768036153 Chr.4: 792641-792641 on Build GRCh38
rs1047136926 Chr.4: 792333-792333 on Build GRCh38
rs370860166 Chr.4: 792632-792632 on Build GRCh38
rs369649751 Chr.4: 792511-792511 on Build GRCh38
rs376139437 Chr.4: 792470-792470 on Build GRCh38
rs376067463 Chr.4: 792754-792754 on Build GRCh38
rs757689645 Chr.4: 792403-792403 on Build GRCh38
rs775215228 Chr.4: 792535-792535 on Build GRCh38
rs754069852 Chr.4: 792560-792560 on Build GRCh38

View Full Product Details