Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 850594-851091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAK
Entrez Gene ID
2580
Gene Name
cyclin G associated kinase
Gene Aliases
DNAJ26, DNAJC26
UniGene
Hs.369607
Gene Chromosome Location
Chr.4: 849275-932390 on Build GRCh38
Transcripts
NM_001318134.1 NM_005255.3 XM_005272268.2 XM_005272270.2 XM_011513425.2 XM_011513426.2 XM_011513427.2 XM_011513428.2 XM_011513429.2 XM_011513430.1 XM_011513431.2 XM_011513432.2 XM_011513434.2 XM_017007991.1 XM_017007992.1 XM_017007993.1 XM_017007995.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4901540 c.3593C>T p.P1198L Chr.4: 851000-851000 on Build GRCh38
6101282 c.3612G>T p.M1204I Chr.4: 850981-850981 on Build GRCh38
7167614 c.3559C>G p.Q1187E Chr.4: 851034-851034 on Build GRCh38
6889025 c.3531G>A p.E1177E Chr.4: 851062-851062 on Build GRCh38
6469718 c.3621G>T p.Q1207H Chr.4: 850972-850972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914596920 Chr.4: 850851-850851 on Build GRCh38
rs745713926 Chr.4: 851004-851004 on Build GRCh38
rs765018640 Chr.4: 851024-851024 on Build GRCh38
rs756641298 Chr.4: 851065-851065 on Build GRCh38
rs765183365 Chr.4: 850860-850860 on Build GRCh38
rs553311068 Chr.4: 850640-850640 on Build GRCh38
rs1052679108 Chr.4: 850868-850868 on Build GRCh38
rs753290164 Chr.4: 850954-850954 on Build GRCh38
rs749629132 Chr.4: 850783-850783 on Build GRCh38
rs954051565 Chr.4: 850853-850853 on Build GRCh38
rs781776214 Chr.4: 850951-850951 on Build GRCh38
rs1045041912 Chr.4: 850947-850947 on Build GRCh38
rs749480032 Chr.4: 850892-850892 on Build GRCh38
rs541463716 Chr.4: 850639-850639 on Build GRCh38
rs757337983 Chr.4: 850844-850844 on Build GRCh38
rs971963757 Chr.4: 850796-850796 on Build GRCh38
rs779077568 Chr.4: 851000-851000 on Build GRCh38
rs776063176 Chr.4: 851018-851018 on Build GRCh38
rs891584632 Chr.4: 850701-850701 on Build GRCh38
rs1011651588 Chr.4: 851002-851002 on Build GRCh38
rs775366773 Chr.4: 850622-850622 on Build GRCh38
rs986442413 Chr.4: 850854-850854 on Build GRCh38
rs139521838 Chr.4: 851046-851046 on Build GRCh38
rs994089843 Chr.4: 850955-850955 on Build GRCh38
rs911365643 Chr.4: 850808-850808 on Build GRCh38
rs372960827 Chr.4: 850922-850922 on Build GRCh38
rs775385885 Chr.4: 850912-850912 on Build GRCh38
rs369002990 Chr.4: 851059-851059 on Build GRCh38
rs777732237 Chr.4: 850975-850975 on Build GRCh38
rs574672209 Chr.4: 850633-850633 on Build GRCh38
rs760532650 Chr.4: 851014-851014 on Build GRCh38
rs369909897 Chr.4: 850890-850890 on Build GRCh38
rs915128455 Chr.4: 850641-850641 on Build GRCh38
rs767484229 Chr.4: 850907-850907 on Build GRCh38
rs932983908 Chr.4: 850861-850861 on Build GRCh38
rs918975745 Chr.4: 850883-850883 on Build GRCh38
rs139558681 Chr.4: 850957-850957 on Build GRCh38
rs377387724 Chr.4: 851011-851011 on Build GRCh38
rs563088646 Chr.4: 850744-850744 on Build GRCh38
rs1001432608 Chr.4: 850736-850736 on Build GRCh38
rs549936853 Chr.4: 850923-850923 on Build GRCh38
rs756580105 Chr.4: 850968-850968 on Build GRCh38
rs1011743113 Chr.4: 850712-850712 on Build GRCh38
rs527799207 Chr.4: 850813-850813 on Build GRCh38
rs182378795 Chr.4: 850668-850668 on Build GRCh38
rs1031857213 Chr.4: 850958-850958 on Build GRCh38
rs748169956 Chr.4: 850889-850890 on Build GRCh38
rs566842422 Chr.4: 850797-850797 on Build GRCh38
rs773305512 Chr.4: 850889-850889 on Build GRCh38
rs747566624 Chr.4: 850642-850643 on Build GRCh38
rs368035537 Chr.4: 850920-850920 on Build GRCh38
rs532631162 Chr.4: 850714-850714 on Build GRCh38
rs1041706469 Chr.4: 850618-850618 on Build GRCh38
rs750074717 Chr.4: 851028-851028 on Build GRCh38
rs762643513 Chr.4: 851042-851042 on Build GRCh38
rs990151880 Chr.4: 850627-850627 on Build GRCh38
rs771878376 Chr.4: 851005-851005 on Build GRCh38
rs1052795829 Chr.4: 850681-850681 on Build GRCh38
rs569155402 Chr.4: 850922-850923 on Build GRCh38
rs767423280 Chr.4: 850617-850617 on Build GRCh38
rs138652612 Chr.4: 850835-850835 on Build GRCh38
rs75072999 Chr.4: 850993-850993 on Build GRCh38
rs1031723269 Chr.4: 850824-850824 on Build GRCh38
rs539813887 Chr.4: 850966-850966 on Build GRCh38
rs779580275 Chr.4: 850766-850766 on Build GRCh38
rs544773314 Chr.4: 850724-850724 on Build GRCh38
rs1042859802 Chr.4: 850719-850719 on Build GRCh38
rs933016470 Chr.4: 850680-850680 on Build GRCh38
rs912896043 Chr.4: 850876-850876 on Build GRCh38
rs149731617 Chr.4: 850988-850988 on Build GRCh38
rs549021707 Chr.4: 850824-850824 on Build GRCh38
rs531130793 Chr.4: 850827-850827 on Build GRCh38
rs774194119 Chr.4: 850896-850896 on Build GRCh38
rs1025733197 Chr.4: 850793-850793 on Build GRCh38
rs1025617036 Chr.4: 850967-850967 on Build GRCh38
rs150598346 Chr.4: 850940-850940 on Build GRCh38
rs888006143 Chr.4: 850752-850752 on Build GRCh38
rs530410176 Chr.4: 850774-850774 on Build GRCh38
rs548613779 Chr.4: 850794-850794 on Build GRCh38
rs761377801 Chr.4: 851020-851020 on Build GRCh38
rs994206969 Chr.4: 850782-850782 on Build GRCh38
rs1040523034 Chr.4: 850686-850686 on Build GRCh38
rs760609467 Chr.4: 850913-850913 on Build GRCh38
rs1050249067 Chr.4: 850649-850649 on Build GRCh38
rs375884686 Chr.4: 850798-850798 on Build GRCh38
rs748275740 Chr.4: 850885-850885 on Build GRCh38
rs993088278 Chr.4: 850733-850733 on Build GRCh38
rs545466700 Chr.4: 850653-850653 on Build GRCh38
rs759322908 Chr.4: 850898-850898 on Build GRCh38
rs771124847 Chr.4: 850997-850997 on Build GRCh38
rs905703928 Chr.4: 850723-850723 on Build GRCh38
rs201736600 Chr.4: 850725-850725 on Build GRCh38
rs571812839 Chr.4: 850647-850647 on Build GRCh38
rs944496913 Chr.4: 850870-850870 on Build GRCh38
rs752033233 Chr.4: 850929-850929 on Build GRCh38
rs768579483 Chr.4: 851015-851015 on Build GRCh38
rs771131121 Chr.4: 850895-850895 on Build GRCh38
rs753905701 Chr.4: 850767-850767 on Build GRCh38

View Full Product Details