Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 960301-960788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKQ
Entrez Gene ID
1609
Gene Name
diacylglycerol kinase theta
Gene Aliases
DAGK, DAGK4, DAGK7
UniGene
Hs.584858
Gene Chromosome Location
Chr.4: 958884-973560 on Build GRCh38
Transcripts
NM_001347.3 XM_011513411.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4449609 c.2794delG p.D932fs*>11 Chr.4: 960655-960655 on Build GRCh38
4677243 c.2771G>T p.G924V Chr.4: 960678-960678 on Build GRCh38
6643137 c.2793G>A p.A931A Chr.4: 960656-960656 on Build GRCh38
7417520 c.2803C>A p.P935T Chr.4: 960646-960646 on Build GRCh38
6481879 c.2798C>T p.A933V Chr.4: 960651-960651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767253967 Chr.4: 960647-960647 on Build GRCh38
rs35717664 Chr.4: 960542-960543 on Build GRCh38
rs556587375 Chr.4: 960576-960576 on Build GRCh38
rs761857857 Chr.4: 960682-960682 on Build GRCh38
rs967624151 Chr.4: 960465-960465 on Build GRCh38
rs762517736 Chr.4: 960577-960577 on Build GRCh38
rs775270693 Chr.4: 960716-960716 on Build GRCh38
rs919458343 Chr.4: 960611-960611 on Build GRCh38
rs935254800 Chr.4: 960458-960458 on Build GRCh38
rs200371098 Chr.4: 960636-960636 on Build GRCh38
rs373041472 Chr.4: 960525-960525 on Build GRCh38
rs1012921300 Chr.4: 960600-960600 on Build GRCh38
rs376247716 Chr.4: 960656-960656 on Build GRCh38
rs775944734 Chr.4: 960334-960334 on Build GRCh38
rs376731382 Chr.4: 960713-960713 on Build GRCh38
rs1009387732 Chr.4: 960721-960721 on Build GRCh38
rs371649620 Chr.4: 960678-960678 on Build GRCh38
rs3733347 Chr.4: 960489-960489 on Build GRCh38
rs572800869 Chr.4: 960631-960631 on Build GRCh38
rs776434593 Chr.4: 960607-960607 on Build GRCh38
rs868769563 Chr.4: 960690-960690 on Build GRCh38
rs781385965 Chr.4: 960674-960674 on Build GRCh38
rs765918344 Chr.4: 960632-960632 on Build GRCh38
rs768151699 Chr.4: 960616-960621 on Build GRCh38
rs759254355 Chr.4: 960638-960638 on Build GRCh38
rs749278325 Chr.4: 960689-960689 on Build GRCh38
rs367632247 Chr.4: 960590-960590 on Build GRCh38
rs535809420 Chr.4: 960594-960594 on Build GRCh38
rs750717630 Chr.4: 960736-960736 on Build GRCh38
rs113661942 Chr.4: 960488-960488 on Build GRCh38
rs899031748 Chr.4: 960377-960377 on Build GRCh38
rs751725859 Chr.4: 960758-960758 on Build GRCh38
rs777360834 Chr.4: 960680-960680 on Build GRCh38
rs776347949 Chr.4: 960572-960572 on Build GRCh38
rs762121542 Chr.4: 960648-960648 on Build GRCh38
rs758768735 Chr.4: 960657-960657 on Build GRCh38
rs752762706 Chr.4: 960638-960639 on Build GRCh38
rs768513842 Chr.4: 960570-960570 on Build GRCh38
rs915236131 Chr.4: 960466-960466 on Build GRCh38
rs757886916 Chr.4: 960593-960593 on Build GRCh38
rs189127018 Chr.4: 960373-960373 on Build GRCh38
rs752917637 Chr.4: 960764-960764 on Build GRCh38
rs145217127 Chr.4: 960524-960524 on Build GRCh38
rs3733346 Chr.4: 960487-960487 on Build GRCh38
rs773959064 Chr.4: 960700-960700 on Build GRCh38
rs759085232 Chr.4: 960703-960703 on Build GRCh38
rs370905438 Chr.4: 960664-960664 on Build GRCh38
rs1051033191 Chr.4: 960342-960342 on Build GRCh38
rs754520219 Chr.4: 960584-960584 on Build GRCh38
rs759622869 Chr.4: 960508-960508 on Build GRCh38
rs113484716 Chr.4: 960381-960381 on Build GRCh38
rs3733349 Chr.4: 960523-960523 on Build GRCh38
rs771918682 Chr.4: 960710-960710 on Build GRCh38
rs1018019289 Chr.4: 960379-960379 on Build GRCh38
rs3733348 Chr.4: 960510-960510 on Build GRCh38
rs760072238 Chr.4: 960572-960573 on Build GRCh38
rs765701087 Chr.4: 960730-960730 on Build GRCh38
rs756332292 Chr.4: 960756-960757 on Build GRCh38
rs776868936 Chr.4: 960545-960545 on Build GRCh38
rs755244449 Chr.4: 960667-960667 on Build GRCh38
rs766001892 Chr.4: 960581-960581 on Build GRCh38
rs112411212 Chr.4: 960634-960634 on Build GRCh38
rs34415614 Chr.4: 960627-960627 on Build GRCh38
rs778227457 Chr.4: 960567-960567 on Build GRCh38
rs201925522 Chr.4: 960661-960661 on Build GRCh38
rs769587311 Chr.4: 960679-960679 on Build GRCh38
rs867749016 Chr.4: 960471-960471 on Build GRCh38
rs199617751 Chr.4: 960614-960614 on Build GRCh38
rs147173078 Chr.4: 960562-960562 on Build GRCh38
rs922399134 Chr.4: 960444-960444 on Build GRCh38
rs534840904 Chr.4: 960527-960527 on Build GRCh38
rs992505147 Chr.4: 960457-960457 on Build GRCh38
rs760346126 Chr.4: 960718-960718 on Build GRCh38
rs866335008 Chr.4: 960691-960691 on Build GRCh38
rs142985195 Chr.4: 960719-960719 on Build GRCh38
rs769750390 Chr.4: 960435-960436 on Build GRCh38
rs190782037 Chr.4: 960660-960660 on Build GRCh38
rs926693204 Chr.4: 960490-960490 on Build GRCh38
rs934049581 Chr.4: 960504-960504 on Build GRCh38
rs958430367 Chr.4: 960424-960424 on Build GRCh38
rs140396044 Chr.4: 960740-960740 on Build GRCh38
rs939553020 Chr.4: 960496-960496 on Build GRCh38
rs563670132 Chr.4: 960512-960512 on Build GRCh38
rs762265749 Chr.4: 960654-960654 on Build GRCh38
rs571529572 Chr.4: 960546-960546 on Build GRCh38
rs1036320127 Chr.4: 960538-960538 on Build GRCh38
rs898070851 Chr.4: 960323-960323 on Build GRCh38
rs968477892 Chr.4: 960579-960579 on Build GRCh38
rs374750606 Chr.4: 960583-960583 on Build GRCh38
rs386670446 Chr.4: 960487-960489 on Build GRCh38
rs1034126553 Chr.4: 960394-960394 on Build GRCh38
rs764583357 Chr.4: 960587-960587 on Build GRCh38
rs1004734585 Chr.4: 960370-960370 on Build GRCh38
rs373939199 Chr.4: 960598-960598 on Build GRCh38
rs181922370 Chr.4: 960745-960745 on Build GRCh38
rs755035611 Chr.4: 960763-960763 on Build GRCh38
rs561975676 Chr.4: 960395-960395 on Build GRCh38
rs977490591 Chr.4: 960613-960613 on Build GRCh38
rs187117518 Chr.4: 960597-960597 on Build GRCh38
rs769787190 Chr.4: 960624-960624 on Build GRCh38
rs761833369 Chr.4: 960340-960340 on Build GRCh38
rs952377804 Chr.4: 960369-960369 on Build GRCh38
rs3733345 Chr.4: 960459-960459 on Build GRCh38
rs541236371 Chr.4: 960470-960470 on Build GRCh38
rs976568101 Chr.4: 960382-960382 on Build GRCh38
rs181085028 Chr.4: 960539-960539 on Build GRCh38
rs770724079 Chr.4: 960697-960697 on Build GRCh38
rs150778154 Chr.4: 960362-960362 on Build GRCh38
rs1018469628 Chr.4: 960530-960530 on Build GRCh38
rs902915728 Chr.4: 960770-960770 on Build GRCh38
rs993716427 Chr.4: 960511-960511 on Build GRCh38

View Full Product Details