Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 960682-961171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKQ
Entrez Gene ID
1609
Gene Name
diacylglycerol kinase theta
Gene Aliases
DAGK, DAGK4, DAGK7
UniGene
Hs.584858
Gene Chromosome Location
Chr.4: 958884-973560 on Build GRCh38
Transcripts
NM_001347.3 XM_011513411.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6351290 c.2679C>A p.P893P Chr.4: 961097-961097 on Build GRCh38
4159312 c.2657C>T p.P886L Chr.4: 961119-961119 on Build GRCh38
481690 c.2722C>G p.P908A Chr.4: 961054-961054 on Build GRCh38
4746445 c.2693delC p.P898fs*4 Chr.4: 961083-961083 on Build GRCh38
4746444 c.2764_2766delAGG p.R922delR Chr.4: 960683-960685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045580203 Chr.4: 960936-960936 on Build GRCh38
rs950848495 Chr.4: 960972-960972 on Build GRCh38
rs933675807 Chr.4: 961063-961063 on Build GRCh38
rs892204855 Chr.4: 960895-960895 on Build GRCh38
rs1022148034 Chr.4: 960910-960910 on Build GRCh38
rs955575885 Chr.4: 960796-960796 on Build GRCh38
rs996462512 Chr.4: 960959-960959 on Build GRCh38
rs775074273 Chr.4: 961031-961031 on Build GRCh38
rs756990762 Chr.4: 961105-961105 on Build GRCh38
rs775270693 Chr.4: 960716-960716 on Build GRCh38
rs908661134 Chr.4: 960806-960806 on Build GRCh38
rs926632364 Chr.4: 960787-960787 on Build GRCh38
rs764189716 Chr.4: 961100-961100 on Build GRCh38
rs1009387732 Chr.4: 960721-960721 on Build GRCh38
rs376731382 Chr.4: 960713-960713 on Build GRCh38
rs533646879 Chr.4: 960980-960980 on Build GRCh38
rs933642961 Chr.4: 960868-960868 on Build GRCh38
rs892104187 Chr.4: 961086-961086 on Build GRCh38
rs1019327151 Chr.4: 960795-960808 on Build GRCh38
rs141366864 Chr.4: 961119-961119 on Build GRCh38
rs201491669 Chr.4: 961078-961078 on Build GRCh38
rs546816060 Chr.4: 960837-960837 on Build GRCh38
rs775584332 Chr.4: 961144-961144 on Build GRCh38
rs750717630 Chr.4: 960736-960736 on Build GRCh38
rs185767693 Chr.4: 960965-960965 on Build GRCh38
rs760056995 Chr.4: 961139-961139 on Build GRCh38
rs77382558 Chr.4: 961097-961097 on Build GRCh38
rs532303644 Chr.4: 960847-960847 on Build GRCh38
rs751725859 Chr.4: 960758-960758 on Build GRCh38
rs766964182 Chr.4: 961082-961082 on Build GRCh38
rs550448016 Chr.4: 960871-960871 on Build GRCh38
rs760439844 Chr.4: 961046-961046 on Build GRCh38
rs200569018 Chr.4: 961136-961136 on Build GRCh38
rs56039006 Chr.4: 960953-960953 on Build GRCh38
rs752917637 Chr.4: 960764-960764 on Build GRCh38
rs770939341 Chr.4: 961009-961009 on Build GRCh38
rs543054060 Chr.4: 961143-961143 on Build GRCh38
rs756288705 Chr.4: 960773-960773 on Build GRCh38
rs144242490 Chr.4: 960780-960780 on Build GRCh38
rs759085232 Chr.4: 960703-960703 on Build GRCh38
rs919322843 Chr.4: 960797-960797 on Build GRCh38
rs768148689 Chr.4: 961055-961055 on Build GRCh38
rs747776860 Chr.4: 961133-961133 on Build GRCh38
rs754141506 Chr.4: 961102-961102 on Build GRCh38
rs759964942 Chr.4: 961084-961084 on Build GRCh38
rs771918682 Chr.4: 960710-960710 on Build GRCh38
rs920014728 Chr.4: 961022-961022 on Build GRCh38
rs1054133143 Chr.4: 960954-960954 on Build GRCh38
rs745805465 Chr.4: 961118-961118 on Build GRCh38
rs765701087 Chr.4: 960730-960730 on Build GRCh38
rs773677269 Chr.4: 961058-961058 on Build GRCh38
rs756332292 Chr.4: 960756-960757 on Build GRCh38
rs776131078 Chr.4: 961132-961132 on Build GRCh38
rs528537824 Chr.4: 960792-960792 on Build GRCh38
rs949889221 Chr.4: 960928-960928 on Build GRCh38
rs929493733 Chr.4: 960805-960805 on Build GRCh38
rs752723580 Chr.4: 961096-961096 on Build GRCh38
rs535845211 Chr.4: 960955-960955 on Build GRCh38
rs899534662 Chr.4: 960866-960866 on Build GRCh38
rs367975881 Chr.4: 961106-961106 on Build GRCh38
rs35715763 Chr.4: 961012-961012 on Build GRCh38
rs575962681 Chr.4: 961137-961137 on Build GRCh38
rs200300192 Chr.4: 961093-961093 on Build GRCh38
rs199884506 Chr.4: 961010-961010 on Build GRCh38
rs530944465 Chr.4: 960778-960778 on Build GRCh38
rs760346126 Chr.4: 960718-960718 on Build GRCh38
rs554224678 Chr.4: 960960-960960 on Build GRCh38
rs986910646 Chr.4: 960801-960801 on Build GRCh38
rs142985195 Chr.4: 960719-960719 on Build GRCh38
rs919879511 Chr.4: 960848-960848 on Build GRCh38
rs1050855834 Chr.4: 960884-960884 on Build GRCh38
rs770866029 Chr.4: 961141-961141 on Build GRCh38
rs140396044 Chr.4: 960740-960740 on Build GRCh38
rs774849818 Chr.4: 961083-961083 on Build GRCh38
rs984989776 Chr.4: 960807-960807 on Build GRCh38
rs753732553 Chr.4: 961104-961104 on Build GRCh38
rs1009705168 Chr.4: 960902-960902 on Build GRCh38
rs909347297 Chr.4: 960823-960823 on Build GRCh38
rs765672981 Chr.4: 960781-960781 on Build GRCh38
rs974529527 Chr.4: 961019-961019 on Build GRCh38
rs746499591 Chr.4: 961130-961130 on Build GRCh38
rs181922370 Chr.4: 960745-960745 on Build GRCh38
rs779736954 Chr.4: 961128-961128 on Build GRCh38
rs777645203 Chr.4: 960999-960999 on Build GRCh38
rs561436831 Chr.4: 960784-960784 on Build GRCh38
rs960686103 Chr.4: 960791-960791 on Build GRCh38
rs755035611 Chr.4: 960763-960763 on Build GRCh38
rs745431809 Chr.4: 961021-961021 on Build GRCh38
rs571354000 Chr.4: 960838-960838 on Build GRCh38
rs764436507 Chr.4: 961145-961145 on Build GRCh38
rs778131030 Chr.4: 960773-960773 on Build GRCh38
rs902946136 Chr.4: 960948-960948 on Build GRCh38
rs1026244338 Chr.4: 960967-960967 on Build GRCh38
rs553557976 Chr.4: 961004-961004 on Build GRCh38
rs771722313 Chr.4: 961026-961026 on Build GRCh38
rs902915728 Chr.4: 960770-960770 on Build GRCh38
rs903646840 Chr.4: 960926-960926 on Build GRCh38

View Full Product Details