Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1011632-1012167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFRL1
Entrez Gene ID
53834
Gene Name
fibroblast growth factor receptor-like 1
Gene Aliases
FGFR5, FHFR
UniGene
Hs.193326
Gene Chromosome Location
Chr.4: 1011822-1026898 on Build GRCh38
Transcripts
NM_001004356.2 NM_001004358.1 XM_011513486.1

SNP Detail

SNP ID Chromosome Location
rs1055374153 Chr.4: 1012132-1012132 on Build GRCh38
rs939719953 Chr.4: 1011919-1011919 on Build GRCh38
rs960509929 Chr.4: 1012047-1012047 on Build GRCh38
rs908298059 Chr.4: 1012022-1012031 on Build GRCh38
rs1038412278 Chr.4: 1011923-1011923 on Build GRCh38
rs940293579 Chr.4: 1011729-1011729 on Build GRCh38
rs1013706327 Chr.4: 1011960-1011960 on Build GRCh38
rs886766051 Chr.4: 1011707-1011713 on Build GRCh38
rs996710820 Chr.4: 1012001-1012001 on Build GRCh38
rs1000886618 Chr.4: 1011982-1011982 on Build GRCh38
rs977766751 Chr.4: 1011876-1011876 on Build GRCh38
rs1021002074 Chr.4: 1011855-1011855 on Build GRCh38
rs199614217 Chr.4: 1012133-1012133 on Build GRCh38
rs1028141594 Chr.4: 1012014-1012024 on Build GRCh38
rs896576916 Chr.4: 1011840-1011840 on Build GRCh38
rs957487572 Chr.4: 1011898-1011898 on Build GRCh38
rs1033816005 Chr.4: 1011824-1011824 on Build GRCh38
rs1038598118 Chr.4: 1011733-1011733 on Build GRCh38
rs1055342156 Chr.4: 1011948-1011948 on Build GRCh38
rs1002386811 Chr.4: 1011816-1011816 on Build GRCh38
rs896650292 Chr.4: 1011959-1011960 on Build GRCh38
rs927803598 Chr.4: 1012040-1012040 on Build GRCh38
rs866777921 Chr.4: 1011667-1011667 on Build GRCh38
rs868584324 Chr.4: 1011779-1011779 on Build GRCh38
rs990725146 Chr.4: 1012054-1012054 on Build GRCh38
rs973770365 Chr.4: 1012074-1012074 on Build GRCh38
rs918113837 Chr.4: 1012134-1012134 on Build GRCh38
rs966866812 Chr.4: 1011677-1011677 on Build GRCh38
rs916489773 Chr.4: 1012069-1012069 on Build GRCh38
rs919585189 Chr.4: 1012110-1012110 on Build GRCh38
rs986572152 Chr.4: 1012020-1012021 on Build GRCh38
rs977838144 Chr.4: 1011921-1011921 on Build GRCh38
rs938279276 Chr.4: 1012125-1012125 on Build GRCh38
rs996540315 Chr.4: 1011939-1011955 on Build GRCh38
rs966723866 Chr.4: 1011869-1011869 on Build GRCh38
rs902572354 Chr.4: 1011978-1011979 on Build GRCh38
rs933776302 Chr.4: 1011913-1011913 on Build GRCh38
rs955188821 Chr.4: 1012014-1012014 on Build GRCh38
rs921093494 Chr.4: 1011908-1011908 on Build GRCh38
rs961381244 Chr.4: 1012067-1012067 on Build GRCh38
rs1032718321 Chr.4: 1011984-1011984 on Build GRCh38
rs900851026 Chr.4: 1011854-1011875 on Build GRCh38
rs908226689 Chr.4: 1011918-1011918 on Build GRCh38
rs975219128 Chr.4: 1011900-1011900 on Build GRCh38
rs957349862 Chr.4: 1011985-1011985 on Build GRCh38
rs986500099 Chr.4: 1011917-1011917 on Build GRCh38
rs1040947903 Chr.4: 1011787-1011796 on Build GRCh38
rs938225829 Chr.4: 1011942-1011943 on Build GRCh38
rs865846599 Chr.4: 1011881-1011881 on Build GRCh38

View Full Product Details