Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1023989-1024482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFRL1
Entrez Gene ID
53834
Gene Name
fibroblast growth factor receptor-like 1
Gene Aliases
FGFR5, FHFR
UniGene
Hs.193326
Gene Chromosome Location
Chr.4: 1011822-1026898 on Build GRCh38
Transcripts
NM_001004356.2 NM_001004358.1 NM_021923.3 XM_011513486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
332934 c.820G>T p.V274L Chr.4: 1024412-1024412 on Build GRCh38
2951706 c.645G>T p.K215N Chr.4: 1024327-1024327 on Build GRCh38
6663844 c.812G>A p.R271H Chr.4: 1024404-1024404 on Build GRCh38
4560500 c.846G>A p.K282K Chr.4: 1024438-1024438 on Build GRCh38
6663825 c.609C>A p.T203T Chr.4: 1024082-1024082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368039756 Chr.4: 1024331-1024331 on Build GRCh38
rs138109269 Chr.4: 1024456-1024456 on Build GRCh38
rs114935241 Chr.4: 1024330-1024330 on Build GRCh38
rs919553923 Chr.4: 1024168-1024168 on Build GRCh38
rs764556581 Chr.4: 1024377-1024377 on Build GRCh38
rs757215836 Chr.4: 1024344-1024344 on Build GRCh38
rs774266040 Chr.4: 1024017-1024017 on Build GRCh38
rs371754554 Chr.4: 1024372-1024372 on Build GRCh38
rs754793602 Chr.4: 1024359-1024359 on Build GRCh38
rs947346158 Chr.4: 1024249-1024249 on Build GRCh38
rs553859561 Chr.4: 1024273-1024273 on Build GRCh38
rs766259424 Chr.4: 1024409-1024409 on Build GRCh38
rs201667493 Chr.4: 1024059-1024059 on Build GRCh38
rs777318066 Chr.4: 1024439-1024439 on Build GRCh38
rs754138761 Chr.4: 1024274-1024275 on Build GRCh38
rs200852436 Chr.4: 1024457-1024457 on Build GRCh38
rs768355541 Chr.4: 1024262-1024262 on Build GRCh38
rs370485607 Chr.4: 1024294-1024294 on Build GRCh38
rs146608674 Chr.4: 1024060-1024060 on Build GRCh38
rs776300610 Chr.4: 1024314-1024314 on Build GRCh38
rs1047230801 Chr.4: 1024178-1024178 on Build GRCh38
rs540991862 Chr.4: 1024384-1024384 on Build GRCh38
rs747464840 Chr.4: 1024049-1024049 on Build GRCh38
rs773991255 Chr.4: 1024076-1024076 on Build GRCh38
rs755405023 Chr.4: 1024125-1024125 on Build GRCh38
rs773213801 Chr.4: 1024063-1024063 on Build GRCh38
rs372119720 Chr.4: 1024440-1024440 on Build GRCh38
rs777032994 Chr.4: 1024451-1024451 on Build GRCh38
rs543347696 Chr.4: 1024323-1024323 on Build GRCh38
rs747940022 Chr.4: 1024424-1024424 on Build GRCh38
rs745598351 Chr.4: 1024149-1024149 on Build GRCh38
rs540985292 Chr.4: 1024254-1024256 on Build GRCh38
rs115146999 Chr.4: 1024269-1024269 on Build GRCh38
rs982252104 Chr.4: 1024175-1024175 on Build GRCh38
rs538143852 Chr.4: 1024234-1024234 on Build GRCh38
rs769387185 Chr.4: 1024364-1024364 on Build GRCh38
rs1044715701 Chr.4: 1024162-1024162 on Build GRCh38
rs748784356 Chr.4: 1024371-1024371 on Build GRCh38
rs140460823 Chr.4: 1024351-1024351 on Build GRCh38
rs762124519 Chr.4: 1024453-1024453 on Build GRCh38
rs749230766 Chr.4: 1024064-1024064 on Build GRCh38
rs776162191 Chr.4: 1024373-1024373 on Build GRCh38
rs750528701 Chr.4: 1024347-1024347 on Build GRCh38
rs58679007 Chr.4: 1024420-1024420 on Build GRCh38
rs746856237 Chr.4: 1024301-1024301 on Build GRCh38
rs761356781 Chr.4: 1024268-1024268 on Build GRCh38
rs756576255 Chr.4: 1024266-1024266 on Build GRCh38
rs765429291 Chr.4: 1024403-1024403 on Build GRCh38
rs753413026 Chr.4: 1024116-1024116 on Build GRCh38
rs751514315 Chr.4: 1024411-1024411 on Build GRCh38
rs535412527 Chr.4: 1024272-1024272 on Build GRCh38
rs892405098 Chr.4: 1024292-1024292 on Build GRCh38
rs753928771 Chr.4: 1024278-1024278 on Build GRCh38
rs751842953 Chr.4: 1024120-1024120 on Build GRCh38
rs183356176 Chr.4: 1024291-1024291 on Build GRCh38
rs770019120 Chr.4: 1024454-1024454 on Build GRCh38
rs759415964 Chr.4: 1024079-1024079 on Build GRCh38
rs772753740 Chr.4: 1024366-1024366 on Build GRCh38
rs757380523 Chr.4: 1024279-1024279 on Build GRCh38
rs758442194 Chr.4: 1024408-1024408 on Build GRCh38
rs749425365 Chr.4: 1024134-1024134 on Build GRCh38
rs767173391 Chr.4: 1024137-1024152 on Build GRCh38
rs770577862 Chr.4: 1024441-1024441 on Build GRCh38
rs771970929 Chr.4: 1024293-1024293 on Build GRCh38
rs532862696 Chr.4: 1024426-1024426 on Build GRCh38
rs781224592 Chr.4: 1024126-1024126 on Build GRCh38
rs373839707 Chr.4: 1024300-1024300 on Build GRCh38
rs770742296 Chr.4: 1024135-1024135 on Build GRCh38
rs909385227 Chr.4: 1024390-1024390 on Build GRCh38
rs116837147 Chr.4: 1024315-1024315 on Build GRCh38
rs149112646 Chr.4: 1024460-1024460 on Build GRCh38
rs1002275417 Chr.4: 1024311-1024311 on Build GRCh38
rs763601127 Chr.4: 1024115-1024115 on Build GRCh38
rs752969422 Chr.4: 1024129-1024129 on Build GRCh38
rs771584275 Chr.4: 1024085-1024085 on Build GRCh38
rs371198708 Chr.4: 1024271-1024271 on Build GRCh38
rs1052424420 Chr.4: 1024201-1024201 on Build GRCh38
rs893784130 Chr.4: 1024246-1024246 on Build GRCh38
rs752653233 Chr.4: 1024032-1024032 on Build GRCh38
rs769346621 Chr.4: 1024450-1024450 on Build GRCh38
rs768269824 Chr.4: 1024313-1024313 on Build GRCh38
rs758196459 Chr.4: 1024375-1024375 on Build GRCh38
rs759143430 Chr.4: 1024116-1024148 on Build GRCh38
rs148556821 Chr.4: 1024047-1024047 on Build GRCh38
rs752117207 Chr.4: 1024022-1024022 on Build GRCh38
rs552464132 Chr.4: 1024130-1024130 on Build GRCh38
rs935344059 Chr.4: 1024197-1024197 on Build GRCh38
rs931093442 Chr.4: 1024174-1024174 on Build GRCh38
rs747844379 Chr.4: 1024363-1024363 on Build GRCh38
rs754124213 Chr.4: 1024341-1024341 on Build GRCh38
rs776925588 Chr.4: 1024383-1024383 on Build GRCh38
rs765681967 Chr.4: 1024031-1024031 on Build GRCh38
rs756377360 Chr.4: 1024274-1024274 on Build GRCh38
rs780767290 Chr.4: 1024360-1024360 on Build GRCh38
rs755650629 Chr.4: 1024025-1024025 on Build GRCh38
rs549806859 Chr.4: 1024261-1024261 on Build GRCh38
rs778177752 Chr.4: 1024277-1024277 on Build GRCh38
rs370772573 Chr.4: 1024404-1024404 on Build GRCh38
rs759093103 Chr.4: 1024018-1024018 on Build GRCh38
rs766962187 Chr.4: 1024119-1024119 on Build GRCh38
rs144361678 Chr.4: 1024055-1024055 on Build GRCh38
rs564517711 Chr.4: 1024459-1024459 on Build GRCh38
rs943843273 Chr.4: 1024190-1024190 on Build GRCh38
rs571149220 Chr.4: 1024184-1024184 on Build GRCh38
rs928033143 Chr.4: 1024189-1024189 on Build GRCh38
rs768184121 Chr.4: 1024329-1024329 on Build GRCh38
rs757673940 Chr.4: 1024434-1024435 on Build GRCh38
rs760118278 Chr.4: 1024112-1024112 on Build GRCh38
rs771912282 Chr.4: 1024260-1024260 on Build GRCh38
rs746770205 Chr.4: 1024352-1024352 on Build GRCh38
rs767248157 Chr.4: 1024021-1024021 on Build GRCh38
rs754703776 Chr.4: 1024412-1024412 on Build GRCh38
rs565792759 Chr.4: 1024041-1024041 on Build GRCh38
rs775228687 Chr.4: 1024110-1024110 on Build GRCh38
rs763437948 Chr.4: 1024118-1024118 on Build GRCh38
rs764362615 Chr.4: 1024335-1024335 on Build GRCh38
rs919516654 Chr.4: 1024071-1024071 on Build GRCh38
rs778940041 Chr.4: 1024147-1024147 on Build GRCh38
rs143714694 Chr.4: 1024319-1024319 on Build GRCh38
rs777982818 Chr.4: 1024133-1024133 on Build GRCh38
rs868650247 Chr.4: 1024023-1024023 on Build GRCh38
rs565801368 Chr.4: 1024289-1024289 on Build GRCh38
rs765357209 Chr.4: 1024345-1024345 on Build GRCh38
rs376289054 Chr.4: 1024276-1024276 on Build GRCh38
rs771046121 Chr.4: 1024067-1024067 on Build GRCh38
rs909604476 Chr.4: 1024257-1024257 on Build GRCh38
rs780279438 Chr.4: 1024048-1024048 on Build GRCh38
rs935417684 Chr.4: 1024299-1024299 on Build GRCh38
rs376072293 Chr.4: 1024442-1024442 on Build GRCh38

View Full Product Details