Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1024082-1024602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFRL1
Entrez Gene ID
53834
Gene Name
fibroblast growth factor receptor-like 1
Gene Aliases
FGFR5, FHFR
UniGene
Hs.193326
Gene Chromosome Location
Chr.4: 1011822-1026898 on Build GRCh38
Transcripts
NM_001004356.2 NM_001004358.1 NM_021923.3 XM_011513486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
332934 c.820G>T p.V274L Chr.4: 1024412-1024412 on Build GRCh38
2951706 c.645G>T p.K215N Chr.4: 1024327-1024327 on Build GRCh38
6663844 c.812G>A p.R271H Chr.4: 1024404-1024404 on Build GRCh38
4560500 c.846G>A p.K282K Chr.4: 1024438-1024438 on Build GRCh38
325344 c.946G>T p.D316Y Chr.4: 1024538-1024538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368039756 Chr.4: 1024331-1024331 on Build GRCh38
rs138109269 Chr.4: 1024456-1024456 on Build GRCh38
rs114935241 Chr.4: 1024330-1024330 on Build GRCh38
rs919553923 Chr.4: 1024168-1024168 on Build GRCh38
rs764556581 Chr.4: 1024377-1024377 on Build GRCh38
rs756715366 Chr.4: 1024561-1024561 on Build GRCh38
rs759443799 Chr.4: 1024464-1024464 on Build GRCh38
rs757215836 Chr.4: 1024344-1024344 on Build GRCh38
rs371754554 Chr.4: 1024372-1024372 on Build GRCh38
rs753437773 Chr.4: 1024478-1024478 on Build GRCh38
rs745887685 Chr.4: 1024571-1024571 on Build GRCh38
rs754793602 Chr.4: 1024359-1024359 on Build GRCh38
rs947346158 Chr.4: 1024249-1024249 on Build GRCh38
rs553859561 Chr.4: 1024273-1024273 on Build GRCh38
rs766259424 Chr.4: 1024409-1024409 on Build GRCh38
rs754138761 Chr.4: 1024274-1024275 on Build GRCh38
rs777318066 Chr.4: 1024439-1024439 on Build GRCh38
rs200852436 Chr.4: 1024457-1024457 on Build GRCh38
rs768355541 Chr.4: 1024262-1024262 on Build GRCh38
rs370485607 Chr.4: 1024294-1024294 on Build GRCh38
rs147385493 Chr.4: 1024540-1024540 on Build GRCh38
rs774380288 Chr.4: 1024515-1024515 on Build GRCh38
rs776300610 Chr.4: 1024314-1024314 on Build GRCh38
rs1047230801 Chr.4: 1024178-1024178 on Build GRCh38
rs540991862 Chr.4: 1024384-1024384 on Build GRCh38
rs755405023 Chr.4: 1024125-1024125 on Build GRCh38
rs777032994 Chr.4: 1024451-1024451 on Build GRCh38
rs372119720 Chr.4: 1024440-1024440 on Build GRCh38
rs543347696 Chr.4: 1024323-1024323 on Build GRCh38
rs747940022 Chr.4: 1024424-1024424 on Build GRCh38
rs745598351 Chr.4: 1024149-1024149 on Build GRCh38
rs540985292 Chr.4: 1024254-1024256 on Build GRCh38
rs115146999 Chr.4: 1024269-1024269 on Build GRCh38
rs982252104 Chr.4: 1024175-1024175 on Build GRCh38
rs200863947 Chr.4: 1024530-1024530 on Build GRCh38
rs777425704 Chr.4: 1024471-1024471 on Build GRCh38
rs538143852 Chr.4: 1024234-1024234 on Build GRCh38
rs769387185 Chr.4: 1024364-1024364 on Build GRCh38
rs1044715701 Chr.4: 1024162-1024162 on Build GRCh38
rs140460823 Chr.4: 1024351-1024351 on Build GRCh38
rs748784356 Chr.4: 1024371-1024371 on Build GRCh38
rs762124519 Chr.4: 1024453-1024453 on Build GRCh38
rs200153294 Chr.4: 1024574-1024574 on Build GRCh38
rs781684420 Chr.4: 1024549-1024549 on Build GRCh38
rs532080560 Chr.4: 1024481-1024481 on Build GRCh38
rs776162191 Chr.4: 1024373-1024373 on Build GRCh38
rs368634271 Chr.4: 1024579-1024579 on Build GRCh38
rs750528701 Chr.4: 1024347-1024347 on Build GRCh38
rs58679007 Chr.4: 1024420-1024420 on Build GRCh38
rs529265854 Chr.4: 1024523-1024523 on Build GRCh38
rs746856237 Chr.4: 1024301-1024301 on Build GRCh38
rs761356781 Chr.4: 1024268-1024268 on Build GRCh38
rs756576255 Chr.4: 1024266-1024266 on Build GRCh38
rs765429291 Chr.4: 1024403-1024403 on Build GRCh38
rs753413026 Chr.4: 1024116-1024116 on Build GRCh38
rs751514315 Chr.4: 1024411-1024411 on Build GRCh38
rs535412527 Chr.4: 1024272-1024272 on Build GRCh38
rs375942692 Chr.4: 1024516-1024516 on Build GRCh38
rs892405098 Chr.4: 1024292-1024292 on Build GRCh38
rs753928771 Chr.4: 1024278-1024278 on Build GRCh38
rs763160215 Chr.4: 1024511-1024511 on Build GRCh38
rs751842953 Chr.4: 1024120-1024120 on Build GRCh38
rs770019120 Chr.4: 1024454-1024454 on Build GRCh38
rs183356176 Chr.4: 1024291-1024291 on Build GRCh38
rs1026267415 Chr.4: 1024510-1024510 on Build GRCh38
rs772753740 Chr.4: 1024366-1024366 on Build GRCh38
rs757380523 Chr.4: 1024279-1024279 on Build GRCh38
rs749425365 Chr.4: 1024134-1024134 on Build GRCh38
rs758442194 Chr.4: 1024408-1024408 on Build GRCh38
rs767173391 Chr.4: 1024137-1024152 on Build GRCh38
rs770577862 Chr.4: 1024441-1024441 on Build GRCh38
rs761732244 Chr.4: 1024581-1024581 on Build GRCh38
rs760516188 Chr.4: 1024577-1024577 on Build GRCh38
rs771970929 Chr.4: 1024293-1024293 on Build GRCh38
rs532862696 Chr.4: 1024426-1024426 on Build GRCh38
rs781224592 Chr.4: 1024126-1024126 on Build GRCh38
rs373839707 Chr.4: 1024300-1024300 on Build GRCh38
rs770742296 Chr.4: 1024135-1024135 on Build GRCh38
rs909385227 Chr.4: 1024390-1024390 on Build GRCh38
rs116837147 Chr.4: 1024315-1024315 on Build GRCh38
rs149112646 Chr.4: 1024460-1024460 on Build GRCh38
rs1002275417 Chr.4: 1024311-1024311 on Build GRCh38
rs752969422 Chr.4: 1024129-1024129 on Build GRCh38
rs763601127 Chr.4: 1024115-1024115 on Build GRCh38
rs370405185 Chr.4: 1024468-1024468 on Build GRCh38
rs371198708 Chr.4: 1024271-1024271 on Build GRCh38
rs1052424420 Chr.4: 1024201-1024201 on Build GRCh38
rs756949305 Chr.4: 1024536-1024536 on Build GRCh38
rs781580876 Chr.4: 1024486-1024486 on Build GRCh38
rs893784130 Chr.4: 1024246-1024246 on Build GRCh38
rs769346621 Chr.4: 1024450-1024450 on Build GRCh38
rs1013555862 Chr.4: 1024504-1024504 on Build GRCh38
rs768269824 Chr.4: 1024313-1024313 on Build GRCh38
rs758196459 Chr.4: 1024375-1024375 on Build GRCh38
rs759143430 Chr.4: 1024116-1024148 on Build GRCh38
rs935344059 Chr.4: 1024197-1024197 on Build GRCh38
rs552464132 Chr.4: 1024130-1024130 on Build GRCh38
rs931093442 Chr.4: 1024174-1024174 on Build GRCh38
rs374222149 Chr.4: 1024578-1024578 on Build GRCh38
rs754124213 Chr.4: 1024341-1024341 on Build GRCh38
rs747844379 Chr.4: 1024363-1024363 on Build GRCh38
rs776925588 Chr.4: 1024383-1024383 on Build GRCh38
rs756377360 Chr.4: 1024274-1024274 on Build GRCh38
rs778748309 Chr.4: 1024537-1024537 on Build GRCh38
rs757035366 Chr.4: 1024480-1024480 on Build GRCh38
rs780767290 Chr.4: 1024360-1024360 on Build GRCh38
rs767391975 Chr.4: 1024465-1024465 on Build GRCh38
rs771097582 Chr.4: 1024566-1024566 on Build GRCh38
rs748610298 Chr.4: 1024552-1024552 on Build GRCh38
rs549806859 Chr.4: 1024261-1024261 on Build GRCh38
rs147980100 Chr.4: 1024572-1024572 on Build GRCh38
rs141649779 Chr.4: 1024582-1024582 on Build GRCh38
rs778177752 Chr.4: 1024277-1024277 on Build GRCh38
rs370772573 Chr.4: 1024404-1024404 on Build GRCh38
rs766962187 Chr.4: 1024119-1024119 on Build GRCh38
rs564517711 Chr.4: 1024459-1024459 on Build GRCh38
rs202228086 Chr.4: 1024564-1024564 on Build GRCh38
rs943843273 Chr.4: 1024190-1024190 on Build GRCh38
rs571149220 Chr.4: 1024184-1024184 on Build GRCh38
rs568375042 Chr.4: 1024512-1024512 on Build GRCh38
rs768184121 Chr.4: 1024329-1024329 on Build GRCh38
rs928033143 Chr.4: 1024189-1024189 on Build GRCh38
rs757673940 Chr.4: 1024434-1024435 on Build GRCh38
rs760118278 Chr.4: 1024112-1024112 on Build GRCh38
rs771912282 Chr.4: 1024260-1024260 on Build GRCh38
rs746770205 Chr.4: 1024352-1024352 on Build GRCh38
rs773419444 Chr.4: 1024490-1024490 on Build GRCh38
rs372708838 Chr.4: 1024482-1024482 on Build GRCh38
rs752493793 Chr.4: 1024469-1024469 on Build GRCh38
rs142743042 Chr.4: 1024522-1024522 on Build GRCh38
rs754703776 Chr.4: 1024412-1024412 on Build GRCh38
rs775228687 Chr.4: 1024110-1024110 on Build GRCh38
rs370584141 Chr.4: 1024533-1024533 on Build GRCh38
rs749914986 Chr.4: 1024538-1024538 on Build GRCh38
rs763437948 Chr.4: 1024118-1024118 on Build GRCh38
rs138843839 Chr.4: 1024489-1024489 on Build GRCh38
rs764362615 Chr.4: 1024335-1024335 on Build GRCh38
rs778940041 Chr.4: 1024147-1024147 on Build GRCh38
rs143714694 Chr.4: 1024319-1024319 on Build GRCh38
rs4647946 Chr.4: 1024483-1024483 on Build GRCh38
rs777982818 Chr.4: 1024133-1024133 on Build GRCh38
rs565801368 Chr.4: 1024289-1024289 on Build GRCh38
rs765357209 Chr.4: 1024345-1024345 on Build GRCh38
rs376289054 Chr.4: 1024276-1024276 on Build GRCh38
rs755880461 Chr.4: 1024470-1024470 on Build GRCh38
rs909604476 Chr.4: 1024257-1024257 on Build GRCh38
rs774575940 Chr.4: 1024567-1024567 on Build GRCh38
rs935417684 Chr.4: 1024299-1024299 on Build GRCh38
rs376072293 Chr.4: 1024442-1024442 on Build GRCh38
rs750115262 Chr.4: 1024584-1024584 on Build GRCh38

View Full Product Details