Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1657074-1657564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM53A
Entrez Gene ID
152877
Gene Name
family with sequence similarity 53 member A
Gene Aliases
DNTNP
UniGene
Hs.143314
Gene Chromosome Location
Chr.4: 1574055-1684313 on Build GRCh38
Transcripts
NM_001013622.3 NM_001174070.2 NM_001297435.1 XM_005247948.3 XM_011513402.2 XM_011513403.1 XM_011513404.2 XM_011513405.1 XM_017007802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4515721 c.126_127GG>AA p.E43K Chr.4: 1657417-1657418 on Build GRCh38
6314846 c.102C>A p.N34K Chr.4: 1657442-1657442 on Build GRCh38
232298 c.121C>T p.P41S Chr.4: 1657423-1657423 on Build GRCh38
7243780 c.136+7A>G p.? Chr.4: 1657401-1657401 on Build GRCh38
6208811 c.130C>T p.L44F Chr.4: 1657414-1657414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910818026 Chr.4: 1657258-1657258 on Build GRCh38
rs756617129 Chr.4: 1657412-1657412 on Build GRCh38
rs9997609 Chr.4: 1657270-1657270 on Build GRCh38
rs201842677 Chr.4: 1657414-1657414 on Build GRCh38
rs934700374 Chr.4: 1657313-1657313 on Build GRCh38
rs993239826 Chr.4: 1657213-1657213 on Build GRCh38
rs924809807 Chr.4: 1657290-1657290 on Build GRCh38
rs1027243046 Chr.4: 1657231-1657231 on Build GRCh38
rs561359054 Chr.4: 1657207-1657207 on Build GRCh38
rs762892435 Chr.4: 1657479-1657482 on Build GRCh38
rs764920347 Chr.4: 1657445-1657445 on Build GRCh38
rs559781828 Chr.4: 1657104-1657104 on Build GRCh38
rs763982029 Chr.4: 1657397-1657397 on Build GRCh38
rs780163571 Chr.4: 1657501-1657501 on Build GRCh38
rs1018897807 Chr.4: 1657194-1657194 on Build GRCh38
rs535962796 Chr.4: 1657332-1657332 on Build GRCh38
rs1054453131 Chr.4: 1657117-1657117 on Build GRCh38
rs763990800 Chr.4: 1657439-1657439 on Build GRCh38
rs554334549 Chr.4: 1657357-1657357 on Build GRCh38
rs905089464 Chr.4: 1657393-1657393 on Build GRCh38
rs79448086 Chr.4: 1657410-1657410 on Build GRCh38
rs370219018 Chr.4: 1657506-1657506 on Build GRCh38
rs377200630 Chr.4: 1657496-1657496 on Build GRCh38
rs776476580 Chr.4: 1657441-1657441 on Build GRCh38
rs759678696 Chr.4: 1657383-1657383 on Build GRCh38
rs546899562 Chr.4: 1657241-1657241 on Build GRCh38
rs372503770 Chr.4: 1657491-1657491 on Build GRCh38
rs189847803 Chr.4: 1657516-1657516 on Build GRCh38
rs750119361 Chr.4: 1657449-1657449 on Build GRCh38
rs1040973575 Chr.4: 1657095-1657095 on Build GRCh38
rs368312670 Chr.4: 1657475-1657475 on Build GRCh38
rs1027379645 Chr.4: 1657419-1657419 on Build GRCh38
rs191636211 Chr.4: 1657178-1657178 on Build GRCh38
rs1023578128 Chr.4: 1657156-1657156 on Build GRCh38
rs916592683 Chr.4: 1657279-1657279 on Build GRCh38
rs1035286501 Chr.4: 1657192-1657192 on Build GRCh38
rs3860715 Chr.4: 1657435-1657435 on Build GRCh38
rs761771174 Chr.4: 1657442-1657442 on Build GRCh38
rs771189690 Chr.4: 1657364-1657364 on Build GRCh38
rs779059193 Chr.4: 1657495-1657495 on Build GRCh38
rs765045155 Chr.4: 1657402-1657402 on Build GRCh38
rs924765520 Chr.4: 1657472-1657472 on Build GRCh38
rs372584138 Chr.4: 1657507-1657507 on Build GRCh38
rs143720003 Chr.4: 1657455-1657455 on Build GRCh38
rs541726666 Chr.4: 1657096-1657096 on Build GRCh38
rs893152143 Chr.4: 1657134-1657134 on Build GRCh38
rs774398707 Chr.4: 1657371-1657371 on Build GRCh38
rs545677074 Chr.4: 1657432-1657432 on Build GRCh38
rs571445919 Chr.4: 1657250-1657250 on Build GRCh38
rs767377697 Chr.4: 1657384-1657384 on Build GRCh38
rs760340494 Chr.4: 1657389-1657389 on Build GRCh38
rs745891724 Chr.4: 1657433-1657433 on Build GRCh38
rs1010673863 Chr.4: 1657344-1657344 on Build GRCh38
rs781743881 Chr.4: 1657409-1657409 on Build GRCh38
rs951850400 Chr.4: 1657461-1657461 on Build GRCh38
rs549453787 Chr.4: 1657203-1657203 on Build GRCh38
rs2895461 Chr.4: 1657401-1657401 on Build GRCh38
rs749670472 Chr.4: 1657416-1657416 on Build GRCh38
rs762508949 Chr.4: 1657454-1657454 on Build GRCh38
rs935599071 Chr.4: 1657534-1657534 on Build GRCh38
rs951716486 Chr.4: 1657234-1657234 on Build GRCh38
rs751076162 Chr.4: 1657463-1657463 on Build GRCh38
rs528443625 Chr.4: 1657230-1657230 on Build GRCh38
rs545615775 Chr.4: 1657174-1657174 on Build GRCh38
rs754164133 Chr.4: 1657476-1657476 on Build GRCh38
rs961817491 Chr.4: 1657212-1657212 on Build GRCh38
rs550368510 Chr.4: 1657314-1657314 on Build GRCh38
rs756629181 Chr.4: 1657464-1657464 on Build GRCh38
rs934781172 Chr.4: 1657107-1657107 on Build GRCh38
rs568663341 Chr.4: 1657318-1657318 on Build GRCh38
rs893589078 Chr.4: 1657133-1657133 on Build GRCh38
rs771138237 Chr.4: 1657431-1657431 on Build GRCh38
rs769738709 Chr.4: 1657517-1657517 on Build GRCh38
rs971958479 Chr.4: 1657208-1657208 on Build GRCh38
rs189064359 Chr.4: 1657143-1657143 on Build GRCh38
rs745620476 Chr.4: 1657289-1657289 on Build GRCh38
rs1003362966 Chr.4: 1657187-1657187 on Build GRCh38
rs916685971 Chr.4: 1657489-1657489 on Build GRCh38
rs1054548810 Chr.4: 1657317-1657317 on Build GRCh38
rs531038351 Chr.4: 1657202-1657202 on Build GRCh38
rs184143162 Chr.4: 1657437-1657437 on Build GRCh38
rs775511102 Chr.4: 1657386-1657386 on Build GRCh38
rs200002164 Chr.4: 1657408-1657408 on Build GRCh38
rs948715358 Chr.4: 1657331-1657331 on Build GRCh38
rs370249819 Chr.4: 1657436-1657436 on Build GRCh38

View Full Product Details