Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73406494-73406992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALB
Entrez Gene ID
213
Gene Name
albumin
Gene Aliases
ANALBA, FDAH, PRO0883, PRO0903, PRO1341
UniGene
Hs.418167
Gene Chromosome Location
Chr.4: 73404255-73421412 on Build GRCh38
Transcripts
NM_000477.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1619045 c.254_255insT p.C86fs*2 Chr.4: 73406745-73406746 on Build GRCh38
4956408 c.238G>C p.D80H Chr.4: 73406729-73406729 on Build GRCh38
5939197 c.205G>A p.E69K Chr.4: 73406696-73406696 on Build GRCh38
5769810 c.175C>G p.P59A Chr.4: 73406666-73406666 on Build GRCh38
1057397 c.161A>T p.Y54F Chr.4: 73406652-73406652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755945494 Chr.4: 73406759-73406759 on Build GRCh38
rs531857468 Chr.4: 73406833-73406833 on Build GRCh38
rs778723170 Chr.4: 73406880-73406880 on Build GRCh38
rs747493375 Chr.4: 73406626-73406626 on Build GRCh38
rs77050410 Chr.4: 73406741-73406741 on Build GRCh38
rs1002158644 Chr.4: 73406903-73406903 on Build GRCh38
rs1021427237 Chr.4: 73406809-73406809 on Build GRCh38
rs1009214817 Chr.4: 73406893-73406893 on Build GRCh38
rs752917056 Chr.4: 73406769-73406769 on Build GRCh38
rs943798594 Chr.4: 73406546-73406546 on Build GRCh38
rs753746833 Chr.4: 73406719-73406719 on Build GRCh38
rs777843094 Chr.4: 73406773-73406773 on Build GRCh38
rs763937998 Chr.4: 73406677-73406677 on Build GRCh38
rs775319852 Chr.4: 73406682-73406682 on Build GRCh38
rs550326970 Chr.4: 73406933-73406933 on Build GRCh38
rs77892378 Chr.4: 73406760-73406760 on Build GRCh38
rs541871018 Chr.4: 73406548-73406548 on Build GRCh38
rs1041035428 Chr.4: 73406781-73406781 on Build GRCh38
rs746808096 Chr.4: 73406795-73406795 on Build GRCh38
rs770240753 Chr.4: 73406813-73406813 on Build GRCh38
rs961117652 Chr.4: 73406796-73406796 on Build GRCh38
rs777738553 Chr.4: 73406623-73406623 on Build GRCh38
rs1049850142 Chr.4: 73406812-73406812 on Build GRCh38
rs79230429 Chr.4: 73406627-73406627 on Build GRCh38
rs890911697 Chr.4: 73406539-73406539 on Build GRCh38
rs769747342 Chr.4: 73406549-73406549 on Build GRCh38
rs200736287 Chr.4: 73406739-73406739 on Build GRCh38
rs780157233 Chr.4: 73406946-73406946 on Build GRCh38
rs56787531 Chr.4: 73406603-73406603 on Build GRCh38
rs952820899 Chr.4: 73406543-73406543 on Build GRCh38
rs568694816 Chr.4: 73406965-73406965 on Build GRCh38
rs560085213 Chr.4: 73406675-73406675 on Build GRCh38
rs11538221 Chr.4: 73406733-73406733 on Build GRCh38
rs367783945 Chr.4: 73406613-73406613 on Build GRCh38
rs780979109 Chr.4: 73406587-73406587 on Build GRCh38
rs758589275 Chr.4: 73406894-73406894 on Build GRCh38
rs772153212 Chr.4: 73406618-73406618 on Build GRCh38
rs952674971 Chr.4: 73406909-73406909 on Build GRCh38
rs75470261 Chr.4: 73406657-73406657 on Build GRCh38
rs763606901 Chr.4: 73406714-73406714 on Build GRCh38
rs984205914 Chr.4: 73406941-73406941 on Build GRCh38
rs372694396 Chr.4: 73406630-73406630 on Build GRCh38
rs59439476 Chr.4: 73406758-73406758 on Build GRCh38
rs1033518390 Chr.4: 73406907-73406907 on Build GRCh38
rs1053975073 Chr.4: 73406860-73406860 on Build GRCh38
rs545416911 Chr.4: 73406854-73406854 on Build GRCh38
rs966919944 Chr.4: 73406828-73406828 on Build GRCh38
rs1037295127 Chr.4: 73406567-73406567 on Build GRCh38
rs932880975 Chr.4: 73406891-73406891 on Build GRCh38
rs773743765 Chr.4: 73406722-73406722 on Build GRCh38
rs750290382 Chr.4: 73406594-73406594 on Build GRCh38
rs777130511 Chr.4: 73406632-73406632 on Build GRCh38
rs78574148 Chr.4: 73406750-73406750 on Build GRCh38
rs926458737 Chr.4: 73406531-73406531 on Build GRCh38
rs912212748 Chr.4: 73406906-73406906 on Build GRCh38
rs368276725 Chr.4: 73406699-73406699 on Build GRCh38
rs868161245 Chr.4: 73406705-73406705 on Build GRCh38
rs995720395 Chr.4: 73406606-73406606 on Build GRCh38
rs979966514 Chr.4: 73406847-73406852 on Build GRCh38
rs779158168 Chr.4: 73406604-73406604 on Build GRCh38
rs906415383 Chr.4: 73406895-73406895 on Build GRCh38
rs374651285 Chr.4: 73406645-73406645 on Build GRCh38
rs749692149 Chr.4: 73406738-73406738 on Build GRCh38
rs756174669 Chr.4: 73406654-73406654 on Build GRCh38
rs747124517 Chr.4: 73406788-73406788 on Build GRCh38
rs755183143 Chr.4: 73406600-73406600 on Build GRCh38
rs11538229 Chr.4: 73406725-73406725 on Build GRCh38
rs760035423 Chr.4: 73406651-73406651 on Build GRCh38
rs140266237 Chr.4: 73406798-73406798 on Build GRCh38
rs77067638 Chr.4: 73406628-73406628 on Build GRCh38
rs746122361 Chr.4: 73406886-73406886 on Build GRCh38
rs766195754 Chr.4: 73406768-73406768 on Build GRCh38
rs59066571 Chr.4: 73406706-73406706 on Build GRCh38
rs758562878 Chr.4: 73406770-73406770 on Build GRCh38
rs75152012 Chr.4: 73406719-73406720 on Build GRCh38
rs552212414 Chr.4: 73406789-73406789 on Build GRCh38

View Full Product Details