Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74308278-74308793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPGN
Entrez Gene ID
255324
Gene Name
epithelial mitogen
Gene Aliases
ALGV3072, EPG, PRO9904
UniGene
Hs.401237
Gene Chromosome Location
Chr.4: 74307571-74317298 on Build GRCh38
Transcripts
NM_001270989.1 NM_001270990.1 NM_001270991.1 NM_001270992.1 NM_001270993.1 NR_073114.1 NR_073117.1 XM_005265666.4 XM_011531817.2

Gene Symbol
LOC105377276
Entrez Gene ID
105377276
Gene Name
uncharacterized LOC105377276
Gene Aliases
-
UniGene
Hs.662627
Gene Chromosome Location
Chr.4: 74278045-74330322 on Build GRCh38
Transcripts
NM_001270989.1 NM_001270990.1 NM_001270991.1 NM_001270992.1 NM_001270993.1 NR_073114.1 NR_073117.1 XM_005265666.4 XM_011531817.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4937861 c.17C>T p.P6L Chr.4: 74308550-74308550 on Build GRCh38
5826285 c.33T>C p.L11L Chr.4: 74308566-74308566 on Build GRCh38
226970 c.16_17CC>TT p.P6>? Chr.4: 74308549-74308550 on Build GRCh38
1430976 c.5C>T p.A2V Chr.4: 74308538-74308538 on Build GRCh38
3428650 c.33T>C p.L11L Chr.4: 74308566-74308566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013060356 Chr.4: 74308340-74308340 on Build GRCh38
rs750995898 Chr.4: 74308590-74308594 on Build GRCh38
rs774712005 Chr.4: 74308521-74308521 on Build GRCh38
rs963062778 Chr.4: 74308357-74308357 on Build GRCh38
rs76183531 Chr.4: 74308298-74308298 on Build GRCh38
rs761155952 Chr.4: 74308557-74308557 on Build GRCh38
rs538928962 Chr.4: 74308447-74308447 on Build GRCh38
rs1022697594 Chr.4: 74308350-74308350 on Build GRCh38
rs1035971673 Chr.4: 74308306-74308306 on Build GRCh38
rs762420877 Chr.4: 74308537-74308539 on Build GRCh38
rs773647342 Chr.4: 74308554-74308554 on Build GRCh38
rs754415961 Chr.4: 74308563-74308563 on Build GRCh38
rs1020339386 Chr.4: 74308396-74308396 on Build GRCh38
rs200884976 Chr.4: 74308583-74308583 on Build GRCh38
rs1019174955 Chr.4: 74308737-74308737 on Build GRCh38
rs753596161 Chr.4: 74308562-74308562 on Build GRCh38
rs994508563 Chr.4: 74308629-74308629 on Build GRCh38
rs994500360 Chr.4: 74308364-74308364 on Build GRCh38
rs1025952476 Chr.4: 74308673-74308673 on Build GRCh38
rs138592325 Chr.4: 74308608-74308608 on Build GRCh38
rs372137371 Chr.4: 74308488-74308488 on Build GRCh38
rs757089688 Chr.4: 74308595-74308595 on Build GRCh38
rs953494445 Chr.4: 74308744-74308744 on Build GRCh38
rs867031720 Chr.4: 74308531-74308531 on Build GRCh38
rs969214014 Chr.4: 74308400-74308400 on Build GRCh38
rs768816494 Chr.4: 74308520-74308520 on Build GRCh38
rs754704540 Chr.4: 74308613-74308613 on Build GRCh38
rs1036156939 Chr.4: 74308478-74308478 on Build GRCh38
rs62312546 Chr.4: 74308665-74308665 on Build GRCh38
rs757765926 Chr.4: 74308374-74308374 on Build GRCh38
rs1002214153 Chr.4: 74308300-74308300 on Build GRCh38
rs1044712917 Chr.4: 74308539-74308539 on Build GRCh38
rs72654838 Chr.4: 74308765-74308765 on Build GRCh38
rs960183632 Chr.4: 74308359-74308359 on Build GRCh38
rs768608578 Chr.4: 74308618-74308618 on Build GRCh38
rs371391618 Chr.4: 74308363-74308363 on Build GRCh38
rs765885233 Chr.4: 74308559-74308562 on Build GRCh38
rs377304390 Chr.4: 74308630-74308630 on Build GRCh38
rs895571794 Chr.4: 74308299-74308299 on Build GRCh38
rs374777351 Chr.4: 74308559-74308559 on Build GRCh38
rs750361240 Chr.4: 74308496-74308498 on Build GRCh38
rs746754153 Chr.4: 74308593-74308593 on Build GRCh38
rs774435887 Chr.4: 74308627-74308627 on Build GRCh38
rs79024984 Chr.4: 74308611-74308611 on Build GRCh38
rs911841960 Chr.4: 74308476-74308476 on Build GRCh38
rs1044205533 Chr.4: 74308609-74308609 on Build GRCh38
rs1026600137 Chr.4: 74308421-74308421 on Build GRCh38
rs749344493 Chr.4: 74308501-74308501 on Build GRCh38
rs987336500 Chr.4: 74308466-74308466 on Build GRCh38
rs770651717 Chr.4: 74308677-74308677 on Build GRCh38
rs1010444809 Chr.4: 74308394-74308394 on Build GRCh38
rs767927586 Chr.4: 74308552-74308552 on Build GRCh38
rs775708748 Chr.4: 74308492-74308492 on Build GRCh38
rs762444180 Chr.4: 74308522-74308522 on Build GRCh38
rs144683986 Chr.4: 74308575-74308575 on Build GRCh38
rs150076616 Chr.4: 74308430-74308430 on Build GRCh38
rs964837339 Chr.4: 74308477-74308477 on Build GRCh38
rs557170647 Chr.4: 74308457-74308457 on Build GRCh38
rs781434693 Chr.4: 74308576-74308576 on Build GRCh38
rs780036476 Chr.4: 74308487-74308487 on Build GRCh38
rs191289578 Chr.4: 74308700-74308700 on Build GRCh38
rs758970981 Chr.4: 74308429-74308429 on Build GRCh38

View Full Product Details