Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74311952-74312448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPGN
Entrez Gene ID
255324
Gene Name
epithelial mitogen
Gene Aliases
ALGV3072, EPG, PRO9904
UniGene
Hs.401237
Gene Chromosome Location
Chr.4: 74307571-74317298 on Build GRCh38
Transcripts
NM_001270989.1 NM_001270990.1 NM_001270991.1 NM_001270992.1 NM_001270993.1 NR_073114.1 NR_073117.1 XM_005265666.4 XM_011531817.2

Gene Symbol
LOC105377276
Entrez Gene ID
105377276
Gene Name
uncharacterized LOC105377276
Gene Aliases
-
UniGene
Hs.662627
Gene Chromosome Location
Chr.4: 74278045-74330322 on Build GRCh38
Transcripts
NM_001270989.1 NM_001270990.1 NM_001270991.1 NM_001270992.1 NM_001270993.1 NR_073114.1 NR_073117.1 XM_005265666.4 XM_011531817.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6654867 c.179T>C p.I60T Chr.4: 74312257-74312257 on Build GRCh38
1619063 c.157_158insA p.D54fs*3 Chr.4: 74312235-74312236 on Build GRCh38
1642518 c.206A>G p.E69G Chr.4: 74312284-74312284 on Build GRCh38
214085 c.122G>A p.G41E Chr.4: 74312200-74312200 on Build GRCh38
4597818 c.180C>G p.I60M Chr.4: 74312258-74312258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746220058 Chr.4: 74312139-74312139 on Build GRCh38
rs989831901 Chr.4: 74312090-74312090 on Build GRCh38
rs760589727 Chr.4: 74312289-74312289 on Build GRCh38
rs200559518 Chr.4: 74312246-74312246 on Build GRCh38
rs985438940 Chr.4: 74312011-74312011 on Build GRCh38
rs768989140 Chr.4: 74312138-74312138 on Build GRCh38
rs982931080 Chr.4: 74311981-74311984 on Build GRCh38
rs780222225 Chr.4: 74312252-74312252 on Build GRCh38
rs771610784 Chr.4: 74312269-74312269 on Build GRCh38
rs751639068 Chr.4: 74312239-74312239 on Build GRCh38
rs367967851 Chr.4: 74312352-74312352 on Build GRCh38
rs777934643 Chr.4: 74312040-74312040 on Build GRCh38
rs913809256 Chr.4: 74312128-74312128 on Build GRCh38
rs773787972 Chr.4: 74312191-74312191 on Build GRCh38
rs991131732 Chr.4: 74312385-74312385 on Build GRCh38
rs760488798 Chr.4: 74312201-74312201 on Build GRCh38
rs1007850807 Chr.4: 74312115-74312115 on Build GRCh38
rs564670597 Chr.4: 74312262-74312262 on Build GRCh38
rs949731150 Chr.4: 74312408-74312408 on Build GRCh38
rs554123160 Chr.4: 74312063-74312063 on Build GRCh38
rs541887508 Chr.4: 74311990-74311990 on Build GRCh38
rs758158040 Chr.4: 74312425-74312425 on Build GRCh38
rs1055477723 Chr.4: 74312057-74312057 on Build GRCh38
rs925931921 Chr.4: 74312021-74312021 on Build GRCh38
rs913810201 Chr.4: 74312410-74312410 on Build GRCh38
rs915563923 Chr.4: 74312402-74312402 on Build GRCh38
rs948304592 Chr.4: 74312141-74312141 on Build GRCh38
rs774809448 Chr.4: 74312140-74312140 on Build GRCh38
rs572815446 Chr.4: 74312112-74312112 on Build GRCh38
rs766155803 Chr.4: 74312205-74312205 on Build GRCh38
rs762567723 Chr.4: 74312179-74312179 on Build GRCh38
rs377708890 Chr.4: 74312223-74312223 on Build GRCh38
rs999735216 Chr.4: 74312192-74312192 on Build GRCh38
rs976624612 Chr.4: 74312276-74312276 on Build GRCh38
rs897365737 Chr.4: 74312237-74312237 on Build GRCh38
rs201296634 Chr.4: 74312352-74312352 on Build GRCh38
rs759425587 Chr.4: 74312325-74312325 on Build GRCh38
rs1033044027 Chr.4: 74312295-74312295 on Build GRCh38
rs951287142 Chr.4: 74312302-74312302 on Build GRCh38
rs757459599 Chr.4: 74312241-74312241 on Build GRCh38
rs781699681 Chr.4: 74312242-74312242 on Build GRCh38
rs957401534 Chr.4: 74312311-74312311 on Build GRCh38
rs201736074 Chr.4: 74312261-74312261 on Build GRCh38
rs945241407 Chr.4: 74312146-74312146 on Build GRCh38
rs771292324 Chr.4: 74311989-74311991 on Build GRCh38
rs935984535 Chr.4: 74312059-74312059 on Build GRCh38
rs368486590 Chr.4: 74312258-74312258 on Build GRCh38
rs895372985 Chr.4: 74312092-74312092 on Build GRCh38
rs767547212 Chr.4: 74312345-74312345 on Build GRCh38
rs868302602 Chr.4: 74312225-74312225 on Build GRCh38
rs575917871 Chr.4: 74312104-74312104 on Build GRCh38
rs149098980 Chr.4: 74312314-74312314 on Build GRCh38
rs775201768 Chr.4: 74312163-74312163 on Build GRCh38
rs1046706067 Chr.4: 74312153-74312153 on Build GRCh38
rs757404341 Chr.4: 74312338-74312338 on Build GRCh38
rs371408244 Chr.4: 74312234-74312234 on Build GRCh38
rs147639214 Chr.4: 74312288-74312288 on Build GRCh38
rs751869673 Chr.4: 74312336-74312336 on Build GRCh38
rs1046188120 Chr.4: 74312159-74312159 on Build GRCh38
rs940985233 Chr.4: 74312032-74312032 on Build GRCh38
rs768306672 Chr.4: 74312185-74312185 on Build GRCh38
rs374832891 Chr.4: 74312335-74312335 on Build GRCh38
rs750380039 Chr.4: 74312200-74312200 on Build GRCh38
rs201278786 Chr.4: 74312194-74312194 on Build GRCh38
rs758636783 Chr.4: 74312226-74312226 on Build GRCh38
rs927704149 Chr.4: 74312168-74312168 on Build GRCh38
rs779344447 Chr.4: 74312146-74312147 on Build GRCh38
rs768259628 Chr.4: 74312260-74312260 on Build GRCh38
rs937721881 Chr.4: 74312180-74312180 on Build GRCh38
rs767396373 Chr.4: 74312195-74312195 on Build GRCh38
rs766354884 Chr.4: 74312303-74312303 on Build GRCh38
rs770092400 Chr.4: 74312155-74312155 on Build GRCh38
rs546515255 Chr.4: 74312230-74312230 on Build GRCh38

View Full Product Details