Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 75577095-75577633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL2
Entrez Gene ID
8999
Gene Name
cyclin dependent kinase like 2
Gene Aliases
KKIAMRE, P56
UniGene
Hs.591698
Gene Chromosome Location
Chr.4: 75576494-75630537 on Build GRCh38
Transcripts
NM_003948.3 XM_005263335.2 XM_006714406.2 XM_017008809.1 XM_017008810.1 XM_017008811.1

SNP Detail

SNP ID Chromosome Location
rs776445643 Chr.4: 75577323-75577323 on Build GRCh38
rs1052740064 Chr.4: 75577228-75577228 on Build GRCh38
rs763427249 Chr.4: 75577524-75577524 on Build GRCh38
rs77002343 Chr.4: 75577240-75577240 on Build GRCh38
rs1004262579 Chr.4: 75577599-75577599 on Build GRCh38
rs1023458809 Chr.4: 75577252-75577252 on Build GRCh38
rs930518353 Chr.4: 75577301-75577301 on Build GRCh38
rs138541328 Chr.4: 75577547-75577547 on Build GRCh38
rs372485332 Chr.4: 75577142-75577142 on Build GRCh38
rs773746291 Chr.4: 75577518-75577518 on Build GRCh38
rs961734771 Chr.4: 75577539-75577539 on Build GRCh38
rs533840739 Chr.4: 75577275-75577275 on Build GRCh38
rs574145928 Chr.4: 75577470-75577470 on Build GRCh38
rs904128953 Chr.4: 75577401-75577401 on Build GRCh38
rs925391189 Chr.4: 75577449-75577449 on Build GRCh38
rs193014888 Chr.4: 75577578-75577578 on Build GRCh38
rs951840594 Chr.4: 75577585-75577585 on Build GRCh38
rs1026594644 Chr.4: 75577567-75577567 on Build GRCh38
rs535183914 Chr.4: 75577508-75577508 on Build GRCh38
rs1001103158 Chr.4: 75577409-75577409 on Build GRCh38
rs926300189 Chr.4: 75577339-75577339 on Build GRCh38
rs4859536 Chr.4: 75577242-75577242 on Build GRCh38
rs185297225 Chr.4: 75577605-75577605 on Build GRCh38
rs1027116667 Chr.4: 75577579-75577579 on Build GRCh38
rs1042928872 Chr.4: 75577391-75577391 on Build GRCh38
rs904065271 Chr.4: 75577158-75577158 on Build GRCh38
rs552473151 Chr.4: 75577318-75577318 on Build GRCh38
rs935302231 Chr.4: 75577461-75577461 on Build GRCh38
rs191187309 Chr.4: 75577173-75577173 on Build GRCh38
rs551078948 Chr.4: 75577121-75577121 on Build GRCh38
rs951176072 Chr.4: 75577587-75577587 on Build GRCh38
rs147077226 Chr.4: 75577387-75577387 on Build GRCh38
rs919198939 Chr.4: 75577280-75577280 on Build GRCh38
rs995661350 Chr.4: 75577269-75577269 on Build GRCh38
rs896190273 Chr.4: 75577363-75577363 on Build GRCh38
rs1044193730 Chr.4: 75577410-75577410 on Build GRCh38
rs896513110 Chr.4: 75577507-75577507 on Build GRCh38
rs937659395 Chr.4: 75577347-75577347 on Build GRCh38
rs867758526 Chr.4: 75577556-75577556 on Build GRCh38
rs986884273 Chr.4: 75577243-75577243 on Build GRCh38
rs768366190 Chr.4: 75577303-75577303 on Build GRCh38
rs984745060 Chr.4: 75577607-75577607 on Build GRCh38
rs114094350 Chr.4: 75577351-75577351 on Build GRCh38
rs184183472 Chr.4: 75577258-75577258 on Build GRCh38
rs922086463 Chr.4: 75577361-75577361 on Build GRCh38
rs965911832 Chr.4: 75577259-75577259 on Build GRCh38
rs189108276 Chr.4: 75577322-75577322 on Build GRCh38
rs1052455798 Chr.4: 75577487-75577487 on Build GRCh38
rs1000842030 Chr.4: 75577171-75577171 on Build GRCh38
rs1026769033 Chr.4: 75577281-75577281 on Build GRCh38
rs375764535 Chr.4: 75577151-75577151 on Build GRCh38
rs868020153 Chr.4: 75577525-75577525 on Build GRCh38
rs889816686 Chr.4: 75577521-75577521 on Build GRCh38
rs746503021 Chr.4: 75577251-75577252 on Build GRCh38
rs753312449 Chr.4: 75577501-75577502 on Build GRCh38
rs1028514917 Chr.4: 75577453-75577453 on Build GRCh38
rs1871114 Chr.4: 75577538-75577538 on Build GRCh38

View Full Product Details