Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76007307-76007821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL9
Entrez Gene ID
4283
Gene Name
C-X-C motif chemokine ligand 9
Gene Aliases
CMK, Humig, MIG, SCYB9, crg-10
UniGene
Hs.77367
Gene Chromosome Location
Chr.4: 76001342-76007523 on Build GRCh38
Transcripts
NM_002416.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7131865 c.19C>G p.L7V Chr.4: 76007431-76007431 on Build GRCh38
6469684 c.63A>G p.Q21Q Chr.4: 76007387-76007387 on Build GRCh38
3605904 c.30G>T p.L10F Chr.4: 76007420-76007420 on Build GRCh38
6638457 c.38T>A p.I13N Chr.4: 76007412-76007412 on Build GRCh38
3605905 c.24C>T p.F8F Chr.4: 76007426-76007426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765556357 Chr.4: 76007466-76007466 on Build GRCh38
rs2276885 Chr.4: 76007576-76007576 on Build GRCh38
rs529239167 Chr.4: 76007442-76007442 on Build GRCh38
rs746469634 Chr.4: 76007381-76007381 on Build GRCh38
rs569566593 Chr.4: 76007365-76007365 on Build GRCh38
rs180882661 Chr.4: 76007770-76007770 on Build GRCh38
rs915945861 Chr.4: 76007550-76007550 on Build GRCh38
rs899537561 Chr.4: 76007746-76007746 on Build GRCh38
rs755178145 Chr.4: 76007351-76007352 on Build GRCh38
rs948811107 Chr.4: 76007564-76007564 on Build GRCh38
rs371607962 Chr.4: 76007518-76007518 on Build GRCh38
rs868227269 Chr.4: 76007767-76007767 on Build GRCh38
rs1019284317 Chr.4: 76007680-76007680 on Build GRCh38
rs767022882 Chr.4: 76007454-76007454 on Build GRCh38
rs899293023 Chr.4: 76007577-76007577 on Build GRCh38
rs753097276 Chr.4: 76007467-76007467 on Build GRCh38
rs758004149 Chr.4: 76007490-76007490 on Build GRCh38
rs916955354 Chr.4: 76007618-76007618 on Build GRCh38
rs35322408 Chr.4: 76007731-76007731 on Build GRCh38
rs747909938 Chr.4: 76007461-76007461 on Build GRCh38
rs770078590 Chr.4: 76007725-76007725 on Build GRCh38
rs1057046270 Chr.4: 76007520-76007520 on Build GRCh38
rs1009183939 Chr.4: 76007648-76007648 on Build GRCh38
rs115820833 Chr.4: 76007610-76007610 on Build GRCh38
rs966026034 Chr.4: 76007509-76007509 on Build GRCh38
rs1019000835 Chr.4: 76007794-76007794 on Build GRCh38
rs909702730 Chr.4: 76007559-76007559 on Build GRCh38
rs769528250 Chr.4: 76007405-76007405 on Build GRCh38
rs770862403 Chr.4: 76007388-76007388 on Build GRCh38
rs968583779 Chr.4: 76007352-76007352 on Build GRCh38
rs953418741 Chr.4: 76007349-76007349 on Build GRCh38
rs752643352 Chr.4: 76007355-76007355 on Build GRCh38
rs548781775 Chr.4: 76007789-76007789 on Build GRCh38
rs949705021 Chr.4: 76007626-76007626 on Build GRCh38
rs536469764 Chr.4: 76007449-76007449 on Build GRCh38
rs918478134 Chr.4: 76007513-76007513 on Build GRCh38
rs762009700 Chr.4: 76007409-76007409 on Build GRCh38
rs767765394 Chr.4: 76007412-76007412 on Build GRCh38
rs755486724 Chr.4: 76007464-76007464 on Build GRCh38
rs778647490 Chr.4: 76007361-76007361 on Build GRCh38
rs766932866 Chr.4: 76007341-76007341 on Build GRCh38
rs776514039 Chr.4: 76007404-76007404 on Build GRCh38
rs531081695 Chr.4: 76007754-76007754 on Build GRCh38
rs777209756 Chr.4: 76007380-76007380 on Build GRCh38
rs1018984190 Chr.4: 76007461-76007461 on Build GRCh38
rs754344963 Chr.4: 76007457-76007457 on Build GRCh38
rs777816216 Chr.4: 76007732-76007732 on Build GRCh38
rs376172940 Chr.4: 76007356-76007356 on Build GRCh38
rs189282200 Chr.4: 76007750-76007750 on Build GRCh38
rs564923591 Chr.4: 76007632-76007632 on Build GRCh38
rs745996350 Chr.4: 76007749-76007749 on Build GRCh38
rs868868411 Chr.4: 76007458-76007458 on Build GRCh38
rs977078118 Chr.4: 76007511-76007511 on Build GRCh38
rs775325063 Chr.4: 76007407-76007407 on Build GRCh38
rs371643297 Chr.4: 76007777-76007777 on Build GRCh38
rs561283727 Chr.4: 76007795-76007795 on Build GRCh38
rs780860302 Chr.4: 76007451-76007451 on Build GRCh38
rs139049627 Chr.4: 76007527-76007527 on Build GRCh38
rs968932548 Chr.4: 76007773-76007773 on Build GRCh38
rs576883313 Chr.4: 76007638-76007638 on Build GRCh38
rs758033522 Chr.4: 76007375-76007375 on Build GRCh38
rs545748270 Chr.4: 76007730-76007730 on Build GRCh38

View Full Product Details