Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76021421-76021920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART3
Entrez Gene ID
419
Gene Name
ADP-ribosyltransferase 3
Gene Aliases
ARTC3
UniGene
Hs.24976
Gene Chromosome Location
Chr.4: 76011180-76112802 on Build GRCh38
Transcripts
NM_001130017.2 NM_001565.3

Gene Symbol
CXCL10
Entrez Gene ID
3627
Gene Name
C-X-C motif chemokine ligand 10
Gene Aliases
C7, IFI10, INP10, IP-10, SCYB10, crg-2, gIP-10, mob-1
UniGene
Hs.632586
Gene Chromosome Location
Chr.4: 76021116-76023536 on Build GRCh38
Transcripts
NM_001130017.2 NM_001565.3

SNP Detail

SNP ID Chromosome Location
rs34836828 Chr.4: 76021609-76021609 on Build GRCh38
rs925811426 Chr.4: 76021452-76021452 on Build GRCh38
rs988752539 Chr.4: 76021506-76021506 on Build GRCh38
rs891597826 Chr.4: 76021518-76021518 on Build GRCh38
rs1049209628 Chr.4: 76021681-76021681 on Build GRCh38
rs182871280 Chr.4: 76021511-76021511 on Build GRCh38
rs902101671 Chr.4: 76021759-76021759 on Build GRCh38
rs987709095 Chr.4: 76021633-76021633 on Build GRCh38
rs747913501 Chr.4: 76021829-76021829 on Build GRCh38
rs988655671 Chr.4: 76021857-76021857 on Build GRCh38
rs768874782 Chr.4: 76021812-76021812 on Build GRCh38
rs572732596 Chr.4: 76021734-76021734 on Build GRCh38
rs968427234 Chr.4: 76021559-76021559 on Build GRCh38
rs750469707 Chr.4: 76021724-76021724 on Build GRCh38
rs891687993 Chr.4: 76021819-76021819 on Build GRCh38
rs1057024366 Chr.4: 76021800-76021800 on Build GRCh38
rs768569309 Chr.4: 76021565-76021565 on Build GRCh38
rs546563216 Chr.4: 76021474-76021474 on Build GRCh38
rs141971505 Chr.4: 76021843-76021843 on Build GRCh38
rs58658570 Chr.4: 76021608-76021608 on Build GRCh38
rs765410135 Chr.4: 76021608-76021609 on Build GRCh38
rs35795399 Chr.4: 76021524-76021524 on Build GRCh38
rs1002751668 Chr.4: 76021479-76021479 on Build GRCh38
rs542690649 Chr.4: 76021893-76021893 on Build GRCh38
rs1028448445 Chr.4: 76021792-76021792 on Build GRCh38
rs373833272 Chr.4: 76021709-76021709 on Build GRCh38
rs1045078030 Chr.4: 76021456-76021456 on Build GRCh38
rs953813423 Chr.4: 76021807-76021807 on Build GRCh38
rs370049710 Chr.4: 76021881-76021881 on Build GRCh38
rs187517470 Chr.4: 76021629-76021629 on Build GRCh38
rs1021353261 Chr.4: 76021548-76021548 on Build GRCh38
rs920709283 Chr.4: 76021720-76021720 on Build GRCh38
rs879068672 Chr.4: 76021838-76021838 on Build GRCh38
rs3921 Chr.4: 76021790-76021790 on Build GRCh38
rs191522507 Chr.4: 76021448-76021448 on Build GRCh38
rs1005072012 Chr.4: 76021700-76021700 on Build GRCh38
rs999975208 Chr.4: 76021808-76021808 on Build GRCh38
rs905908311 Chr.4: 76021644-76021644 on Build GRCh38
rs372556969 Chr.4: 76021885-76021885 on Build GRCh38
rs60445587 Chr.4: 76021609-76021609 on Build GRCh38
rs578163076 Chr.4: 76021834-76021834 on Build GRCh38
rs569014097 Chr.4: 76021659-76021659 on Build GRCh38
rs147499398 Chr.4: 76021670-76021670 on Build GRCh38
rs906499395 Chr.4: 76021778-76021778 on Build GRCh38
rs886636633 Chr.4: 76021689-76021689 on Build GRCh38
rs974414524 Chr.4: 76021584-76021584 on Build GRCh38
rs530961534 Chr.4: 76021892-76021892 on Build GRCh38
rs148141229 Chr.4: 76021677-76021677 on Build GRCh38
rs767501324 Chr.4: 76021696-76021696 on Build GRCh38
rs553410352 Chr.4: 76021820-76021820 on Build GRCh38
rs193269641 Chr.4: 76021860-76021860 on Build GRCh38
rs751103662 Chr.4: 76021887-76021887 on Build GRCh38
rs973552099 Chr.4: 76021718-76021718 on Build GRCh38

View Full Product Details