Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 77158442-77158937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNG2
Entrez Gene ID
901
Gene Name
cyclin G2
Gene Aliases
-
UniGene
Hs.740456
Gene Chromosome Location
Chr.4: 77157204-77170060 on Build GRCh38
Transcripts
NM_004354.2 XM_011532398.1 XM_011532399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7233933 c.87G>A p.E29E Chr.4: 77158619-77158619 on Build GRCh38
5550405 c.17C>T p.A6V Chr.4: 77158549-77158549 on Build GRCh38
3714948 c.21G>A p.E7E Chr.4: 77158553-77158553 on Build GRCh38
7233934 c.87G>A p.E29E Chr.4: 77158619-77158619 on Build GRCh38
481534 c.71A>G p.Y24C Chr.4: 77158603-77158603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754545084 Chr.4: 77158676-77158676 on Build GRCh38
rs4150054 Chr.4: 77158751-77158751 on Build GRCh38
rs1015381872 Chr.4: 77158728-77158728 on Build GRCh38
rs1037944286 Chr.4: 77158561-77158561 on Build GRCh38
rs957015331 Chr.4: 77158915-77158915 on Build GRCh38
rs767932428 Chr.4: 77158643-77158643 on Build GRCh38
rs778512926 Chr.4: 77158680-77158680 on Build GRCh38
rs376667218 Chr.4: 77158700-77158700 on Build GRCh38
rs890785645 Chr.4: 77158846-77158846 on Build GRCh38
rs200354808 Chr.4: 77158698-77158698 on Build GRCh38
rs374959105 Chr.4: 77158586-77158586 on Build GRCh38
rs761644165 Chr.4: 77158600-77158600 on Build GRCh38
rs759620752 Chr.4: 77158509-77158509 on Build GRCh38
rs755836994 Chr.4: 77158646-77158646 on Build GRCh38
rs765346917 Chr.4: 77158612-77158612 on Build GRCh38
rs775577155 Chr.4: 77158717-77158717 on Build GRCh38
rs757745413 Chr.4: 77158629-77158629 on Build GRCh38
rs185302259 Chr.4: 77158858-77158858 on Build GRCh38
rs1040080118 Chr.4: 77158815-77158815 on Build GRCh38
rs756729821 Chr.4: 77158489-77158489 on Build GRCh38
rs879560351 Chr.4: 77158664-77158664 on Build GRCh38
rs892588521 Chr.4: 77158793-77158793 on Build GRCh38
rs908017303 Chr.4: 77158527-77158527 on Build GRCh38
rs535933836 Chr.4: 77158781-77158781 on Build GRCh38
rs948126191 Chr.4: 77158607-77158607 on Build GRCh38
rs1052461776 Chr.4: 77158792-77158792 on Build GRCh38
rs4150050 Chr.4: 77158542-77158542 on Build GRCh38
rs529555770 Chr.4: 77158905-77158905 on Build GRCh38
rs752716488 Chr.4: 77158520-77158520 on Build GRCh38
rs1048862972 Chr.4: 77158795-77158795 on Build GRCh38
rs4150052 Chr.4: 77158619-77158619 on Build GRCh38
rs745588311 Chr.4: 77158569-77158569 on Build GRCh38
rs981273956 Chr.4: 77158754-77158754 on Build GRCh38
rs541668747 Chr.4: 77158474-77158474 on Build GRCh38
rs564627222 Chr.4: 77158494-77158494 on Build GRCh38
rs768166341 Chr.4: 77158581-77158581 on Build GRCh38
rs955114846 Chr.4: 77158480-77158480 on Build GRCh38
rs764688581 Chr.4: 77158522-77158523 on Build GRCh38
rs770718337 Chr.4: 77158702-77158702 on Build GRCh38
rs745455684 Chr.4: 77158482-77158482 on Build GRCh38
rs1050190863 Chr.4: 77158854-77158854 on Build GRCh38
rs539214084 Chr.4: 77158740-77158740 on Build GRCh38
rs560094130 Chr.4: 77158730-77158730 on Build GRCh38
rs768443085 Chr.4: 77158496-77158496 on Build GRCh38
rs773102024 Chr.4: 77158699-77158699 on Build GRCh38
rs4150053 Chr.4: 77158698-77158699 on Build GRCh38
rs371620068 Chr.4: 77158575-77158575 on Build GRCh38
rs979571921 Chr.4: 77158723-77158723 on Build GRCh38
rs139604990 Chr.4: 77158801-77158801 on Build GRCh38
rs570293932 Chr.4: 77158547-77158547 on Build GRCh38
rs564765302 Chr.4: 77158822-77158822 on Build GRCh38
rs544153561 Chr.4: 77158836-77158836 on Build GRCh38
rs570550357 Chr.4: 77158727-77158728 on Build GRCh38
rs754120290 Chr.4: 77158504-77158504 on Build GRCh38
rs773127821 Chr.4: 77158597-77158597 on Build GRCh38
rs1030019075 Chr.4: 77158839-77158839 on Build GRCh38
rs759871299 Chr.4: 77158604-77158604 on Build GRCh38
rs748934178 Chr.4: 77158655-77158655 on Build GRCh38
rs761724456 Chr.4: 77158497-77158497 on Build GRCh38
rs765416218 Chr.4: 77158518-77158518 on Build GRCh38
rs531415074 Chr.4: 77158580-77158580 on Build GRCh38
rs541087081 Chr.4: 77158810-77158810 on Build GRCh38
rs968168120 Chr.4: 77158533-77158533 on Build GRCh38
rs750719979 Chr.4: 77158644-77158644 on Build GRCh38
rs761779386 Chr.4: 77158584-77158584 on Build GRCh38
rs892286137 Chr.4: 77158605-77158605 on Build GRCh38
rs752926174 Chr.4: 77158783-77158783 on Build GRCh38
rs752020200 Chr.4: 77158551-77158551 on Build GRCh38
rs758986160 Chr.4: 77158538-77158538 on Build GRCh38
rs1001060202 Chr.4: 77158867-77158867 on Build GRCh38
rs4150051 Chr.4: 77158615-77158615 on Build GRCh38
rs533534756 Chr.4: 77158495-77158495 on Build GRCh38
rs1031165738 Chr.4: 77158868-77158868 on Build GRCh38
rs957162241 Chr.4: 77158735-77158735 on Build GRCh38
rs886337299 Chr.4: 77158637-77158637 on Build GRCh38
rs757990917 Chr.4: 77158702-77158703 on Build GRCh38
rs34727878 Chr.4: 77158583-77158583 on Build GRCh38
rs149751721 Chr.4: 77158828-77158828 on Build GRCh38
rs1045039176 Chr.4: 77158578-77158578 on Build GRCh38
rs141389944 Chr.4: 77158741-77158741 on Build GRCh38
rs4150055 Chr.4: 77158861-77158861 on Build GRCh38
rs373722482 Chr.4: 77158490-77158490 on Build GRCh38
rs1004576713 Chr.4: 77158663-77158663 on Build GRCh38
rs533564969 Chr.4: 77158616-77158616 on Build GRCh38
rs974968820 Chr.4: 77158470-77158470 on Build GRCh38
rs545669968 Chr.4: 77158791-77158791 on Build GRCh38
rs767384682 Chr.4: 77158499-77158499 on Build GRCh38
rs773700435 Chr.4: 77158658-77158658 on Build GRCh38
rs748155176 Chr.4: 77158696-77158696 on Build GRCh38
rs968220068 Chr.4: 77158714-77158714 on Build GRCh38
rs901628336 Chr.4: 77158682-77158682 on Build GRCh38
rs142033422 Chr.4: 77158568-77158568 on Build GRCh38
rs962198113 Chr.4: 77158728-77158728 on Build GRCh38
rs375142582 Chr.4: 77158562-77158562 on Build GRCh38
rs769323971 Chr.4: 77158577-77158577 on Build GRCh38
rs144429077 Chr.4: 77158695-77158696 on Build GRCh38

View Full Product Details