Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 79906684-79907182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANTXR2
Entrez Gene ID
118429
Gene Name
anthrax toxin receptor 2
Gene Aliases
CMG-2, CMG2, HFS, ISH, JHF
UniGene
Hs.162963
Gene Chromosome Location
Chr.4: 79901617-80073472 on Build GRCh38
Transcripts
NM_001286780.1 NM_001286781.1 NM_058172.5

SNP Detail

SNP ID Chromosome Location
rs746819365 Chr.4: 79907059-79907059 on Build GRCh38
rs932331806 Chr.4: 79906723-79906723 on Build GRCh38
rs904839992 Chr.4: 79906730-79906730 on Build GRCh38
rs935512377 Chr.4: 79906708-79906708 on Build GRCh38
rs748970937 Chr.4: 79906888-79906888 on Build GRCh38
rs996670112 Chr.4: 79907066-79907066 on Build GRCh38
rs149404014 Chr.4: 79906752-79906752 on Build GRCh38
rs754910280 Chr.4: 79906724-79906724 on Build GRCh38
rs947186737 Chr.4: 79907092-79907092 on Build GRCh38
rs1057133988 Chr.4: 79907118-79907118 on Build GRCh38
rs963505681 Chr.4: 79906957-79906957 on Build GRCh38
rs953399350 Chr.4: 79906862-79906862 on Build GRCh38
rs1040964075 Chr.4: 79907049-79907049 on Build GRCh38
rs965203498 Chr.4: 79906820-79906820 on Build GRCh38
rs895785022 Chr.4: 79907126-79907126 on Build GRCh38
rs144763113 Chr.4: 79906925-79906925 on Build GRCh38
rs574677718 Chr.4: 79906868-79906868 on Build GRCh38
rs889162016 Chr.4: 79907080-79907080 on Build GRCh38
rs779898165 Chr.4: 79907053-79907053 on Build GRCh38
rs1015051862 Chr.4: 79906942-79906942 on Build GRCh38
rs926398259 Chr.4: 79906944-79906944 on Build GRCh38
rs1005049416 Chr.4: 79906940-79906940 on Build GRCh38
rs772511957 Chr.4: 79907039-79907039 on Build GRCh38
rs1036068823 Chr.4: 79906893-79906893 on Build GRCh38
rs1029033260 Chr.4: 79907068-79907068 on Build GRCh38
rs980501615 Chr.4: 79906943-79906943 on Build GRCh38
rs1019378098 Chr.4: 79906815-79906815 on Build GRCh38
rs1001532542 Chr.4: 79906890-79906890 on Build GRCh38
rs956746856 Chr.4: 79907120-79907120 on Build GRCh38
rs919668018 Chr.4: 79906994-79906994 on Build GRCh38
rs563681812 Chr.4: 79906901-79906901 on Build GRCh38
rs4690108 Chr.4: 79907093-79907093 on Build GRCh38
rs1001999653 Chr.4: 79907111-79907111 on Build GRCh38
rs745896907 Chr.4: 79906726-79906726 on Build GRCh38
rs1029064348 Chr.4: 79906844-79906844 on Build GRCh38
rs879716190 Chr.4: 79906972-79906972 on Build GRCh38
rs953872349 Chr.4: 79907010-79907010 on Build GRCh38
rs1000968129 Chr.4: 79907085-79907085 on Build GRCh38
rs886785939 Chr.4: 79907034-79907034 on Build GRCh38
rs997539236 Chr.4: 79906836-79906836 on Build GRCh38
rs1048298243 Chr.4: 79906743-79906743 on Build GRCh38
rs755763454 Chr.4: 79906870-79906870 on Build GRCh38
rs912193117 Chr.4: 79907028-79907028 on Build GRCh38
rs537492648 Chr.4: 79906727-79906727 on Build GRCh38
rs542816123 Chr.4: 79906955-79906955 on Build GRCh38
rs560060945 Chr.4: 79907113-79907113 on Build GRCh38
rs188337138 Chr.4: 79907011-79907011 on Build GRCh38
rs891601909 Chr.4: 79906774-79906774 on Build GRCh38
rs1043146665 Chr.4: 79906819-79906819 on Build GRCh38
rs777254585 Chr.4: 79906872-79906872 on Build GRCh38
rs543914223 Chr.4: 79906789-79906789 on Build GRCh38
rs995607519 Chr.4: 79906734-79906734 on Build GRCh38
rs943944740 Chr.4: 79907031-79907031 on Build GRCh38
rs1004136165 Chr.4: 79906762-79906762 on Build GRCh38
rs905958998 Chr.4: 79907104-79907104 on Build GRCh38
rs1011704171 Chr.4: 79906776-79906776 on Build GRCh38
rs925312947 Chr.4: 79906881-79906881 on Build GRCh38

View Full Product Details