Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83462301-83462852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM175A
Entrez Gene ID
84142
Gene Name
family with sequence similarity 175 member A
Gene Aliases
ABRA1, CCDC98
UniGene
Hs.334772
Gene Chromosome Location
Chr.4: 83459515-83485181 on Build GRCh38
Transcripts
NM_139076.2 XM_005263280.4 XM_017008694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6210940 c.1000G>A p.E334K Chr.4: 83462699-83462699 on Build GRCh38
6956398 c.908delA p.N303fs*17 Chr.4: 83462791-83462791 on Build GRCh38
3760790 c.1042G>A p.A348T Chr.4: 83462657-83462657 on Build GRCh38
6011539 c.1096C>T p.Q366* Chr.4: 83462603-83462603 on Build GRCh38
6101284 c.985C>A p.H329N Chr.4: 83462714-83462714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749943018 Chr.4: 83462774-83462774 on Build GRCh38
rs1022505540 Chr.4: 83462402-83462402 on Build GRCh38
rs778357689 Chr.4: 83462713-83462713 on Build GRCh38
rs759694768 Chr.4: 83462635-83462635 on Build GRCh38
rs760957137 Chr.4: 83462504-83462504 on Build GRCh38
rs773812911 Chr.4: 83462643-83462643 on Build GRCh38
rs779168599 Chr.4: 83462598-83462598 on Build GRCh38
rs748616526 Chr.4: 83462486-83462486 on Build GRCh38
rs900639769 Chr.4: 83462694-83462694 on Build GRCh38
rs756946633 Chr.4: 83462467-83462467 on Build GRCh38
rs781307588 Chr.4: 83462803-83462803 on Build GRCh38
rs771082457 Chr.4: 83462734-83462734 on Build GRCh38
rs756892998 Chr.4: 83462576-83462576 on Build GRCh38
rs764951890 Chr.4: 83462537-83462537 on Build GRCh38
rs941211611 Chr.4: 83462498-83462498 on Build GRCh38
rs772754954 Chr.4: 83462607-83462607 on Build GRCh38
rs774216794 Chr.4: 83462764-83462764 on Build GRCh38
rs966702733 Chr.4: 83462405-83462406 on Build GRCh38
rs747527820 Chr.4: 83462718-83462718 on Build GRCh38
rs867481466 Chr.4: 83462506-83462507 on Build GRCh38
rs758296226 Chr.4: 83462541-83462541 on Build GRCh38
rs766975460 Chr.4: 83462771-83462771 on Build GRCh38
rs142910445 Chr.4: 83462608-83462608 on Build GRCh38
rs781160014 Chr.4: 83462740-83462740 on Build GRCh38
rs368663402 Chr.4: 83462476-83462476 on Build GRCh38
rs140407442 Chr.4: 83462802-83462802 on Build GRCh38
rs762929045 Chr.4: 83462641-83462641 on Build GRCh38
rs374462211 Chr.4: 83462454-83462454 on Build GRCh38
rs747151396 Chr.4: 83462609-83462609 on Build GRCh38
rs755313466 Chr.4: 83462593-83462593 on Build GRCh38
rs367958380 Chr.4: 83462680-83462680 on Build GRCh38
rs752752926 Chr.4: 83462602-83462603 on Build GRCh38
rs776562122 Chr.4: 83462529-83462529 on Build GRCh38
rs529156082 Chr.4: 83462497-83462497 on Build GRCh38
rs79357787 Chr.4: 83462748-83462748 on Build GRCh38
rs1017716903 Chr.4: 83462341-83462341 on Build GRCh38
rs755755614 Chr.4: 83462663-83462663 on Build GRCh38
rs759305711 Chr.4: 83462536-83462536 on Build GRCh38
rs929635919 Chr.4: 83462622-83462622 on Build GRCh38
rs1038252853 Chr.4: 83462555-83462555 on Build GRCh38
rs13125836 Chr.4: 83462582-83462582 on Build GRCh38
rs766764068 Chr.4: 83462510-83462510 on Build GRCh38
rs765284026 Chr.4: 83462448-83462448 on Build GRCh38
rs774564381 Chr.4: 83462396-83462396 on Build GRCh38
rs778300694 Chr.4: 83462787-83462787 on Build GRCh38
rs557827320 Chr.4: 83462381-83462381 on Build GRCh38
rs587780261 Chr.4: 83462666-83462667 on Build GRCh38
rs138986552 Chr.4: 83462782-83462782 on Build GRCh38
rs761761558 Chr.4: 83462767-83462767 on Build GRCh38
rs750046185 Chr.4: 83462661-83462661 on Build GRCh38
rs201627097 Chr.4: 83462617-83462617 on Build GRCh38
rs769962886 Chr.4: 83462746-83462746 on Build GRCh38
rs751122263 Chr.4: 83462575-83462575 on Build GRCh38
rs587780262 Chr.4: 83462592-83462593 on Build GRCh38
rs760360071 Chr.4: 83462779-83462779 on Build GRCh38
rs758777622 Chr.4: 83462783-83462783 on Build GRCh38
rs775185945 Chr.4: 83462639-83462639 on Build GRCh38
rs72931491 Chr.4: 83462358-83462358 on Build GRCh38
rs752068094 Chr.4: 83462795-83462795 on Build GRCh38
rs763567031 Chr.4: 83462547-83462547 on Build GRCh38
rs748650098 Chr.4: 83462604-83462604 on Build GRCh38
rs779312880 Chr.4: 83462669-83462669 on Build GRCh38
rs775345294 Chr.4: 83462747-83462747 on Build GRCh38
rs766724020 Chr.4: 83462507-83462508 on Build GRCh38
rs746621063 Chr.4: 83462463-83462463 on Build GRCh38
rs772142848 Chr.4: 83462494-83462494 on Build GRCh38
rs764429803 Chr.4: 83462688-83462688 on Build GRCh38
rs377162113 Chr.4: 83462809-83462809 on Build GRCh38
rs774960680 Chr.4: 83462563-83462564 on Build GRCh38
rs752627041 Chr.4: 83462538-83462538 on Build GRCh38
rs111720506 Chr.4: 83462408-83462408 on Build GRCh38
rs376489465 Chr.4: 83462758-83462758 on Build GRCh38
rs745996212 Chr.4: 83462810-83462810 on Build GRCh38
rs587780264 Chr.4: 83462560-83462560 on Build GRCh38
rs12642536 Chr.4: 83462657-83462657 on Build GRCh38
rs143012786 Chr.4: 83462804-83462804 on Build GRCh38
rs200281698 Chr.4: 83462485-83462485 on Build GRCh38
rs746068286 Chr.4: 83462742-83462742 on Build GRCh38
rs878855206 Chr.4: 83462595-83462597 on Build GRCh38
rs756152827 Chr.4: 83462608-83462609 on Build GRCh38
rs758969039 Chr.4: 83462681-83462681 on Build GRCh38
rs754855587 Chr.4: 83462436-83462436 on Build GRCh38
rs769589027 Chr.4: 83462637-83462637 on Build GRCh38
rs587780263 Chr.4: 83462591-83462591 on Build GRCh38
rs76266580 Chr.4: 83462336-83462336 on Build GRCh38
rs745512435 Chr.4: 83462587-83462587 on Build GRCh38
rs768755623 Chr.4: 83462348-83462348 on Build GRCh38
rs769078691 Chr.4: 83462819-83462820 on Build GRCh38
rs766011574 Chr.4: 83462781-83462781 on Build GRCh38
rs763365161 Chr.4: 83462454-83462454 on Build GRCh38
rs545866366 Chr.4: 83462372-83462372 on Build GRCh38
rs751068863 Chr.4: 83462651-83462651 on Build GRCh38
rs561858434 Chr.4: 83462477-83462477 on Build GRCh38
rs535462348 Chr.4: 83462330-83462330 on Build GRCh38
rs759939936 Chr.4: 83462566-83462566 on Build GRCh38
rs587780269 Chr.4: 83462784-83462784 on Build GRCh38
rs776208491 Chr.4: 83462448-83462450 on Build GRCh38
rs771137917 Chr.4: 83462615-83462615 on Build GRCh38
rs753734498 Chr.4: 83462434-83462434 on Build GRCh38

View Full Product Details