Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83470279-83470792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM175A
Entrez Gene ID
84142
Gene Name
family with sequence similarity 175 member A
Gene Aliases
ABRA1, CCDC98
UniGene
Hs.334772
Gene Chromosome Location
Chr.4: 83459515-83485181 on Build GRCh38
Transcripts
NM_139076.2 XM_005263280.4 XM_017008694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
211445 c.377C>T p.S126L Chr.4: 83470302-83470302 on Build GRCh38
6917100 c.324G>C p.Q108H Chr.4: 83470355-83470355 on Build GRCh38
1291977 c.342G>A p.E114E Chr.4: 83470337-83470337 on Build GRCh38
6937683 c.332C>T p.T111M Chr.4: 83470347-83470347 on Build GRCh38
734440 c.313C>T p.H105Y Chr.4: 83470366-83470366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115192589 Chr.4: 83470504-83470504 on Build GRCh38
rs201884868 Chr.4: 83470486-83470486 on Build GRCh38
rs996716972 Chr.4: 83470542-83470542 on Build GRCh38
rs778038758 Chr.4: 83470413-83470413 on Build GRCh38
rs137876115 Chr.4: 83470315-83470315 on Build GRCh38
rs149402633 Chr.4: 83470493-83470493 on Build GRCh38
rs759499928 Chr.4: 83470319-83470319 on Build GRCh38
rs907497479 Chr.4: 83470442-83470442 on Build GRCh38
rs544130487 Chr.4: 83470617-83470617 on Build GRCh38
rs948890862 Chr.4: 83470720-83470720 on Build GRCh38
rs1025170783 Chr.4: 83470646-83470646 on Build GRCh38
rs748835757 Chr.4: 83470614-83470614 on Build GRCh38
rs748680448 Chr.4: 83470372-83470372 on Build GRCh38
rs751510689 Chr.4: 83470435-83470436 on Build GRCh38
rs952725384 Chr.4: 83470699-83470699 on Build GRCh38
rs542570943 Chr.4: 83470368-83470368 on Build GRCh38
rs957081607 Chr.4: 83470626-83470626 on Build GRCh38
rs796478814 Chr.4: 83470464-83470464 on Build GRCh38
rs1045946337 Chr.4: 83470431-83470431 on Build GRCh38
rs998638331 Chr.4: 83470387-83470387 on Build GRCh38
rs878855207 Chr.4: 83470380-83470380 on Build GRCh38
rs138846806 Chr.4: 83470446-83470446 on Build GRCh38
rs989932107 Chr.4: 83470396-83470396 on Build GRCh38
rs193082384 Chr.4: 83470724-83470724 on Build GRCh38
rs528119988 Chr.4: 83470472-83470475 on Build GRCh38
rs1043106380 Chr.4: 83470596-83470596 on Build GRCh38
rs756903113 Chr.4: 83470351-83470351 on Build GRCh38
rs565014701 Chr.4: 83470698-83470698 on Build GRCh38
rs57818535 Chr.4: 83470641-83470641 on Build GRCh38
rs531933896 Chr.4: 83470580-83470580 on Build GRCh38
rs779913611 Chr.4: 83470417-83470418 on Build GRCh38
rs763619387 Chr.4: 83470336-83470336 on Build GRCh38
rs969690987 Chr.4: 83470463-83470467 on Build GRCh38
rs572536976 Chr.4: 83470531-83470531 on Build GRCh38
rs928809286 Chr.4: 83470752-83470752 on Build GRCh38
rs919974076 Chr.4: 83470474-83470474 on Build GRCh38
rs200068246 Chr.4: 83470371-83470371 on Build GRCh38
rs188900531 Chr.4: 83470533-83470533 on Build GRCh38
rs779497289 Chr.4: 83470369-83470369 on Build GRCh38
rs28540634 Chr.4: 83470479-83470479 on Build GRCh38
rs368005955 Chr.4: 83470601-83470601 on Build GRCh38
rs367592316 Chr.4: 83470390-83470390 on Build GRCh38
rs533564631 Chr.4: 83470739-83470739 on Build GRCh38
rs955018036 Chr.4: 83470308-83470308 on Build GRCh38
rs1047680851 Chr.4: 83470458-83470458 on Build GRCh38
rs747228318 Chr.4: 83470423-83470423 on Build GRCh38
rs199992927 Chr.4: 83470426-83470426 on Build GRCh38
rs573603690 Chr.4: 83470357-83470357 on Build GRCh38
rs545108219 Chr.4: 83470548-83470548 on Build GRCh38
rs1013744034 Chr.4: 83470622-83470622 on Build GRCh38
rs762389463 Chr.4: 83470334-83470334 on Build GRCh38
rs915690309 Chr.4: 83470690-83470690 on Build GRCh38
rs990285334 Chr.4: 83470662-83470662 on Build GRCh38
rs745581822 Chr.4: 83470444-83470444 on Build GRCh38
rs751175389 Chr.4: 83470346-83470346 on Build GRCh38
rs924277966 Chr.4: 83470352-83470352 on Build GRCh38
rs375331034 Chr.4: 83470364-83470364 on Build GRCh38
rs561419479 Chr.4: 83470558-83470558 on Build GRCh38
rs765150557 Chr.4: 83470330-83470330 on Build GRCh38
rs918182945 Chr.4: 83470459-83470463 on Build GRCh38
rs981612898 Chr.4: 83470737-83470737 on Build GRCh38
rs996802966 Chr.4: 83470592-83470592 on Build GRCh38
rs977345649 Chr.4: 83470605-83470605 on Build GRCh38
rs981087841 Chr.4: 83470687-83470687 on Build GRCh38
rs747293271 Chr.4: 83470528-83470528 on Build GRCh38
rs970038292 Chr.4: 83470668-83470668 on Build GRCh38
rs776481250 Chr.4: 83470437-83470437 on Build GRCh38

View Full Product Details