Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 84635073-84635574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDS1
Entrez Gene ID
1040
Gene Name
CDP-diacylglycerol synthase 1
Gene Aliases
CDS
UniGene
Hs.654899
Gene Chromosome Location
Chr.4: 84582904-84651340 on Build GRCh38
Transcripts
NM_001263.3 XM_005262687.2 XM_017007648.1 XM_017007649.1 XM_017007650.1 XM_017007651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
33082 c.789_790insT p.G264fs*7 Chr.4: 84635330-84635331 on Build GRCh38
5351128 c.723-9T>A p.? Chr.4: 84635255-84635255 on Build GRCh38
1058330 c.780A>T p.G260G Chr.4: 84635321-84635321 on Build GRCh38
6845344 c.723-2A>G p.? Chr.4: 84635262-84635262 on Build GRCh38
1736673 c.723-1G>A p.? Chr.4: 84635263-84635263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187037308 Chr.4: 84635127-84635127 on Build GRCh38
rs751607512 Chr.4: 84635372-84635373 on Build GRCh38
rs760620010 Chr.4: 84635302-84635302 on Build GRCh38
rs887894659 Chr.4: 84635535-84635535 on Build GRCh38
rs746782356 Chr.4: 84635207-84635207 on Build GRCh38
rs374317419 Chr.4: 84635457-84635457 on Build GRCh38
rs952027956 Chr.4: 84635179-84635179 on Build GRCh38
rs774094919 Chr.4: 84635256-84635257 on Build GRCh38
rs11310518 Chr.4: 84635240-84635240 on Build GRCh38
rs758946183 Chr.4: 84635363-84635363 on Build GRCh38
rs922328146 Chr.4: 84635193-84635193 on Build GRCh38
rs758510708 Chr.4: 84635367-84635368 on Build GRCh38
rs924548808 Chr.4: 84635230-84635230 on Build GRCh38
rs984304341 Chr.4: 84635146-84635146 on Build GRCh38
rs376285323 Chr.4: 84635255-84635255 on Build GRCh38
rs201506801 Chr.4: 84635373-84635373 on Build GRCh38
rs758269580 Chr.4: 84635380-84635380 on Build GRCh38
rs1003830596 Chr.4: 84635120-84635120 on Build GRCh38
rs750874122 Chr.4: 84635232-84635232 on Build GRCh38
rs770563380 Chr.4: 84635256-84635256 on Build GRCh38
rs951504123 Chr.4: 84635145-84635145 on Build GRCh38
rs773293764 Chr.4: 84635295-84635295 on Build GRCh38
rs7683338 Chr.4: 84635233-84635233 on Build GRCh38
rs777776191 Chr.4: 84635382-84635382 on Build GRCh38
rs760629917 Chr.4: 84635418-84635418 on Build GRCh38
rs368292788 Chr.4: 84635213-84635213 on Build GRCh38
rs746785761 Chr.4: 84635386-84635386 on Build GRCh38
rs767805161 Chr.4: 84635370-84635370 on Build GRCh38
rs762103217 Chr.4: 84635357-84635357 on Build GRCh38
rs757165524 Chr.4: 84635107-84635107 on Build GRCh38
rs745406815 Chr.4: 84635403-84635403 on Build GRCh38
rs776261160 Chr.4: 84635330-84635330 on Build GRCh38
rs781680387 Chr.4: 84635220-84635220 on Build GRCh38
rs773227678 Chr.4: 84635240-84635242 on Build GRCh38
rs770949394 Chr.4: 84635400-84635400 on Build GRCh38
rs776401991 Chr.4: 84635401-84635401 on Build GRCh38
rs768975884 Chr.4: 84635264-84635264 on Build GRCh38
rs528617646 Chr.4: 84635256-84635256 on Build GRCh38
rs750921286 Chr.4: 84635363-84635364 on Build GRCh38
rs975482521 Chr.4: 84635188-84635188 on Build GRCh38
rs755185369 Chr.4: 84635373-84635374 on Build GRCh38
rs547078642 Chr.4: 84635283-84635283 on Build GRCh38
rs754848200 Chr.4: 84635240-84635241 on Build GRCh38
rs935808202 Chr.4: 84635324-84635324 on Build GRCh38
rs775074696 Chr.4: 84635369-84635369 on Build GRCh38
rs778975938 Chr.4: 84635271-84635271 on Build GRCh38
rs543964678 Chr.4: 84635174-84635174 on Build GRCh38
rs993593144 Chr.4: 84635164-84635164 on Build GRCh38
rs887367162 Chr.4: 84635548-84635548 on Build GRCh38
rs866901321 Chr.4: 84635358-84635358 on Build GRCh38
rs191424064 Chr.4: 84635521-84635521 on Build GRCh38
rs750784443 Chr.4: 84635371-84635371 on Build GRCh38
rs748320604 Chr.4: 84635379-84635379 on Build GRCh38
rs776894995 Chr.4: 84635353-84635354 on Build GRCh38
rs757578210 Chr.4: 84635215-84635215 on Build GRCh38
rs767152003 Chr.4: 84635321-84635322 on Build GRCh38
rs756705270 Chr.4: 84635372-84635372 on Build GRCh38
rs747909544 Chr.4: 84635239-84635240 on Build GRCh38
rs770768820 Chr.4: 84635321-84635321 on Build GRCh38
rs759091263 Chr.4: 84635322-84635322 on Build GRCh38
rs1007540588 Chr.4: 84635536-84635536 on Build GRCh38
rs771940970 Chr.4: 84635290-84635290 on Build GRCh38
rs897679091 Chr.4: 84635421-84635421 on Build GRCh38
rs529325289 Chr.4: 84635368-84635368 on Build GRCh38
rs780249664 Chr.4: 84635234-84635234 on Build GRCh38
rs1008191471 Chr.4: 84635198-84635198 on Build GRCh38
rs779214749 Chr.4: 84635378-84635378 on Build GRCh38
rs909600521 Chr.4: 84635159-84635159 on Build GRCh38
rs201623642 Chr.4: 84635362-84635362 on Build GRCh38
rs1025875459 Chr.4: 84635542-84635542 on Build GRCh38
rs748866234 Chr.4: 84635255-84635256 on Build GRCh38
rs1003565028 Chr.4: 84635529-84635529 on Build GRCh38
rs1014084684 Chr.4: 84635099-84635099 on Build GRCh38
rs552223123 Chr.4: 84635359-84635359 on Build GRCh38
rs550956288 Chr.4: 84635407-84635407 on Build GRCh38
rs866419809 Chr.4: 84635265-84635265 on Build GRCh38
rs933888980 Chr.4: 84635197-84635197 on Build GRCh38
rs893841965 Chr.4: 84635383-84635383 on Build GRCh38

View Full Product Details