Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 169654314-169654825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK2
Entrez Gene ID
1794
Gene Name
dedicator of cytokinesis 2
Gene Aliases
IMD40
UniGene
Hs.586174
Gene Chromosome Location
Chr.5: 169637247-170083382 on Build GRCh38
Transcripts
NM_004946.2 XM_005265830.3 XM_011534448.2 XM_011534449.2 XM_011534450.2 XM_011534451.2 XM_017009189.1 XM_017009190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4943049 c.66C>A p.S22R Chr.5: 169654425-169654425 on Build GRCh38
280565 c.107G>A p.R36Q Chr.5: 169654466-169654466 on Build GRCh38
1065692 c.93C>T p.I31I Chr.5: 169654452-169654452 on Build GRCh38
1665907 c.127+2T>C p.? Chr.5: 169654488-169654488 on Build GRCh38
1180996 c.71delC p.Q26fs*25 Chr.5: 169654430-169654430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003950966 Chr.5: 169654614-169654614 on Build GRCh38
rs756827620 Chr.5: 169654480-169654480 on Build GRCh38
rs770367983 Chr.5: 169654393-169654393 on Build GRCh38
rs776439216 Chr.5: 169654504-169654504 on Build GRCh38
rs997045199 Chr.5: 169654696-169654696 on Build GRCh38
rs560210829 Chr.5: 169654742-169654742 on Build GRCh38
rs978510151 Chr.5: 169654712-169654712 on Build GRCh38
rs1019494244 Chr.5: 169654663-169654663 on Build GRCh38
rs562507776 Chr.5: 169654549-169654549 on Build GRCh38
rs1026741262 Chr.5: 169654348-169654348 on Build GRCh38
rs759644547 Chr.5: 169654512-169654512 on Build GRCh38
rs764520982 Chr.5: 169654478-169654478 on Build GRCh38
rs374789654 Chr.5: 169654458-169654458 on Build GRCh38
rs776586113 Chr.5: 169654429-169654429 on Build GRCh38
rs578109974 Chr.5: 169654384-169654384 on Build GRCh38
rs768442577 Chr.5: 169654503-169654503 on Build GRCh38
rs757957020 Chr.5: 169654492-169654492 on Build GRCh38
rs775126575 Chr.5: 169654421-169654421 on Build GRCh38
rs993418598 Chr.5: 169654613-169654613 on Build GRCh38
rs141894939 Chr.5: 169654449-169654449 on Build GRCh38
rs975349336 Chr.5: 169654464-169654464 on Build GRCh38
rs925614155 Chr.5: 169654743-169654743 on Build GRCh38
rs763142081 Chr.5: 169654434-169654434 on Build GRCh38
rs760709796 Chr.5: 169654425-169654425 on Build GRCh38
rs776680266 Chr.5: 169654382-169654383 on Build GRCh38
rs185449432 Chr.5: 169654410-169654410 on Build GRCh38
rs529148848 Chr.5: 169654612-169654612 on Build GRCh38
rs745570765 Chr.5: 169654491-169654491 on Build GRCh38
rs771834553 Chr.5: 169654389-169654389 on Build GRCh38
rs370494583 Chr.5: 169654452-169654452 on Build GRCh38
rs756449193 Chr.5: 169654466-169654466 on Build GRCh38
rs369301781 Chr.5: 169654536-169654536 on Build GRCh38
rs766504591 Chr.5: 169654439-169654439 on Build GRCh38
rs781409133 Chr.5: 169654364-169654364 on Build GRCh38
rs934359496 Chr.5: 169654763-169654763 on Build GRCh38
rs531692992 Chr.5: 169654479-169654479 on Build GRCh38
rs905918483 Chr.5: 169654575-169654575 on Build GRCh38
rs772916136 Chr.5: 169654677-169654677 on Build GRCh38
rs879757157 Chr.5: 169654694-169654694 on Build GRCh38
rs1052788819 Chr.5: 169654775-169654775 on Build GRCh38
rs778300082 Chr.5: 169654377-169654377 on Build GRCh38
rs374674113 Chr.5: 169654531-169654531 on Build GRCh38
rs958168436 Chr.5: 169654699-169654699 on Build GRCh38
rs202240320 Chr.5: 169654521-169654521 on Build GRCh38
rs376950305 Chr.5: 169654426-169654426 on Build GRCh38
rs755402670 Chr.5: 169654357-169654357 on Build GRCh38
rs745515617 Chr.5: 169654402-169654402 on Build GRCh38
rs576137078 Chr.5: 169654561-169654561 on Build GRCh38
rs546901899 Chr.5: 169654737-169654737 on Build GRCh38
rs753293125 Chr.5: 169654368-169654368 on Build GRCh38
rs759741411 Chr.5: 169654429-169654430 on Build GRCh38
rs1055381312 Chr.5: 169654550-169654550 on Build GRCh38
rs779719029 Chr.5: 169654498-169654498 on Build GRCh38
rs748096920 Chr.5: 169654506-169654506 on Build GRCh38
rs771611960 Chr.5: 169654697-169654697 on Build GRCh38
rs542631592 Chr.5: 169654538-169654538 on Build GRCh38
rs989517037 Chr.5: 169654720-169654720 on Build GRCh38
rs771196980 Chr.5: 169654510-169654510 on Build GRCh38
rs543944724 Chr.5: 169654562-169654562 on Build GRCh38
rs4867873 Chr.5: 169654602-169654602 on Build GRCh38
rs774538414 Chr.5: 169654511-169654511 on Build GRCh38
rs746803598 Chr.5: 169654502-169654502 on Build GRCh38
rs756543562 Chr.5: 169654376-169654376 on Build GRCh38
rs775719760 Chr.5: 169654527-169654527 on Build GRCh38
rs987325718 Chr.5: 169654481-169654481 on Build GRCh38
rs778380712 Chr.5: 169654485-169654485 on Build GRCh38
rs767961785 Chr.5: 169654455-169654455 on Build GRCh38
rs145786696 Chr.5: 169654788-169654788 on Build GRCh38
rs778542070 Chr.5: 169654394-169654394 on Build GRCh38
rs925645273 Chr.5: 169654517-169654517 on Build GRCh38
rs945441705 Chr.5: 169654514-169654514 on Build GRCh38
rs928125181 Chr.5: 169654547-169654547 on Build GRCh38
rs954659710 Chr.5: 169654651-169654651 on Build GRCh38
rs896001842 Chr.5: 169654356-169654356 on Build GRCh38

View Full Product Details