Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 169674210-169674715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK2
Entrez Gene ID
1794
Gene Name
dedicator of cytokinesis 2
Gene Aliases
IMD40
UniGene
Hs.586174
Gene Chromosome Location
Chr.5: 169637247-170083382 on Build GRCh38
Transcripts
NM_004946.2 XM_005265830.3 XM_011534448.2 XM_011534449.2 XM_011534450.2 XM_011534451.2 XM_017009189.1 XM_017009190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5819836 c.390G>C p.Q130H Chr.5: 169674365-169674365 on Build GRCh38
4547086 c.414G>A p.K138K Chr.5: 169674389-169674389 on Build GRCh38
3776519 c.372G>C p.L124L Chr.5: 169674347-169674347 on Build GRCh38
7428732 c.328delA p.K111fs*10 Chr.5: 169674303-169674303 on Build GRCh38
3614033 c.401G>A p.G134E Chr.5: 169674376-169674376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775108769 Chr.5: 169674265-169674265 on Build GRCh38
rs545379811 Chr.5: 169674255-169674255 on Build GRCh38
rs188953420 Chr.5: 169674612-169674612 on Build GRCh38
rs922613863 Chr.5: 169674606-169674606 on Build GRCh38
rs770738793 Chr.5: 169674469-169674472 on Build GRCh38
rs762701119 Chr.5: 169674443-169674443 on Build GRCh38
rs746532024 Chr.5: 169674394-169674394 on Build GRCh38
rs563866643 Chr.5: 169674284-169674284 on Build GRCh38
rs575409388 Chr.5: 169674656-169674656 on Build GRCh38
rs746362515 Chr.5: 169674263-169674263 on Build GRCh38
rs757696002 Chr.5: 169674344-169674344 on Build GRCh38
rs772905218 Chr.5: 169674258-169674258 on Build GRCh38
rs779123178 Chr.5: 169674346-169674346 on Build GRCh38
rs1018090120 Chr.5: 169674379-169674379 on Build GRCh38
rs924809831 Chr.5: 169674277-169674277 on Build GRCh38
rs200514075 Chr.5: 169674268-169674268 on Build GRCh38
rs1037406523 Chr.5: 169674270-169674270 on Build GRCh38
rs893987798 Chr.5: 169674324-169674324 on Build GRCh38
rs78450053 Chr.5: 169674364-169674364 on Build GRCh38
rs746844645 Chr.5: 169674457-169674477 on Build GRCh38
rs750421267 Chr.5: 169674313-169674313 on Build GRCh38
rs768272691 Chr.5: 169674481-169674481 on Build GRCh38
rs763830472 Chr.5: 169674274-169674274 on Build GRCh38
rs751428460 Chr.5: 169674450-169674450 on Build GRCh38
rs2244445 Chr.5: 169674595-169674595 on Build GRCh38
rs61512497 Chr.5: 169674685-169674685 on Build GRCh38
rs760872058 Chr.5: 169674464-169674464 on Build GRCh38
rs552925477 Chr.5: 169674590-169674590 on Build GRCh38
rs1049895394 Chr.5: 169674288-169674288 on Build GRCh38
rs956047250 Chr.5: 169674484-169674484 on Build GRCh38
rs913839233 Chr.5: 169674568-169674568 on Build GRCh38
rs749463827 Chr.5: 169674342-169674342 on Build GRCh38
rs878984678 Chr.5: 169674307-169674307 on Build GRCh38
rs756125120 Chr.5: 169674336-169674336 on Build GRCh38
rs774987244 Chr.5: 169674246-169674246 on Build GRCh38
rs932857824 Chr.5: 169674614-169674614 on Build GRCh38
rs981996723 Chr.5: 169674575-169674575 on Build GRCh38
rs1050008284 Chr.5: 169674626-169674626 on Build GRCh38
rs769682916 Chr.5: 169674432-169674432 on Build GRCh38
rs112922689 Chr.5: 169674326-169674326 on Build GRCh38
rs377117509 Chr.5: 169674281-169674281 on Build GRCh38
rs780547647 Chr.5: 169674486-169674486 on Build GRCh38
rs1012793578 Chr.5: 169674513-169674513 on Build GRCh38
rs562031933 Chr.5: 169674647-169674647 on Build GRCh38
rs751169724 Chr.5: 169674646-169674646 on Build GRCh38
rs140206456 Chr.5: 169674374-169674374 on Build GRCh38
rs776090099 Chr.5: 169674278-169674278 on Build GRCh38
rs566107879 Chr.5: 169674510-169674510 on Build GRCh38
rs371312059 Chr.5: 169674445-169674445 on Build GRCh38
rs771755377 Chr.5: 169674264-169674264 on Build GRCh38
rs767985225 Chr.5: 169674398-169674398 on Build GRCh38
rs961504809 Chr.5: 169674596-169674596 on Build GRCh38
rs565477115 Chr.5: 169674591-169674591 on Build GRCh38
rs544475548 Chr.5: 169674604-169674604 on Build GRCh38
rs764064687 Chr.5: 169674471-169674471 on Build GRCh38
rs1017809991 Chr.5: 169674682-169674682 on Build GRCh38
rs776741593 Chr.5: 169674310-169674310 on Build GRCh38
rs1026577454 Chr.5: 169674338-169674338 on Build GRCh38
rs146486187 Chr.5: 169674474-169674474 on Build GRCh38
rs367573686 Chr.5: 169674437-169674437 on Build GRCh38
rs760229980 Chr.5: 169674621-169674621 on Build GRCh38
rs987988813 Chr.5: 169674508-169674508 on Build GRCh38
rs549670923 Chr.5: 169674422-169674422 on Build GRCh38
rs900668926 Chr.5: 169674687-169674687 on Build GRCh38
rs112654610 Chr.5: 169674569-169674569 on Build GRCh38
rs752809346 Chr.5: 169674335-169674335 on Build GRCh38
rs867401091 Chr.5: 169674350-169674350 on Build GRCh38
rs764939243 Chr.5: 169674293-169674293 on Build GRCh38
rs901403929 Chr.5: 169674242-169674242 on Build GRCh38
rs376223836 Chr.5: 169674472-169674472 on Build GRCh38
rs1022712833 Chr.5: 169674518-169674518 on Build GRCh38
rs761361714 Chr.5: 169674429-169674429 on Build GRCh38
rs750839101 Chr.5: 169674477-169674477 on Build GRCh38
rs35393134 Chr.5: 169674341-169674341 on Build GRCh38
rs373741346 Chr.5: 169674380-169674380 on Build GRCh38
rs780712308 Chr.5: 169674480-169674480 on Build GRCh38
rs998913556 Chr.5: 169674440-169674440 on Build GRCh38
rs767627447 Chr.5: 169674328-169674328 on Build GRCh38
rs758777904 Chr.5: 169674479-169674479 on Build GRCh38
rs1035379247 Chr.5: 169674452-169674452 on Build GRCh38

View Full Product Details