Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 169863626-169864127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK2
Entrez Gene ID
1794
Gene Name
dedicator of cytokinesis 2
Gene Aliases
IMD40
UniGene
Hs.586174
Gene Chromosome Location
Chr.5: 169637247-170083382 on Build GRCh38
Transcripts
NM_004946.2 XM_011534449.2 XM_011534450.2 XM_011534451.2 XM_017009189.1 XM_017009190.1 NM_001129891.1 XM_011534415.2

Gene Symbol
FAM196B
Entrez Gene ID
100131897
Gene Name
family with sequence similarity 196 member B
Gene Aliases
C5orf57
UniGene
Hs.721917
Gene Chromosome Location
Chr.5: 169861304-169980740 on Build GRCh38
Transcripts
NM_004946.2 XM_011534449.2 XM_011534450.2 XM_011534451.2 XM_017009189.1 XM_017009190.1 NM_001129891.1 XM_011534415.2

SNP Detail

SNP ID Chromosome Location
rs193105041 Chr.5: 169864055-169864055 on Build GRCh38
rs116590429 Chr.5: 169863827-169863827 on Build GRCh38
rs990105604 Chr.5: 169863922-169863922 on Build GRCh38
rs74350237 Chr.5: 169863890-169863890 on Build GRCh38
rs1011694814 Chr.5: 169863804-169863804 on Build GRCh38
rs889331151 Chr.5: 169863781-169863781 on Build GRCh38
rs547047416 Chr.5: 169863941-169863941 on Build GRCh38
rs112636019 Chr.5: 169863978-169863978 on Build GRCh38
rs936641441 Chr.5: 169863912-169863912 on Build GRCh38
rs567266380 Chr.5: 169863973-169863973 on Build GRCh38
rs562496621 Chr.5: 169863717-169863717 on Build GRCh38
rs377082495 Chr.5: 169864081-169864081 on Build GRCh38
rs951602474 Chr.5: 169863801-169863801 on Build GRCh38
rs909315662 Chr.5: 169863954-169863954 on Build GRCh38
rs939257450 Chr.5: 169863656-169863656 on Build GRCh38
rs1055523399 Chr.5: 169863987-169863987 on Build GRCh38
rs261029 Chr.5: 169863988-169863988 on Build GRCh38
rs969275580 Chr.5: 169864101-169864101 on Build GRCh38
rs958092249 Chr.5: 169864097-169864097 on Build GRCh38
rs992413550 Chr.5: 169863669-169863669 on Build GRCh38
rs569939141 Chr.5: 169864009-169864009 on Build GRCh38
rs868708255 Chr.5: 169864043-169864043 on Build GRCh38
rs1022031369 Chr.5: 169863810-169863810 on Build GRCh38
rs182459874 Chr.5: 169863927-169863927 on Build GRCh38
rs187685993 Chr.5: 169864040-169864040 on Build GRCh38
rs72842503 Chr.5: 169863874-169863874 on Build GRCh38
rs887853618 Chr.5: 169863727-169863727 on Build GRCh38
rs923259271 Chr.5: 169863976-169863976 on Build GRCh38
rs1001239902 Chr.5: 169863670-169863670 on Build GRCh38
rs940727396 Chr.5: 169863960-169863960 on Build GRCh38
rs889354788 Chr.5: 169864006-169864006 on Build GRCh38
rs887350156 Chr.5: 169864049-169864049 on Build GRCh38
rs1051037241 Chr.5: 169863768-169863768 on Build GRCh38
rs997745956 Chr.5: 169863767-169863767 on Build GRCh38
rs940701163 Chr.5: 169863745-169863745 on Build GRCh38
rs757100616 Chr.5: 169863712-169863712 on Build GRCh38
rs747249754 Chr.5: 169864082-169864082 on Build GRCh38
rs1049214118 Chr.5: 169863722-169863722 on Build GRCh38
rs906940908 Chr.5: 169863649-169863649 on Build GRCh38
rs778536621 Chr.5: 169863828-169863828 on Build GRCh38
rs984609694 Chr.5: 169863953-169863953 on Build GRCh38
rs999195910 Chr.5: 169864077-169864077 on Build GRCh38
rs967750590 Chr.5: 169863816-169863816 on Build GRCh38
rs916480756 Chr.5: 169863891-169863891 on Build GRCh38
rs753358258 Chr.5: 169863748-169863748 on Build GRCh38
rs902107848 Chr.5: 169863759-169863759 on Build GRCh38
rs1023580915 Chr.5: 169864098-169864098 on Build GRCh38
rs1026293940 Chr.5: 169863762-169863762 on Build GRCh38
rs372429267 Chr.5: 169863711-169863711 on Build GRCh38
rs1036341674 Chr.5: 169863755-169863755 on Build GRCh38
rs1031085100 Chr.5: 169864078-169864078 on Build GRCh38
rs780377065 Chr.5: 169864071-169864071 on Build GRCh38

View Full Product Details