Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 170106051-170106553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXI1
Entrez Gene ID
2299
Gene Name
forkhead box I1
Gene Aliases
FKH10, FKHL10, FREAC-6, FREAC6, HFH-3, HFH3
UniGene
Hs.87236
Gene Chromosome Location
Chr.5: 170105897-170109725 on Build GRCh38
Transcripts
NM_012188.4 NM_144769.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3614070 c.99G>A p.E33E Chr.5: 170106056-170106056 on Build GRCh38
7199742 c.393C>A p.L131L Chr.5: 170106350-170106350 on Build GRCh38
5896765 c.151G>A p.G51R Chr.5: 170106108-170106108 on Build GRCh38
1435963 c.421G>A p.D141N Chr.5: 170106378-170106378 on Build GRCh38
1435961 c.232G>A p.G78S Chr.5: 170106189-170106189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200309754 Chr.5: 170106530-170106530 on Build GRCh38
rs142009641 Chr.5: 170106088-170106088 on Build GRCh38
rs745736470 Chr.5: 170106157-170106157 on Build GRCh38
rs934827575 Chr.5: 170106259-170106259 on Build GRCh38
rs774956405 Chr.5: 170106173-170106173 on Build GRCh38
rs772701997 Chr.5: 170106235-170106235 on Build GRCh38
rs750380368 Chr.5: 170106199-170106199 on Build GRCh38
rs769230925 Chr.5: 170106497-170106497 on Build GRCh38
rs749678694 Chr.5: 170106171-170106171 on Build GRCh38
rs748411850 Chr.5: 170106163-170106163 on Build GRCh38
rs748217286 Chr.5: 170106205-170106205 on Build GRCh38
rs765842378 Chr.5: 170106527-170106527 on Build GRCh38
rs562169481 Chr.5: 170106182-170106182 on Build GRCh38
rs747430896 Chr.5: 170106383-170106383 on Build GRCh38
rs754141285 Chr.5: 170106317-170106317 on Build GRCh38
rs770055886 Chr.5: 170106164-170106164 on Build GRCh38
rs759583461 Chr.5: 170106522-170106522 on Build GRCh38
rs774832918 Chr.5: 170106230-170106230 on Build GRCh38
rs1017078423 Chr.5: 170106324-170106324 on Build GRCh38
rs978862736 Chr.5: 170106210-170106210 on Build GRCh38
rs753661318 Chr.5: 170106139-170106139 on Build GRCh38
rs753370431 Chr.5: 170106194-170106194 on Build GRCh38
rs866622107 Chr.5: 170106251-170106251 on Build GRCh38
rs1051087019 Chr.5: 170106130-170106130 on Build GRCh38
rs750217079 Chr.5: 170106272-170106272 on Build GRCh38
rs763788629 Chr.5: 170106192-170106192 on Build GRCh38
rs373094576 Chr.5: 170106485-170106485 on Build GRCh38
rs747629075 Chr.5: 170106496-170106496 on Build GRCh38
rs764898591 Chr.5: 170106198-170106198 on Build GRCh38
rs769041762 Chr.5: 170106494-170106494 on Build GRCh38
rs757195384 Chr.5: 170106154-170106154 on Build GRCh38
rs2277944 Chr.5: 170106236-170106236 on Build GRCh38
rs769427090 Chr.5: 170106413-170106413 on Build GRCh38
rs889870074 Chr.5: 170106142-170106142 on Build GRCh38
rs771134802 Chr.5: 170106172-170106172 on Build GRCh38
rs962863313 Chr.5: 170106353-170106353 on Build GRCh38
rs766123812 Chr.5: 170106425-170106425 on Build GRCh38
rs756249041 Chr.5: 170106301-170106301 on Build GRCh38
rs770196010 Chr.5: 170106094-170106094 on Build GRCh38
rs376307456 Chr.5: 170106492-170106492 on Build GRCh38
rs778919285 Chr.5: 170106155-170106155 on Build GRCh38
rs761978045 Chr.5: 170106093-170106093 on Build GRCh38
rs773511000 Chr.5: 170106169-170106169 on Build GRCh38
rs114293092 Chr.5: 170106275-170106275 on Build GRCh38
rs772882906 Chr.5: 170106502-170106502 on Build GRCh38
rs757388303 Chr.5: 170106329-170106329 on Build GRCh38
rs746221300 Chr.5: 170106349-170106349 on Build GRCh38
rs553561553 Chr.5: 170106399-170106399 on Build GRCh38
rs762742077 Chr.5: 170106419-170106419 on Build GRCh38
rs763835524 Chr.5: 170106134-170106134 on Build GRCh38
rs199665479 Chr.5: 170106464-170106464 on Build GRCh38
rs138668268 Chr.5: 170106158-170106158 on Build GRCh38
rs534008931 Chr.5: 170106385-170106385 on Build GRCh38
rs746415631 Chr.5: 170106231-170106231 on Build GRCh38
rs148101435 Chr.5: 170106515-170106515 on Build GRCh38
rs764273040 Chr.5: 170106452-170106452 on Build GRCh38
rs757556947 Chr.5: 170106465-170106465 on Build GRCh38
rs759808329 Chr.5: 170106218-170106218 on Build GRCh38
rs373137429 Chr.5: 170106159-170106159 on Build GRCh38
rs756090467 Chr.5: 170106221-170106221 on Build GRCh38
rs866775728 Chr.5: 170106288-170106288 on Build GRCh38
rs753856999 Chr.5: 170106463-170106463 on Build GRCh38
rs766320745 Chr.5: 170106291-170106291 on Build GRCh38
rs780815772 Chr.5: 170106495-170106495 on Build GRCh38
rs772507619 Chr.5: 170106377-170106377 on Build GRCh38
rs377178658 Chr.5: 170106446-170106446 on Build GRCh38
rs535372917 Chr.5: 170106077-170106077 on Build GRCh38
rs910640993 Chr.5: 170106193-170106193 on Build GRCh38
rs973083998 Chr.5: 170106177-170106177 on Build GRCh38
rs760272275 Chr.5: 170106245-170106245 on Build GRCh38
rs771350065 Chr.5: 170106227-170106227 on Build GRCh38
rs370450076 Chr.5: 170106265-170106265 on Build GRCh38
rs775106921 Chr.5: 170106189-170106189 on Build GRCh38
rs758303475 Chr.5: 170106201-170106201 on Build GRCh38
rs550209761 Chr.5: 170106188-170106188 on Build GRCh38
rs773987289 Chr.5: 170106519-170106519 on Build GRCh38
rs779485173 Chr.5: 170106334-170106334 on Build GRCh38
rs766896773 Chr.5: 170106111-170106111 on Build GRCh38
rs767980791 Chr.5: 170106257-170106257 on Build GRCh38
rs779291702 Chr.5: 170106469-170106469 on Build GRCh38
rs933979218 Chr.5: 170106120-170106120 on Build GRCh38
rs751432686 Chr.5: 170106299-170106299 on Build GRCh38
rs781320501 Chr.5: 170106202-170106202 on Build GRCh38
rs767098191 Chr.5: 170106119-170106119 on Build GRCh38
rs561448899 Chr.5: 170106117-170106117 on Build GRCh38
rs758560898 Chr.5: 170106303-170106303 on Build GRCh38
rs941579883 Chr.5: 170106092-170106092 on Build GRCh38
rs547608599 Chr.5: 170106108-170106108 on Build GRCh38
rs1053809070 Chr.5: 170106264-170106264 on Build GRCh38
rs1029569674 Chr.5: 170106181-170106181 on Build GRCh38
rs752419262 Chr.5: 170106127-170106127 on Build GRCh38
rs778060920 Chr.5: 170106222-170106222 on Build GRCh38
rs886060398 Chr.5: 170106525-170106525 on Build GRCh38
rs768660570 Chr.5: 170106078-170106078 on Build GRCh38
rs1022683356 Chr.5: 170106086-170106086 on Build GRCh38
rs757076059 Chr.5: 170106196-170106196 on Build GRCh38
rs144559860 Chr.5: 170106440-170106440 on Build GRCh38
rs891739205 Chr.5: 170106213-170106213 on Build GRCh38
rs868704936 Chr.5: 170106268-170106268 on Build GRCh38
rs146116253 Chr.5: 170106116-170106116 on Build GRCh38
rs70979158 Chr.5: 170106128-170106129 on Build GRCh38
rs780706464 Chr.5: 170106382-170106382 on Build GRCh38
rs867801131 Chr.5: 170106450-170106450 on Build GRCh38
rs773599384 Chr.5: 170106098-170106098 on Build GRCh38
rs770762547 Chr.5: 170106518-170106518 on Build GRCh38
rs749433068 Chr.5: 170106226-170106226 on Build GRCh38
rs369758264 Chr.5: 170106075-170106075 on Build GRCh38
rs755803481 Chr.5: 170106133-170106133 on Build GRCh38
rs768231871 Chr.5: 170106384-170106384 on Build GRCh38

View Full Product Details