Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 170109499-170109998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXI1
Entrez Gene ID
2299
Gene Name
forkhead box I1
Gene Aliases
FKH10, FKHL10, FREAC-6, FREAC6, HFH-3, HFH3
UniGene
Hs.87236
Gene Chromosome Location
Chr.5: 170105897-170109725 on Build GRCh38
Transcripts
NM_012188.4 NM_144769.2

SNP Detail

SNP ID Chromosome Location
rs552935794 Chr.5: 170109869-170109869 on Build GRCh38
rs386405655 Chr.5: 170109543-170109544 on Build GRCh38
rs1050755698 Chr.5: 170109656-170109656 on Build GRCh38
rs545967111 Chr.5: 170109847-170109847 on Build GRCh38
rs1056493991 Chr.5: 170109754-170109754 on Build GRCh38
rs561784588 Chr.5: 170109902-170109902 on Build GRCh38
rs1043738882 Chr.5: 170109853-170109853 on Build GRCh38
rs530492087 Chr.5: 170109942-170109942 on Build GRCh38
rs869204485 Chr.5: 170109550-170109551 on Build GRCh38
rs887567341 Chr.5: 170109793-170109793 on Build GRCh38
rs1006471591 Chr.5: 170109962-170109962 on Build GRCh38
rs896663954 Chr.5: 170109883-170109883 on Build GRCh38
rs555325636 Chr.5: 170109764-170109764 on Build GRCh38
rs902055668 Chr.5: 170109834-170109834 on Build GRCh38
rs1005032697 Chr.5: 170109526-170109526 on Build GRCh38
rs35964510 Chr.5: 170109549-170109550 on Build GRCh38
rs977988172 Chr.5: 170109720-170109720 on Build GRCh38
rs867055048 Chr.5: 170109525-170109525 on Build GRCh38
rs947722091 Chr.5: 170109977-170109977 on Build GRCh38
rs113171964 Chr.5: 170109884-170109884 on Build GRCh38
rs778213515 Chr.5: 170109555-170109555 on Build GRCh38
rs571081281 Chr.5: 170109552-170109552 on Build GRCh38
rs111876035 Chr.5: 170109597-170109597 on Build GRCh38
rs1021712793 Chr.5: 170109860-170109860 on Build GRCh38
rs765045710 Chr.5: 170109904-170109904 on Build GRCh38
rs916525806 Chr.5: 170109908-170109908 on Build GRCh38
rs568876793 Chr.5: 170109765-170109765 on Build GRCh38
rs146942268 Chr.5: 170109740-170109740 on Build GRCh38
rs961021431 Chr.5: 170109542-170109543 on Build GRCh38
rs558826653 Chr.5: 170109864-170109864 on Build GRCh38
rs10057744 Chr.5: 170109750-170109750 on Build GRCh38
rs893022438 Chr.5: 170109846-170109846 on Build GRCh38
rs372184845 Chr.5: 170109807-170109807 on Build GRCh38
rs1048779998 Chr.5: 170109789-170109789 on Build GRCh38
rs902571257 Chr.5: 170109746-170109746 on Build GRCh38
rs775308176 Chr.5: 170109621-170109622 on Build GRCh38
rs997798388 Chr.5: 170109838-170109838 on Build GRCh38
rs960883419 Chr.5: 170109901-170109901 on Build GRCh38
rs1022356841 Chr.5: 170109744-170109744 on Build GRCh38
rs533523592 Chr.5: 170109556-170109556 on Build GRCh38
rs975828513 Chr.5: 170109796-170109796 on Build GRCh38
rs3839285 Chr.5: 170109541-170109542 on Build GRCh38
rs946511930 Chr.5: 170109654-170109654 on Build GRCh38
rs386405656 Chr.5: 170109548-170109549 on Build GRCh38
rs1050379839 Chr.5: 170109842-170109842 on Build GRCh38
rs947777750 Chr.5: 170109782-170109782 on Build GRCh38
rs546888065 Chr.5: 170109625-170109625 on Build GRCh38
rs529428860 Chr.5: 170109532-170109532 on Build GRCh38
rs953348322 Chr.5: 170109852-170109852 on Build GRCh38
rs1035240161 Chr.5: 170109536-170109536 on Build GRCh38
rs201086598 Chr.5: 170109541-170109541 on Build GRCh38
rs771802654 Chr.5: 170109562-170109562 on Build GRCh38
rs771997356 Chr.5: 170109774-170109774 on Build GRCh38
rs541982977 Chr.5: 170109896-170109896 on Build GRCh38
rs951763219 Chr.5: 170109792-170109793 on Build GRCh38
rs901666453 Chr.5: 170109645-170109645 on Build GRCh38
rs1011605850 Chr.5: 170109855-170109855 on Build GRCh38
rs10057859 Chr.5: 170109818-170109818 on Build GRCh38
rs914691555 Chr.5: 170109624-170109624 on Build GRCh38
rs537869040 Chr.5: 170109771-170109771 on Build GRCh38
rs952363349 Chr.5: 170109934-170109934 on Build GRCh38
rs1040197567 Chr.5: 170109626-170109626 on Build GRCh38
rs45466695 Chr.5: 170109535-170109535 on Build GRCh38
rs889824081 Chr.5: 170109739-170109739 on Build GRCh38
rs976455348 Chr.5: 170109551-170109551 on Build GRCh38
rs557172279 Chr.5: 170109794-170109794 on Build GRCh38
rs370208923 Chr.5: 170109767-170109767 on Build GRCh38

View Full Product Details