Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 170789051-170789559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRP
Entrez Gene ID
2568
Gene Name
gamma-aminobutyric acid type A receptor pi subunit
Gene Aliases
-
UniGene
Hs.26225
Gene Chromosome Location
Chr.5: 170783719-170814047 on Build GRCh38
Transcripts
NM_001291985.1 NM_014211.2 XM_005265872.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
123310 c.85G>A p.E29K Chr.5: 170789160-170789160 on Build GRCh38
6103407 c.169G>T p.G57C Chr.5: 170789244-170789244 on Build GRCh38
1065776 c.154C>A p.L52I Chr.5: 170789229-170789229 on Build GRCh38
1661942 c.103A>T p.K35* Chr.5: 170789178-170789178 on Build GRCh38
3783889 c.86A>G p.E29G Chr.5: 170789161-170789161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991944073 Chr.5: 170789150-170789150 on Build GRCh38
rs371029333 Chr.5: 170789225-170789225 on Build GRCh38
rs1030608679 Chr.5: 170789412-170789412 on Build GRCh38
rs967749671 Chr.5: 170789477-170789477 on Build GRCh38
rs765736197 Chr.5: 170789243-170789243 on Build GRCh38
rs766004118 Chr.5: 170789255-170789255 on Build GRCh38
rs368998560 Chr.5: 170789158-170789158 on Build GRCh38
rs377082764 Chr.5: 170789117-170789117 on Build GRCh38
rs749901338 Chr.5: 170789159-170789159 on Build GRCh38
rs374035124 Chr.5: 170789306-170789306 on Build GRCh38
rs146765294 Chr.5: 170789244-170789244 on Build GRCh38
rs575917022 Chr.5: 170789320-170789320 on Build GRCh38
rs2303134 Chr.5: 170789239-170789239 on Build GRCh38
rs1022662728 Chr.5: 170789219-170789219 on Build GRCh38
rs759688364 Chr.5: 170789105-170789105 on Build GRCh38
rs41275331 Chr.5: 170789504-170789504 on Build GRCh38
rs994265930 Chr.5: 170789075-170789075 on Build GRCh38
rs1000768261 Chr.5: 170789381-170789381 on Build GRCh38
rs553592344 Chr.5: 170789167-170789167 on Build GRCh38
rs757576786 Chr.5: 170789092-170789095 on Build GRCh38
rs752437512 Chr.5: 170789290-170789290 on Build GRCh38
rs190389282 Chr.5: 170789518-170789518 on Build GRCh38
rs891212770 Chr.5: 170789319-170789319 on Build GRCh38
rs377497557 Chr.5: 170789213-170789213 on Build GRCh38
rs748078594 Chr.5: 170789171-170789171 on Build GRCh38
rs141635070 Chr.5: 170789471-170789471 on Build GRCh38
rs940953504 Chr.5: 170789423-170789423 on Build GRCh38
rs1021630122 Chr.5: 170789468-170789468 on Build GRCh38
rs569473769 Chr.5: 170789512-170789512 on Build GRCh38
rs778878134 Chr.5: 170789190-170789190 on Build GRCh38
rs923624236 Chr.5: 170789144-170789144 on Build GRCh38
rs772234500 Chr.5: 170789216-170789216 on Build GRCh38
rs538571628 Chr.5: 170789076-170789076 on Build GRCh38
rs200679140 Chr.5: 170789133-170789133 on Build GRCh38
rs756019366 Chr.5: 170789181-170789181 on Build GRCh38
rs72837675 Chr.5: 170789186-170789186 on Build GRCh38
rs768730048 Chr.5: 170789296-170789296 on Build GRCh38
rs1006685392 Chr.5: 170789330-170789330 on Build GRCh38
rs373435065 Chr.5: 170789211-170789211 on Build GRCh38
rs143479635 Chr.5: 170789222-170789222 on Build GRCh38
rs939140041 Chr.5: 170789271-170789271 on Build GRCh38
rs189632864 Chr.5: 170789097-170789097 on Build GRCh38
rs999457372 Chr.5: 170789503-170789503 on Build GRCh38
rs775772483 Chr.5: 170789221-170789221 on Build GRCh38
rs909491355 Chr.5: 170789410-170789410 on Build GRCh38
rs768619421 Chr.5: 170789082-170789082 on Build GRCh38
rs761100924 Chr.5: 170789131-170789131 on Build GRCh38
rs979046144 Chr.5: 170789116-170789116 on Build GRCh38
rs1048717394 Chr.5: 170789379-170789379 on Build GRCh38
rs754554458 Chr.5: 170789283-170789283 on Build GRCh38
rs184276802 Chr.5: 170789301-170789301 on Build GRCh38
rs754395896 Chr.5: 170789149-170789149 on Build GRCh38
rs772285896 Chr.5: 170789120-170789120 on Build GRCh38
rs1012025802 Chr.5: 170789460-170789460 on Build GRCh38
rs564137370 Chr.5: 170789436-170789436 on Build GRCh38
rs544943289 Chr.5: 170789431-170789431 on Build GRCh38
rs777079055 Chr.5: 170789238-170789238 on Build GRCh38
rs560403598 Chr.5: 170789480-170789480 on Build GRCh38
rs1051223869 Chr.5: 170789305-170789305 on Build GRCh38
rs751207851 Chr.5: 170789270-170789270 on Build GRCh38
rs781085845 Chr.5: 170789083-170789083 on Build GRCh38
rs549294358 Chr.5: 170789509-170789509 on Build GRCh38
rs756034235 Chr.5: 170789297-170789297 on Build GRCh38
rs749427874 Chr.5: 170789089-170789089 on Build GRCh38
rs1020720829 Chr.5: 170789425-170789425 on Build GRCh38
rs775760884 Chr.5: 170789122-170789122 on Build GRCh38
rs1036536675 Chr.5: 170789429-170789429 on Build GRCh38
rs571307888 Chr.5: 170789140-170789140 on Build GRCh38
rs533159333 Chr.5: 170789469-170789469 on Build GRCh38
rs139832992 Chr.5: 170789166-170789166 on Build GRCh38
rs748247445 Chr.5: 170789430-170789430 on Build GRCh38
rs767308541 Chr.5: 170789287-170789287 on Build GRCh38
rs751402272 Chr.5: 170789165-170789165 on Build GRCh38
rs770991075 Chr.5: 170789091-170789091 on Build GRCh38
rs528267016 Chr.5: 170789508-170789508 on Build GRCh38
rs778561776 Chr.5: 170789156-170789156 on Build GRCh38
rs200140579 Chr.5: 170789160-170789160 on Build GRCh38
rs145941447 Chr.5: 170789157-170789157 on Build GRCh38
rs781318764 Chr.5: 170789352-170789352 on Build GRCh38
rs889193656 Chr.5: 170789456-170789456 on Build GRCh38

View Full Product Details