Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 170805628-170806126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRP
Entrez Gene ID
2568
Gene Name
gamma-aminobutyric acid type A receptor pi subunit
Gene Aliases
-
UniGene
Hs.26225
Gene Chromosome Location
Chr.5: 170783719-170814047 on Build GRCh38
Transcripts
NM_001291985.1 NM_014211.2 XM_005265872.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3429274 c.598C>T p.R200C Chr.5: 170805772-170805772 on Build GRCh38
6211324 c.579G>A p.L193L Chr.5: 170805753-170805753 on Build GRCh38
338352 c.553G>A p.G185R Chr.5: 170805727-170805727 on Build GRCh38
377680 c.559G>A p.D187N Chr.5: 170805733-170805733 on Build GRCh38
4686170 c.589G>A p.D197N Chr.5: 170805763-170805763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751847592 Chr.5: 170805853-170805853 on Build GRCh38
rs984361429 Chr.5: 170805858-170805858 on Build GRCh38
rs766395776 Chr.5: 170805954-170805954 on Build GRCh38
rs1038098604 Chr.5: 170805952-170805952 on Build GRCh38
rs200102540 Chr.5: 170805681-170805681 on Build GRCh38
rs769073805 Chr.5: 170805806-170805806 on Build GRCh38
rs539418830 Chr.5: 170805967-170805967 on Build GRCh38
rs28652836 Chr.5: 170805859-170805859 on Build GRCh38
rs373600124 Chr.5: 170805676-170805676 on Build GRCh38
rs762621670 Chr.5: 170805757-170805757 on Build GRCh38
rs748713828 Chr.5: 170805813-170805813 on Build GRCh38
rs201137222 Chr.5: 170805814-170805814 on Build GRCh38
rs371669810 Chr.5: 170805872-170805872 on Build GRCh38
rs376775791 Chr.5: 170805825-170805825 on Build GRCh38
rs886042237 Chr.5: 170805781-170805781 on Build GRCh38
rs1017886046 Chr.5: 170806053-170806053 on Build GRCh38
rs766181531 Chr.5: 170805677-170805677 on Build GRCh38
rs150667747 Chr.5: 170805791-170805791 on Build GRCh38
rs537718477 Chr.5: 170806094-170806094 on Build GRCh38
rs371756685 Chr.5: 170805685-170805685 on Build GRCh38
rs773791819 Chr.5: 170805887-170805887 on Build GRCh38
rs758743196 Chr.5: 170805790-170805790 on Build GRCh38
rs752383357 Chr.5: 170805836-170805836 on Build GRCh38
rs138553703 Chr.5: 170805751-170805751 on Build GRCh38
rs751400203 Chr.5: 170805763-170805763 on Build GRCh38
rs929475651 Chr.5: 170805999-170805999 on Build GRCh38
rs562309526 Chr.5: 170805865-170805865 on Build GRCh38
rs770623496 Chr.5: 170805817-170805817 on Build GRCh38
rs768447552 Chr.5: 170805714-170805715 on Build GRCh38
rs1041176500 Chr.5: 170805662-170805662 on Build GRCh38
rs751458302 Chr.5: 170806095-170806095 on Build GRCh38
rs932421822 Chr.5: 170805940-170805940 on Build GRCh38
rs772546066 Chr.5: 170805753-170805753 on Build GRCh38
rs137960338 Chr.5: 170805819-170805819 on Build GRCh38
rs74966184 Chr.5: 170805698-170805698 on Build GRCh38
rs149140731 Chr.5: 170805764-170805764 on Build GRCh38
rs112704076 Chr.5: 170805773-170805773 on Build GRCh38
rs954969309 Chr.5: 170805702-170805702 on Build GRCh38
rs556216691 Chr.5: 170805700-170805700 on Build GRCh38
rs747449774 Chr.5: 170805796-170805796 on Build GRCh38
rs758547993 Chr.5: 170805845-170805845 on Build GRCh38
rs761651594 Chr.5: 170805861-170805862 on Build GRCh38
rs149480838 Chr.5: 170805846-170805846 on Build GRCh38
rs757352374 Chr.5: 170805688-170805688 on Build GRCh38
rs879020256 Chr.5: 170805731-170805731 on Build GRCh38
rs756689094 Chr.5: 170805860-170805860 on Build GRCh38
rs376679605 Chr.5: 170805891-170805891 on Build GRCh38
rs201289609 Chr.5: 170805749-170805749 on Build GRCh38
rs781641550 Chr.5: 170805854-170805854 on Build GRCh38
rs764160496 Chr.5: 170805902-170805902 on Build GRCh38
rs754677208 Chr.5: 170805901-170805901 on Build GRCh38
rs771594660 Chr.5: 170805893-170805893 on Build GRCh38
rs752682296 Chr.5: 170805768-170805768 on Build GRCh38
rs778235609 Chr.5: 170805867-170805867 on Build GRCh38
rs569045954 Chr.5: 170806086-170806086 on Build GRCh38
rs1013000809 Chr.5: 170806089-170806089 on Build GRCh38
rs73327149 Chr.5: 170806068-170806068 on Build GRCh38
rs773956363 Chr.5: 170805768-170805769 on Build GRCh38
rs1049508263 Chr.5: 170805946-170805946 on Build GRCh38
rs751242788 Chr.5: 170805907-170805909 on Build GRCh38
rs867138983 Chr.5: 170806033-170806033 on Build GRCh38
rs946581268 Chr.5: 170805991-170805991 on Build GRCh38
rs922311417 Chr.5: 170805931-170805931 on Build GRCh38
rs772816775 Chr.5: 170805815-170805815 on Build GRCh38
rs1042266548 Chr.5: 170806007-170806007 on Build GRCh38
rs780793952 Chr.5: 170805960-170805960 on Build GRCh38
rs368063746 Chr.5: 170805680-170805680 on Build GRCh38
rs143232586 Chr.5: 170805900-170805900 on Build GRCh38
rs776718807 Chr.5: 170805904-170805904 on Build GRCh38
rs755519081 Chr.5: 170806019-170806019 on Build GRCh38
rs781357641 Chr.5: 170805812-170805812 on Build GRCh38
rs759120936 Chr.5: 170805834-170805834 on Build GRCh38
rs369751716 Chr.5: 170805835-170805835 on Build GRCh38
rs770431393 Chr.5: 170805875-170805875 on Build GRCh38
rs767418400 Chr.5: 170805884-170805885 on Build GRCh38
rs79061387 Chr.5: 170805772-170805772 on Build GRCh38
rs747356154 Chr.5: 170805703-170805703 on Build GRCh38
rs555225645 Chr.5: 170806082-170806082 on Build GRCh38
rs765981038 Chr.5: 170805762-170805762 on Build GRCh38
rs190950426 Chr.5: 170805770-170805770 on Build GRCh38
rs570324672 Chr.5: 170805918-170805918 on Build GRCh38
rs763917484 Chr.5: 170805840-170805840 on Build GRCh38
rs546654460 Chr.5: 170805998-170805998 on Build GRCh38
rs762424354 Chr.5: 170805816-170805816 on Build GRCh38
rs368617970 Chr.5: 170805744-170805744 on Build GRCh38
rs201676692 Chr.5: 170805745-170805745 on Build GRCh38
rs748426651 Chr.5: 170805857-170805857 on Build GRCh38
rs548370382 Chr.5: 170805839-170805839 on Build GRCh38
rs747689817 Chr.5: 170806009-170806009 on Build GRCh38
rs746099867 Chr.5: 170805697-170805697 on Build GRCh38
rs778790224 Chr.5: 170805695-170805695 on Build GRCh38
rs566564944 Chr.5: 170806013-170806013 on Build GRCh38
rs769348904 Chr.5: 170805722-170805722 on Build GRCh38
rs1036159812 Chr.5: 170805781-170805781 on Build GRCh38

View Full Product Details