Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 171862458-171863000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW11
Entrez Gene ID
23291
Gene Name
F-box and WD repeat domain containing 11
Gene Aliases
BTRC2, BTRCP2, FBW1B, FBXW1B, Fbw11, Hos
UniGene
Hs.484138
Gene Chromosome Location
Chr.5: 171861549-172006873 on Build GRCh38
Transcripts
NM_012300.2 NM_033644.2 NM_033645.2 XM_005265855.4 XM_005265856.2 XM_005265857.2 XM_005265858.2 XM_017009279.1 XM_017009280.1 XM_017009281.1

SNP Detail

SNP ID Chromosome Location
rs780301641 Chr.5: 171862589-171862589 on Build GRCh38
rs554132977 Chr.5: 171862574-171862574 on Build GRCh38
rs768891795 Chr.5: 171862641-171862641 on Build GRCh38
rs929785376 Chr.5: 171862663-171862663 on Build GRCh38
rs756203448 Chr.5: 171862485-171862485 on Build GRCh38
rs937050296 Chr.5: 171862628-171862628 on Build GRCh38
rs895772183 Chr.5: 171862647-171862647 on Build GRCh38
rs572571640 Chr.5: 171862843-171862843 on Build GRCh38
rs189658721 Chr.5: 171862704-171862704 on Build GRCh38
rs7715661 Chr.5: 171862864-171862864 on Build GRCh38
rs536924358 Chr.5: 171862622-171862622 on Build GRCh38
rs956853565 Chr.5: 171862513-171862513 on Build GRCh38
rs988307769 Chr.5: 171862548-171862548 on Build GRCh38
rs181645006 Chr.5: 171862764-171862764 on Build GRCh38
rs1005659865 Chr.5: 171862848-171862848 on Build GRCh38
rs772385895 Chr.5: 171862893-171862893 on Build GRCh38
rs912976601 Chr.5: 171862552-171862552 on Build GRCh38
rs1022423456 Chr.5: 171862934-171862934 on Build GRCh38
rs956986797 Chr.5: 171862914-171862914 on Build GRCh38
rs1030324953 Chr.5: 171862895-171862895 on Build GRCh38
rs186680490 Chr.5: 171862896-171862896 on Build GRCh38
rs184694895 Chr.5: 171862630-171862630 on Build GRCh38
rs555620930 Chr.5: 171862608-171862608 on Build GRCh38
rs899821137 Chr.5: 171862865-171862865 on Build GRCh38
rs1042664265 Chr.5: 171862607-171862607 on Build GRCh38
rs537383097 Chr.5: 171862579-171862579 on Build GRCh38
rs888491932 Chr.5: 171862748-171862748 on Build GRCh38
rs774486945 Chr.5: 171862674-171862674 on Build GRCh38
rs757150046 Chr.5: 171862723-171862725 on Build GRCh38
rs561252576 Chr.5: 171862916-171862916 on Build GRCh38
rs1046937073 Chr.5: 171862730-171862730 on Build GRCh38
rs565336271 Chr.5: 171862787-171862787 on Build GRCh38

View Full Product Details