Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 171876206-171876611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW11
Entrez Gene ID
23291
Gene Name
F-box and WD repeat domain containing 11
Gene Aliases
BTRC2, BTRCP2, FBW1B, FBXW1B, Fbw11, Hos
UniGene
Hs.484138
Gene Chromosome Location
Chr.5: 171861549-172006873 on Build GRCh38
Transcripts
NM_012300.2 NM_033644.2 NM_033645.2 XM_005265855.4 XM_005265856.2 XM_005265857.2 XM_005265858.2 XM_017009279.1 XM_017009280.1 XM_017009281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4346951 c.1049G>A p.R350Q Chr.5: 171876355-171876355 on Build GRCh38
256949 c.1002G>A p.M334I Chr.5: 171876402-171876402 on Build GRCh38
5808363 c.1096G>T p.A366S Chr.5: 171876308-171876308 on Build GRCh38
1065803 c.1067G>A p.R356H Chr.5: 171876376-171876376 on Build GRCh38
1065805 c.1053C>T p.T351T Chr.5: 171876390-171876390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746684500 Chr.5: 171876516-171876516 on Build GRCh38
rs10052973 Chr.5: 171876473-171876473 on Build GRCh38
rs937377142 Chr.5: 171876572-171876572 on Build GRCh38
rs766311423 Chr.5: 171876549-171876549 on Build GRCh38
rs773251633 Chr.5: 171876546-171876546 on Build GRCh38
rs756315604 Chr.5: 171876303-171876303 on Build GRCh38
rs1043193213 Chr.5: 171876543-171876543 on Build GRCh38
rs779641836 Chr.5: 171876501-171876501 on Build GRCh38
rs201522540 Chr.5: 171876378-171876378 on Build GRCh38
rs868510910 Chr.5: 171876425-171876425 on Build GRCh38
rs746192325 Chr.5: 171876497-171876497 on Build GRCh38
rs762550563 Chr.5: 171876573-171876573 on Build GRCh38
rs758639751 Chr.5: 171876583-171876583 on Build GRCh38
rs935050428 Chr.5: 171876254-171876254 on Build GRCh38
rs367673917 Chr.5: 171876556-171876556 on Build GRCh38
rs764962009 Chr.5: 171876244-171876244 on Build GRCh38
rs772522909 Chr.5: 171876498-171876498 on Build GRCh38
rs778024564 Chr.5: 171876474-171876474 on Build GRCh38
rs776500801 Chr.5: 171876377-171876377 on Build GRCh38
rs571392332 Chr.5: 171876426-171876426 on Build GRCh38
rs769606161 Chr.5: 171876539-171876539 on Build GRCh38
rs750181953 Chr.5: 171876393-171876393 on Build GRCh38
rs771606804 Chr.5: 171876342-171876342 on Build GRCh38
rs749845697 Chr.5: 171876317-171876317 on Build GRCh38
rs769321307 Chr.5: 171876527-171876527 on Build GRCh38
rs548680205 Chr.5: 171876517-171876517 on Build GRCh38
rs377105632 Chr.5: 171876547-171876547 on Build GRCh38
rs199594215 Chr.5: 171876438-171876438 on Build GRCh38
rs891990894 Chr.5: 171876318-171876318 on Build GRCh38
rs764474794 Chr.5: 171876559-171876559 on Build GRCh38
rs188270483 Chr.5: 171876551-171876551 on Build GRCh38
rs146880247 Chr.5: 171876468-171876468 on Build GRCh38
rs779530352 Chr.5: 171876484-171876484 on Build GRCh38
rs757697590 Chr.5: 171876480-171876480 on Build GRCh38
rs781546756 Chr.5: 171876273-171876273 on Build GRCh38
rs974685829 Chr.5: 171876394-171876394 on Build GRCh38
rs748226503 Chr.5: 171876280-171876280 on Build GRCh38
rs776121848 Chr.5: 171876537-171876537 on Build GRCh38
rs771280574 Chr.5: 171876321-171876321 on Build GRCh38
rs538479066 Chr.5: 171876327-171876327 on Build GRCh38
rs775168567 Chr.5: 171876348-171876348 on Build GRCh38
rs373380893 Chr.5: 171876235-171876235 on Build GRCh38
rs757005743 Chr.5: 171876240-171876240 on Build GRCh38
rs768185805 Chr.5: 171876359-171876359 on Build GRCh38
rs371553236 Chr.5: 171876586-171876586 on Build GRCh38
rs200425757 Chr.5: 171876253-171876253 on Build GRCh38
rs760146583 Chr.5: 171876357-171876357 on Build GRCh38
rs779254554 Chr.5: 171876575-171876575 on Build GRCh38
rs750970614 Chr.5: 171876582-171876582 on Build GRCh38
rs61749649 Chr.5: 171876315-171876315 on Build GRCh38
rs371040715 Chr.5: 171876569-171876569 on Build GRCh38
rs372901835 Chr.5: 171876413-171876413 on Build GRCh38
rs751746261 Chr.5: 171876246-171876246 on Build GRCh38
rs369137834 Chr.5: 171876390-171876390 on Build GRCh38
rs549613848 Chr.5: 171876233-171876233 on Build GRCh38
rs746307930 Chr.5: 171876329-171876329 on Build GRCh38
rs754342662 Chr.5: 171876477-171876477 on Build GRCh38
rs761453523 Chr.5: 171876538-171876538 on Build GRCh38
rs755161123 Chr.5: 171876269-171876269 on Build GRCh38

View Full Product Details