Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 172923920-172924426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERGIC1
Entrez Gene ID
57222
Gene Name
endoplasmic reticulum-golgi intermediate compartment 1
Gene Aliases
ERGIC-32, ERGIC32, NET24
UniGene
Hs.509163
Gene Chromosome Location
Chr.5: 172834220-172952685 on Build GRCh38
Transcripts
NM_001031711.2 XM_006714892.1 XM_011534597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4640652 c.380C>A p.P127H Chr.5: 172924009-172924009 on Build GRCh38
1065875 c.394G>A p.V132M Chr.5: 172924023-172924023 on Build GRCh38
4494337 c.430C>T p.P144S Chr.5: 172924059-172924059 on Build GRCh38
737697 c.441G>T p.T147T Chr.5: 172924070-172924070 on Build GRCh38
1642678 c.440C>T p.T147M Chr.5: 172924069-172924069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2302959 Chr.5: 172924311-172924311 on Build GRCh38
rs188394855 Chr.5: 172924399-172924399 on Build GRCh38
rs982841738 Chr.5: 172924343-172924343 on Build GRCh38
rs767023837 Chr.5: 172924011-172924011 on Build GRCh38
rs549668819 Chr.5: 172924209-172924209 on Build GRCh38
rs1035456688 Chr.5: 172924296-172924296 on Build GRCh38
rs758217254 Chr.5: 172924086-172924086 on Build GRCh38
rs769945953 Chr.5: 172923980-172923980 on Build GRCh38
rs775215957 Chr.5: 172924022-172924022 on Build GRCh38
rs772520694 Chr.5: 172924158-172924158 on Build GRCh38
rs760335399 Chr.5: 172924265-172924265 on Build GRCh38
rs186622895 Chr.5: 172924204-172924204 on Build GRCh38
rs934582100 Chr.5: 172924098-172924098 on Build GRCh38
rs897395390 Chr.5: 172923944-172923944 on Build GRCh38
rs2302958 Chr.5: 172924157-172924157 on Build GRCh38
rs565001351 Chr.5: 172924067-172924067 on Build GRCh38
rs566068168 Chr.5: 172924126-172924126 on Build GRCh38
rs965426392 Chr.5: 172924290-172924290 on Build GRCh38
rs191464312 Chr.5: 172924219-172924219 on Build GRCh38
rs754145170 Chr.5: 172924317-172924317 on Build GRCh38
rs144585333 Chr.5: 172924264-172924264 on Build GRCh38
rs569596264 Chr.5: 172924387-172924387 on Build GRCh38
rs550378442 Chr.5: 172924388-172924388 on Build GRCh38
rs373905217 Chr.5: 172923954-172923954 on Build GRCh38
rs201269025 Chr.5: 172924036-172924036 on Build GRCh38
rs867307182 Chr.5: 172924400-172924400 on Build GRCh38
rs892777875 Chr.5: 172924242-172924242 on Build GRCh38
rs889964445 Chr.5: 172924246-172924246 on Build GRCh38
rs755140740 Chr.5: 172923955-172923955 on Build GRCh38
rs771467676 Chr.5: 172923989-172923989 on Build GRCh38
rs781489853 Chr.5: 172923965-172923965 on Build GRCh38
rs887400610 Chr.5: 172924182-172924182 on Build GRCh38
rs767035538 Chr.5: 172924141-172924141 on Build GRCh38
rs991515267 Chr.5: 172924312-172924312 on Build GRCh38
rs1020614621 Chr.5: 172924250-172924250 on Build GRCh38
rs7724177 Chr.5: 172924127-172924127 on Build GRCh38
rs370443963 Chr.5: 172924132-172924132 on Build GRCh38
rs551447874 Chr.5: 172924275-172924275 on Build GRCh38
rs111880705 Chr.5: 172924112-172924112 on Build GRCh38
rs774694436 Chr.5: 172923996-172923996 on Build GRCh38
rs908541218 Chr.5: 172924354-172924354 on Build GRCh38
rs781230483 Chr.5: 172924103-172924103 on Build GRCh38
rs763635234 Chr.5: 172924054-172924054 on Build GRCh38
rs896781755 Chr.5: 172924214-172924214 on Build GRCh38
rs369506310 Chr.5: 172923948-172923948 on Build GRCh38
rs554042356 Chr.5: 172924319-172924319 on Build GRCh38
rs1019627556 Chr.5: 172923999-172923999 on Build GRCh38
rs757251614 Chr.5: 172924069-172924069 on Build GRCh38
rs573894705 Chr.5: 172924323-172924323 on Build GRCh38
rs928472645 Chr.5: 172924028-172924028 on Build GRCh38
rs1011271842 Chr.5: 172924245-172924245 on Build GRCh38
rs748107663 Chr.5: 172923974-172923974 on Build GRCh38
rs773661322 Chr.5: 172923987-172923987 on Build GRCh38
rs777637919 Chr.5: 172923958-172923958 on Build GRCh38
rs967280771 Chr.5: 172924261-172924261 on Build GRCh38
rs944237610 Chr.5: 172924357-172924357 on Build GRCh38
rs149317716 Chr.5: 172924210-172924210 on Build GRCh38
rs562604051 Chr.5: 172923988-172923988 on Build GRCh38
rs777560910 Chr.5: 172924298-172924298 on Build GRCh38
rs190785521 Chr.5: 172924010-172924010 on Build GRCh38
rs373775703 Chr.5: 172924140-172924140 on Build GRCh38
rs1006999068 Chr.5: 172924265-172924266 on Build GRCh38
rs750184723 Chr.5: 172924078-172924078 on Build GRCh38
rs753340202 Chr.5: 172924070-172924070 on Build GRCh38
rs182882572 Chr.5: 172924289-172924289 on Build GRCh38
rs192733506 Chr.5: 172924402-172924402 on Build GRCh38
rs183246461 Chr.5: 172924023-172924023 on Build GRCh38
rs369931035 Chr.5: 172924194-172924194 on Build GRCh38
rs758187129 Chr.5: 172924189-172924189 on Build GRCh38
rs781165252 Chr.5: 172924389-172924389 on Build GRCh38
rs748425641 Chr.5: 172924124-172924124 on Build GRCh38
rs952814468 Chr.5: 172924328-172924328 on Build GRCh38

View Full Product Details