Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 172951813-172952320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERGIC1
Entrez Gene ID
57222
Gene Name
endoplasmic reticulum-golgi intermediate compartment 1
Gene Aliases
ERGIC-32, ERGIC32, NET24
UniGene
Hs.509163
Gene Chromosome Location
Chr.5: 172834220-172952685 on Build GRCh38
Transcripts
NM_001031711.2 XM_006714892.1 XM_011534597.1

SNP Detail

SNP ID Chromosome Location
rs972126399 Chr.5: 172952051-172952051 on Build GRCh38
rs763184236 Chr.5: 172951884-172951884 on Build GRCh38
rs577904946 Chr.5: 172951959-172951959 on Build GRCh38
rs764400185 Chr.5: 172951889-172951889 on Build GRCh38
rs386695023 Chr.5: 172951853-172951854 on Build GRCh38
rs893835128 Chr.5: 172951947-172951947 on Build GRCh38
rs919662450 Chr.5: 172952090-172952090 on Build GRCh38
rs191784867 Chr.5: 172951996-172951996 on Build GRCh38
rs377054252 Chr.5: 172952043-172952043 on Build GRCh38
rs899924441 Chr.5: 172951915-172951915 on Build GRCh38
rs943920511 Chr.5: 172952213-172952213 on Build GRCh38
rs149464075 Chr.5: 172951866-172951866 on Build GRCh38
rs1053820959 Chr.5: 172952269-172952269 on Build GRCh38
rs900954301 Chr.5: 172951930-172951930 on Build GRCh38
rs1023709688 Chr.5: 172952288-172952288 on Build GRCh38
rs566588833 Chr.5: 172952019-172952019 on Build GRCh38
rs1003720973 Chr.5: 172952024-172952024 on Build GRCh38
rs372670359 Chr.5: 172951918-172951918 on Build GRCh38
rs537341026 Chr.5: 172952012-172952012 on Build GRCh38
rs557676674 Chr.5: 172951980-172951981 on Build GRCh38
rs1040922066 Chr.5: 172952195-172952195 on Build GRCh38
rs932486819 Chr.5: 172952263-172952263 on Build GRCh38
rs866128445 Chr.5: 172952127-172952127 on Build GRCh38
rs1041250463 Chr.5: 172951910-172951910 on Build GRCh38
rs893867728 Chr.5: 172952278-172952278 on Build GRCh38
rs943906501 Chr.5: 172951908-172951908 on Build GRCh38
rs901007822 Chr.5: 172952264-172952264 on Build GRCh38
rs548426932 Chr.5: 172951931-172951931 on Build GRCh38
rs539936 Chr.5: 172951854-172951854 on Build GRCh38
rs774748166 Chr.5: 172952074-172952074 on Build GRCh38
rs1036356793 Chr.5: 172951863-172951863 on Build GRCh38
rs908441819 Chr.5: 172951905-172951905 on Build GRCh38
rs138735066 Chr.5: 172952096-172952096 on Build GRCh38
rs976618280 Chr.5: 172952214-172952214 on Build GRCh38
rs991270894 Chr.5: 172952077-172952077 on Build GRCh38
rs142781394 Chr.5: 172952279-172952279 on Build GRCh38
rs962142102 Chr.5: 172952044-172952044 on Build GRCh38
rs532677842 Chr.5: 172951857-172951857 on Build GRCh38
rs921128600 Chr.5: 172952232-172952232 on Build GRCh38
rs1042335240 Chr.5: 172952289-172952289 on Build GRCh38
rs369015985 Chr.5: 172952295-172952297 on Build GRCh38
rs550743249 Chr.5: 172952035-172952035 on Build GRCh38
rs982332078 Chr.5: 172952037-172952037 on Build GRCh38
rs544624235 Chr.5: 172952293-172952295 on Build GRCh38
rs982421495 Chr.5: 172952145-172952145 on Build GRCh38
rs959564328 Chr.5: 172952041-172952041 on Build GRCh38
rs147158105 Chr.5: 172951919-172951919 on Build GRCh38
rs986518050 Chr.5: 172952094-172952094 on Build GRCh38
rs539766065 Chr.5: 172952206-172952206 on Build GRCh38
rs60165841 Chr.5: 172951853-172951853 on Build GRCh38
rs573357861 Chr.5: 172952021-172952022 on Build GRCh38
rs1021179636 Chr.5: 172951960-172951960 on Build GRCh38
rs930679842 Chr.5: 172952070-172952070 on Build GRCh38
rs769797759 Chr.5: 172951851-172951851 on Build GRCh38
rs868598986 Chr.5: 172951885-172951885 on Build GRCh38
rs908470708 Chr.5: 172952152-172952152 on Build GRCh38
rs982664470 Chr.5: 172951838-172951838 on Build GRCh38
rs1030715629 Chr.5: 172951968-172951968 on Build GRCh38
rs893486179 Chr.5: 172951977-172951977 on Build GRCh38
rs920650714 Chr.5: 172952054-172952054 on Build GRCh38

View Full Product Details