Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173086327-173086738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBRF
Entrez Gene ID
153222
Gene Name
CREB3 regulatory factor
Gene Aliases
C5orf41, LRF
UniGene
Hs.484195
Gene Chromosome Location
Chr.5: 173056349-173139288 on Build GRCh38
Transcripts
NM_001168393.1 NM_001168394.1 NM_153607.2 XM_005265821.3 XM_006714822.3 XM_011534442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3339800 c.69G>A p.S23S Chr.5: 173086560-173086560 on Build GRCh38
5663474 c.126G>A p.M42I Chr.5: 173086617-173086617 on Build GRCh38
3744782 c.135+2T>C p.? Chr.5: 173086628-173086628 on Build GRCh38
3414562 c.10-9C>T p.? Chr.5: 173086492-173086492 on Build GRCh38
6635375 c.111G>A p.S37S Chr.5: 173086602-173086602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765387706 Chr.5: 173086479-173086479 on Build GRCh38
rs986088533 Chr.5: 173086406-173086406 on Build GRCh38
rs915443305 Chr.5: 173086513-173086513 on Build GRCh38
rs747504238 Chr.5: 173086509-173086509 on Build GRCh38
rs952366095 Chr.5: 173086687-173086687 on Build GRCh38
rs750373522 Chr.5: 173086480-173086480 on Build GRCh38
rs758436102 Chr.5: 173086492-173086492 on Build GRCh38
rs1053808083 Chr.5: 173086389-173086389 on Build GRCh38
rs747279375 Chr.5: 173086515-173086515 on Build GRCh38
rs774061629 Chr.5: 173086560-173086560 on Build GRCh38
rs756404711 Chr.5: 173086650-173086650 on Build GRCh38
rs938957069 Chr.5: 173086708-173086708 on Build GRCh38
rs770773866 Chr.5: 173086663-173086663 on Build GRCh38
rs760405809 Chr.5: 173086577-173086577 on Build GRCh38
rs1009852422 Chr.5: 173086404-173086404 on Build GRCh38
rs146782259 Chr.5: 173086572-173086572 on Build GRCh38
rs1003230872 Chr.5: 173086358-173086358 on Build GRCh38
rs751645985 Chr.5: 173086608-173086608 on Build GRCh38
rs780159169 Chr.5: 173086498-173086498 on Build GRCh38
rs752605650 Chr.5: 173086464-173086464 on Build GRCh38
rs112043212 Chr.5: 173086373-173086373 on Build GRCh38
rs558772543 Chr.5: 173086362-173086362 on Build GRCh38
rs138913647 Chr.5: 173086593-173086593 on Build GRCh38
rs537391528 Chr.5: 173086524-173086524 on Build GRCh38
rs958450672 Chr.5: 173086595-173086595 on Build GRCh38
rs779460226 Chr.5: 173086445-173086445 on Build GRCh38
rs115235376 Chr.5: 173086510-173086510 on Build GRCh38
rs770323307 Chr.5: 173086554-173086554 on Build GRCh38
rs745380832 Chr.5: 173086678-173086678 on Build GRCh38
rs750465383 Chr.5: 173086489-173086489 on Build GRCh38
rs112359463 Chr.5: 173086418-173086418 on Build GRCh38
rs768810893 Chr.5: 173086667-173086667 on Build GRCh38
rs538900034 Chr.5: 173086437-173086438 on Build GRCh38
rs138562616 Chr.5: 173086512-173086512 on Build GRCh38
rs982685404 Chr.5: 173086708-173086708 on Build GRCh38
rs554866103 Chr.5: 173086400-173086400 on Build GRCh38
rs1003707861 Chr.5: 173086543-173086543 on Build GRCh38
rs765456881 Chr.5: 173086592-173086592 on Build GRCh38
rs143944605 Chr.5: 173086567-173086567 on Build GRCh38
rs371639516 Chr.5: 173086523-173086523 on Build GRCh38
rs376606038 Chr.5: 173086602-173086602 on Build GRCh38
rs769439937 Chr.5: 173086531-173086531 on Build GRCh38
rs772772961 Chr.5: 173086536-173086536 on Build GRCh38
rs755424130 Chr.5: 173086615-173086615 on Build GRCh38
rs778247087 Chr.5: 173086653-173086653 on Build GRCh38
rs748475579 Chr.5: 173086530-173086530 on Build GRCh38
rs748505426 Chr.5: 173086641-173086641 on Build GRCh38
rs748878172 Chr.5: 173086654-173086654 on Build GRCh38
rs748514438 Chr.5: 173086501-173086501 on Build GRCh38
rs200257911 Chr.5: 173086514-173086514 on Build GRCh38
rs758706916 Chr.5: 173086597-173086597 on Build GRCh38
rs775362830 Chr.5: 173086574-173086574 on Build GRCh38
rs897579063 Chr.5: 173086373-173086373 on Build GRCh38
rs192472753 Chr.5: 173086675-173086675 on Build GRCh38
rs577220927 Chr.5: 173086405-173086405 on Build GRCh38
rs749011980 Chr.5: 173086538-173086538 on Build GRCh38
rs534739215 Chr.5: 173086376-173086377 on Build GRCh38
rs781680360 Chr.5: 173086634-173086634 on Build GRCh38
rs757429860 Chr.5: 173086470-173086470 on Build GRCh38

View Full Product Details