Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173090176-173090695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBRF
Entrez Gene ID
153222
Gene Name
CREB3 regulatory factor
Gene Aliases
C5orf41, LRF
UniGene
Hs.484195
Gene Chromosome Location
Chr.5: 173056349-173139288 on Build GRCh38
Transcripts
NM_001168393.1 NM_001168394.1 NM_153607.2 XM_005265821.3 XM_006714822.3 XM_011534442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6200560 c.493C>A p.Q165K Chr.5: 173090672-173090672 on Build GRCh38
4653944 c.357T>C p.G119G Chr.5: 173090536-173090536 on Build GRCh38
1237727 c.317G>T p.S106I Chr.5: 173090496-173090496 on Build GRCh38
6635374 c.246T>C p.G82G Chr.5: 173090425-173090425 on Build GRCh38
5011551 c.273G>T p.W91C Chr.5: 173090452-173090452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746918244 Chr.5: 173090437-173090437 on Build GRCh38
rs779661851 Chr.5: 173090435-173090435 on Build GRCh38
rs764674724 Chr.5: 173090572-173090572 on Build GRCh38
rs538056249 Chr.5: 173090230-173090230 on Build GRCh38
rs1008740292 Chr.5: 173090337-173090337 on Build GRCh38
rs745327754 Chr.5: 173090426-173090426 on Build GRCh38
rs776235463 Chr.5: 173090471-173090471 on Build GRCh38
rs767574002 Chr.5: 173090546-173090546 on Build GRCh38
rs759644003 Chr.5: 173090357-173090357 on Build GRCh38
rs775317490 Chr.5: 173090296-173090296 on Build GRCh38
rs761622677 Chr.5: 173090475-173090475 on Build GRCh38
rs543520103 Chr.5: 173090573-173090573 on Build GRCh38
rs898294213 Chr.5: 173090654-173090654 on Build GRCh38
rs751590431 Chr.5: 173090356-173090356 on Build GRCh38
rs757723698 Chr.5: 173090614-173090614 on Build GRCh38
rs112345986 Chr.5: 173090402-173090402 on Build GRCh38
rs774526740 Chr.5: 173090482-173090482 on Build GRCh38
rs754098904 Chr.5: 173090602-173090602 on Build GRCh38
rs779456608 Chr.5: 173090637-173090637 on Build GRCh38
rs756700986 Chr.5: 173090375-173090375 on Build GRCh38
rs778725931 Chr.5: 173090424-173090424 on Build GRCh38
rs758193769 Chr.5: 173090647-173090647 on Build GRCh38
rs372973206 Chr.5: 173090504-173090504 on Build GRCh38
rs763786078 Chr.5: 173090317-173090317 on Build GRCh38
rs771101800 Chr.5: 173090476-173090476 on Build GRCh38
rs761020358 Chr.5: 173090557-173090557 on Build GRCh38
rs993451214 Chr.5: 173090262-173090262 on Build GRCh38
rs759805262 Chr.5: 173090513-173090513 on Build GRCh38
rs977613806 Chr.5: 173090207-173090207 on Build GRCh38
rs771675272 Chr.5: 173090432-173090432 on Build GRCh38
rs898812389 Chr.5: 173090258-173090258 on Build GRCh38
rs949667853 Chr.5: 173090447-173090447 on Build GRCh38
rs752886692 Chr.5: 173090551-173090551 on Build GRCh38
rs779367195 Chr.5: 173090649-173090649 on Build GRCh38
rs200282773 Chr.5: 173090365-173090365 on Build GRCh38
rs760703944 Chr.5: 173090305-173090305 on Build GRCh38
rs756912524 Chr.5: 173090422-173090422 on Build GRCh38
rs772082257 Chr.5: 173090284-173090284 on Build GRCh38
rs530973126 Chr.5: 173090200-173090200 on Build GRCh38
rs981140064 Chr.5: 173090518-173090518 on Build GRCh38
rs753124918 Chr.5: 173090371-173090371 on Build GRCh38
rs375287212 Chr.5: 173090418-173090418 on Build GRCh38
rs758743746 Chr.5: 173090295-173090295 on Build GRCh38
rs777994509 Chr.5: 173090393-173090393 on Build GRCh38
rs369014574 Chr.5: 173090463-173090463 on Build GRCh38
rs35768619 Chr.5: 173090650-173090650 on Build GRCh38
rs369730429 Chr.5: 173090282-173090282 on Build GRCh38
rs372264494 Chr.5: 173090353-173090353 on Build GRCh38
rs558188397 Chr.5: 173090345-173090345 on Build GRCh38
rs766605392 Chr.5: 173090352-173090352 on Build GRCh38
rs776559458 Chr.5: 173090318-173090318 on Build GRCh38
rs750076305 Chr.5: 173090639-173090639 on Build GRCh38
rs1037383745 Chr.5: 173090225-173090225 on Build GRCh38

View Full Product Details