Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173092167-173092667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBRF
Entrez Gene ID
153222
Gene Name
CREB3 regulatory factor
Gene Aliases
C5orf41, LRF
UniGene
Hs.484195
Gene Chromosome Location
Chr.5: 173056349-173139288 on Build GRCh38
Transcripts
NM_001168393.1 NM_001168394.1 NM_153607.2 XM_005265821.3 XM_006714822.3 XM_011534442.1

SNP Detail

SNP ID Chromosome Location
rs999255648 Chr.5: 173092256-173092256 on Build GRCh38
rs974475300 Chr.5: 173092272-173092272 on Build GRCh38
rs765243824 Chr.5: 173092314-173092314 on Build GRCh38
rs560965141 Chr.5: 173092212-173092212 on Build GRCh38
rs957857677 Chr.5: 173092321-173092321 on Build GRCh38
rs995530067 Chr.5: 173092421-173092421 on Build GRCh38
rs78216725 Chr.5: 173092237-173092237 on Build GRCh38
rs191750701 Chr.5: 173092262-173092262 on Build GRCh38
rs900907728 Chr.5: 173092244-173092244 on Build GRCh38
rs957314673 Chr.5: 173092511-173092511 on Build GRCh38
rs967296714 Chr.5: 173092544-173092544 on Build GRCh38
rs184005428 Chr.5: 173092266-173092266 on Build GRCh38
rs72816119 Chr.5: 173092605-173092605 on Build GRCh38
rs951684701 Chr.5: 173092211-173092211 on Build GRCh38
rs900813834 Chr.5: 173092576-173092576 on Build GRCh38
rs1045554256 Chr.5: 173092373-173092373 on Build GRCh38
rs928681797 Chr.5: 173092437-173092437 on Build GRCh38
rs887091265 Chr.5: 173092630-173092630 on Build GRCh38
rs964187956 Chr.5: 173092486-173092486 on Build GRCh38
rs995965617 Chr.5: 173092498-173092498 on Build GRCh38
rs751646116 Chr.5: 173092606-173092609 on Build GRCh38
rs1031186551 Chr.5: 173092309-173092309 on Build GRCh38
rs990505610 Chr.5: 173092294-173092294 on Build GRCh38
rs367884937 Chr.5: 173092512-173092512 on Build GRCh38
rs142546294 Chr.5: 173092209-173092209 on Build GRCh38
rs969331476 Chr.5: 173092410-173092410 on Build GRCh38
rs890071743 Chr.5: 173092215-173092215 on Build GRCh38
rs1019822245 Chr.5: 173092442-173092442 on Build GRCh38
rs887387704 Chr.5: 173092428-173092428 on Build GRCh38
rs571003806 Chr.5: 173092282-173092282 on Build GRCh38
rs572221189 Chr.5: 173092194-173092194 on Build GRCh38
rs1019768045 Chr.5: 173092238-173092238 on Build GRCh38
rs113347890 Chr.5: 173092539-173092539 on Build GRCh38
rs898526340 Chr.5: 173092416-173092416 on Build GRCh38
rs913156155 Chr.5: 173092299-173092299 on Build GRCh38
rs1047469393 Chr.5: 173092423-173092423 on Build GRCh38
rs1023548641 Chr.5: 173092377-173092378 on Build GRCh38
rs999602108 Chr.5: 173092601-173092601 on Build GRCh38
rs981307910 Chr.5: 173092549-173092549 on Build GRCh38
rs768887788 Chr.5: 173092562-173092562 on Build GRCh38
rs928511729 Chr.5: 173092569-173092569 on Build GRCh38
rs571722771 Chr.5: 173092430-173092430 on Build GRCh38
rs775437869 Chr.5: 173092384-173092384 on Build GRCh38
rs937451385 Chr.5: 173092399-173092399 on Build GRCh38
rs1047068697 Chr.5: 173092627-173092627 on Build GRCh38
rs931366815 Chr.5: 173092608-173092608 on Build GRCh38
rs543430599 Chr.5: 173092254-173092254 on Build GRCh38
rs148670646 Chr.5: 173092337-173092337 on Build GRCh38
rs991699328 Chr.5: 173092598-173092598 on Build GRCh38
rs527889691 Chr.5: 173092439-173092439 on Build GRCh38
rs764496640 Chr.5: 173092507-173092507 on Build GRCh38
rs928458022 Chr.5: 173092393-173092393 on Build GRCh38
rs1055882118 Chr.5: 173092412-173092412 on Build GRCh38
rs752159061 Chr.5: 173092527-173092527 on Build GRCh38
rs567413058 Chr.5: 173092597-173092597 on Build GRCh38
rs527328898 Chr.5: 173092532-173092532 on Build GRCh38
rs552069607 Chr.5: 173092401-173092401 on Build GRCh38
rs759776610 Chr.5: 173092275-173092275 on Build GRCh38
rs1006510345 Chr.5: 173092233-173092233 on Build GRCh38
rs796217410 Chr.5: 173092427-173092427 on Build GRCh38
rs770752017 Chr.5: 173092207-173092207 on Build GRCh38
rs1048585498 Chr.5: 173092538-173092538 on Build GRCh38
rs945927508 Chr.5: 173092354-173092354 on Build GRCh38
rs144281603 Chr.5: 173092597-173092597 on Build GRCh38

View Full Product Details