Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173108503-173109013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBRF
Entrez Gene ID
153222
Gene Name
CREB3 regulatory factor
Gene Aliases
C5orf41, LRF
UniGene
Hs.484195
Gene Chromosome Location
Chr.5: 173056349-173139288 on Build GRCh38
Transcripts
NM_153607.2 XM_005265821.3 XM_006714822.3 XM_011534442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
331451 c.1223-1G>T p.? Chr.5: 173108623-173108623 on Build GRCh38
6635377 c.1233T>C p.N411N Chr.5: 173108634-173108634 on Build GRCh38
449338 c.1306G>C p.E436Q Chr.5: 173108707-173108707 on Build GRCh38
1198887 c.1398G>C p.Q466H Chr.5: 173108799-173108799 on Build GRCh38
6635379 c.1276C>T p.R426W Chr.5: 173108677-173108677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774242116 Chr.5: 173108655-173108655 on Build GRCh38
rs535367225 Chr.5: 173108726-173108726 on Build GRCh38
rs768259269 Chr.5: 173108589-173108589 on Build GRCh38
rs914178966 Chr.5: 173108950-173108950 on Build GRCh38
rs574369785 Chr.5: 173108711-173108711 on Build GRCh38
rs951340334 Chr.5: 173108893-173108893 on Build GRCh38
rs397882503 Chr.5: 173108553-173108554 on Build GRCh38
rs892118914 Chr.5: 173108712-173108712 on Build GRCh38
rs1016765972 Chr.5: 173108829-173108829 on Build GRCh38
rs561292355 Chr.5: 173108984-173108984 on Build GRCh38
rs763177631 Chr.5: 173108931-173108931 on Build GRCh38
rs895540974 Chr.5: 173108748-173108748 on Build GRCh38
rs142630557 Chr.5: 173108770-173108770 on Build GRCh38
rs759680931 Chr.5: 173108861-173108861 on Build GRCh38
rs985464202 Chr.5: 173108581-173108581 on Build GRCh38
rs373863828 Chr.5: 173108771-173108771 on Build GRCh38
rs911223255 Chr.5: 173108919-173108919 on Build GRCh38
rs766300347 Chr.5: 173108583-173108584 on Build GRCh38
rs757553134 Chr.5: 173108695-173108695 on Build GRCh38
rs924160082 Chr.5: 173108941-173108941 on Build GRCh38
rs1054916516 Chr.5: 173108987-173108987 on Build GRCh38
rs765227967 Chr.5: 173108700-173108700 on Build GRCh38
rs530913131 Chr.5: 173108869-173108869 on Build GRCh38
rs961210085 Chr.5: 173108851-173108851 on Build GRCh38
rs1026268127 Chr.5: 173108883-173108883 on Build GRCh38
rs946986410 Chr.5: 173108639-173108639 on Build GRCh38
rs747386867 Chr.5: 173108580-173108580 on Build GRCh38
rs576376433 Chr.5: 173108878-173108878 on Build GRCh38
rs186077834 Chr.5: 173108915-173108915 on Build GRCh38
rs753297387 Chr.5: 173108895-173108895 on Build GRCh38
rs945032065 Chr.5: 173108959-173108959 on Build GRCh38
rs565758536 Chr.5: 173108532-173108532 on Build GRCh38
rs769495244 Chr.5: 173108805-173108805 on Build GRCh38
rs767383054 Chr.5: 173108670-173108670 on Build GRCh38
rs758844507 Chr.5: 173108718-173108718 on Build GRCh38
rs962765978 Chr.5: 173108916-173108916 on Build GRCh38
rs368351952 Chr.5: 173108840-173108840 on Build GRCh38
rs911486088 Chr.5: 173108627-173108627 on Build GRCh38
rs747557168 Chr.5: 173108763-173108763 on Build GRCh38
rs935447459 Chr.5: 173108675-173108675 on Build GRCh38
rs758971295 Chr.5: 173108573-173108573 on Build GRCh38
rs987457568 Chr.5: 173108942-173108942 on Build GRCh38
rs921462865 Chr.5: 173108610-173108610 on Build GRCh38
rs201664801 Chr.5: 173108677-173108677 on Build GRCh38
rs753905877 Chr.5: 173108614-173108614 on Build GRCh38
rs780327846 Chr.5: 173108579-173108579 on Build GRCh38
rs776118512 Chr.5: 173108590-173108590 on Build GRCh38
rs541191843 Chr.5: 173108966-173108966 on Build GRCh38
rs192021908 Chr.5: 173108607-173108607 on Build GRCh38
rs1056627269 Chr.5: 173108971-173108971 on Build GRCh38
rs770949667 Chr.5: 173108847-173108847 on Build GRCh38
rs747755461 Chr.5: 173108791-173108791 on Build GRCh38
rs116460413 Chr.5: 173108932-173108932 on Build GRCh38
rs905816560 Chr.5: 173108774-173108774 on Build GRCh38
rs1016960934 Chr.5: 173108913-173108913 on Build GRCh38
rs180840280 Chr.5: 173108767-173108767 on Build GRCh38
rs982808616 Chr.5: 173108905-173108905 on Build GRCh38
rs937727914 Chr.5: 173108986-173108986 on Build GRCh38
rs1013910649 Chr.5: 173108879-173108879 on Build GRCh38
rs35953058 Chr.5: 173108551-173108552 on Build GRCh38
rs780571169 Chr.5: 173108758-173108758 on Build GRCh38
rs1005473654 Chr.5: 173108802-173108802 on Build GRCh38
rs777363785 Chr.5: 173108826-173108826 on Build GRCh38
rs909989651 Chr.5: 173108578-173108578 on Build GRCh38
rs896304965 Chr.5: 173108988-173108988 on Build GRCh38
rs769174659 Chr.5: 173108596-173108596 on Build GRCh38
rs202157525 Chr.5: 173108738-173108738 on Build GRCh38
rs766434353 Chr.5: 173108608-173108608 on Build GRCh38
rs147734847 Chr.5: 173108668-173108668 on Build GRCh38
rs527752705 Chr.5: 173108965-173108965 on Build GRCh38

View Full Product Details