Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 175878694-175879199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPLX2
Entrez Gene ID
10814
Gene Name
complexin 2
Gene Aliases
921-L, CPX-2, CPX2, Hfb1
UniGene
Hs.193235
Gene Chromosome Location
Chr.5: 175796539-175884021 on Build GRCh38
Transcripts
NM_001008220.1 NM_006650.3 XM_005265798.1 XM_005265799.1 XM_011534419.1 XM_017008964.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4520682 c.106G>A p.E36K Chr.5: 175878982-175878982 on Build GRCh38
362803 c.73G>A p.E25K Chr.5: 175878949-175878949 on Build GRCh38
4803572 c.141G>T p.E47D Chr.5: 175879017-175879017 on Build GRCh38
482534 c.143G>C p.R48P Chr.5: 175879019-175879019 on Build GRCh38
4543551 c.33G>A p.G11G Chr.5: 175878909-175878909 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554204367 Chr.5: 175878903-175878903 on Build GRCh38
rs989448433 Chr.5: 175878829-175878829 on Build GRCh38
rs139443061 Chr.5: 175879139-175879139 on Build GRCh38
rs745518580 Chr.5: 175879122-175879122 on Build GRCh38
rs200872237 Chr.5: 175878875-175878875 on Build GRCh38
rs969198823 Chr.5: 175878840-175878840 on Build GRCh38
rs560682117 Chr.5: 175879141-175879141 on Build GRCh38
rs956830362 Chr.5: 175878884-175878884 on Build GRCh38
rs755926994 Chr.5: 175878858-175878858 on Build GRCh38
rs549555179 Chr.5: 175878728-175878728 on Build GRCh38
rs746752549 Chr.5: 175879090-175879091 on Build GRCh38
rs768638902 Chr.5: 175878992-175878992 on Build GRCh38
rs777613263 Chr.5: 175879091-175879094 on Build GRCh38
rs148443914 Chr.5: 175878981-175878981 on Build GRCh38
rs527762149 Chr.5: 175879145-175879145 on Build GRCh38
rs766571484 Chr.5: 175879050-175879050 on Build GRCh38
rs745898022 Chr.5: 175878737-175878737 on Build GRCh38
rs146284357 Chr.5: 175878960-175878960 on Build GRCh38
rs530072651 Chr.5: 175879138-175879138 on Build GRCh38
rs753126962 Chr.5: 175878823-175878823 on Build GRCh38
rs188817237 Chr.5: 175878948-175878948 on Build GRCh38
rs777197988 Chr.5: 175878790-175878790 on Build GRCh38
rs773985589 Chr.5: 175879121-175879121 on Build GRCh38
rs980478393 Chr.5: 175878938-175878938 on Build GRCh38
rs556552689 Chr.5: 175878809-175878809 on Build GRCh38
rs544890678 Chr.5: 175879100-175879100 on Build GRCh38
rs574866432 Chr.5: 175878850-175878850 on Build GRCh38
rs374027096 Chr.5: 175878734-175878734 on Build GRCh38
rs763871164 Chr.5: 175879034-175879034 on Build GRCh38
rs377227199 Chr.5: 175878791-175878791 on Build GRCh38
rs748664347 Chr.5: 175879093-175879093 on Build GRCh38
rs767394411 Chr.5: 175878931-175878931 on Build GRCh38
rs777111870 Chr.5: 175878882-175878882 on Build GRCh38
rs577502502 Chr.5: 175879097-175879097 on Build GRCh38
rs775793619 Chr.5: 175878748-175878748 on Build GRCh38
rs759047856 Chr.5: 175879010-175879010 on Build GRCh38
rs771232967 Chr.5: 175879107-175879107 on Build GRCh38
rs752539534 Chr.5: 175878820-175878820 on Build GRCh38
rs1040791450 Chr.5: 175879126-175879126 on Build GRCh38
rs752660307 Chr.5: 175878945-175878945 on Build GRCh38
rs901801802 Chr.5: 175878806-175878806 on Build GRCh38
rs746093286 Chr.5: 175878868-175878868 on Build GRCh38
rs748682267 Chr.5: 175878994-175878994 on Build GRCh38
rs750597984 Chr.5: 175878949-175878949 on Build GRCh38
rs943457446 Chr.5: 175879092-175879092 on Build GRCh38
rs751442006 Chr.5: 175878808-175878808 on Build GRCh38
rs781483059 Chr.5: 175879074-175879074 on Build GRCh38
rs934716553 Chr.5: 175879123-175879123 on Build GRCh38
rs781099861 Chr.5: 175878723-175878723 on Build GRCh38
rs113857060 Chr.5: 175879084-175879084 on Build GRCh38
rs780237373 Chr.5: 175878873-175878873 on Build GRCh38
rs199844563 Chr.5: 175878996-175878996 on Build GRCh38
rs565316889 Chr.5: 175879036-175879036 on Build GRCh38
rs758033687 Chr.5: 175878991-175878992 on Build GRCh38
rs762216901 Chr.5: 175878886-175878886 on Build GRCh38
rs1035106998 Chr.5: 175878954-175878954 on Build GRCh38
rs1030691756 Chr.5: 175878814-175878814 on Build GRCh38
rs142629775 Chr.5: 175879051-175879051 on Build GRCh38
rs781738443 Chr.5: 175878993-175878993 on Build GRCh38
rs1019382650 Chr.5: 175878800-175878800 on Build GRCh38
rs750133917 Chr.5: 175878804-175878804 on Build GRCh38
rs984935215 Chr.5: 175879020-175879020 on Build GRCh38
rs747742596 Chr.5: 175878943-175878945 on Build GRCh38
rs774394266 Chr.5: 175878736-175878736 on Build GRCh38
rs759371540 Chr.5: 175878913-175878913 on Build GRCh38
rs930881682 Chr.5: 175879007-175879007 on Build GRCh38
rs771987308 Chr.5: 175878870-175878870 on Build GRCh38
rs761257195 Chr.5: 175878764-175878764 on Build GRCh38
rs749507747 Chr.5: 175878732-175878732 on Build GRCh38
rs751120600 Chr.5: 175878909-175878909 on Build GRCh38
rs767016364 Chr.5: 175879023-175879023 on Build GRCh38
rs780934706 Chr.5: 175878818-175878818 on Build GRCh38
rs751759019 Chr.5: 175879053-175879053 on Build GRCh38
rs868466959 Chr.5: 175879019-175879019 on Build GRCh38
rs4077873 Chr.5: 175879104-175879104 on Build GRCh38
rs920636396 Chr.5: 175878952-175878952 on Build GRCh38
rs772222736 Chr.5: 175878740-175878740 on Build GRCh38
rs778540345 Chr.5: 175878815-175878815 on Build GRCh38
rs770273285 Chr.5: 175878995-175878995 on Build GRCh38
rs376386630 Chr.5: 175879071-175879071 on Build GRCh38
rs370297557 Chr.5: 175878716-175878716 on Build GRCh38
rs755838434 Chr.5: 175878946-175878946 on Build GRCh38
rs1010806285 Chr.5: 175879149-175879149 on Build GRCh38
rs1022140467 Chr.5: 175879162-175879162 on Build GRCh38
rs760662043 Chr.5: 175879033-175879033 on Build GRCh38
rs770667199 Chr.5: 175879000-175879000 on Build GRCh38
rs1051018835 Chr.5: 175878718-175878718 on Build GRCh38
rs773899228 Chr.5: 175879002-175879002 on Build GRCh38
rs373302730 Chr.5: 175878891-175878891 on Build GRCh38
rs765028650 Chr.5: 175878796-175878796 on Build GRCh38
rs779065649 Chr.5: 175878866-175878866 on Build GRCh38
rs1023889773 Chr.5: 175878836-175878836 on Build GRCh38
rs775347080 Chr.5: 175879032-175879032 on Build GRCh38
rs764770402 Chr.5: 175878805-175878805 on Build GRCh38
rs754675476 Chr.5: 175878721-175878721 on Build GRCh38
rs538540266 Chr.5: 175878780-175878780 on Build GRCh38
rs763541194 Chr.5: 175878879-175878879 on Build GRCh38

View Full Product Details